Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118193649-118194160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAML
Entrez Gene ID
120425
Gene Name
junction adhesion molecule like
Gene Aliases
AMICA, AMICA1, CREA7-1, CREA7-4, Gm638
UniGene
Hs.16291
Gene Chromosome Location
Chr.11: 118193727-118225094 on Build GRCh38
Transcripts
NM_001098526.1 NM_001286570.1 NM_001286571.1 NM_153206.2 NR_104479.1 XM_005271407.3 XM_017017213.1 XM_017017214.1 XM_017017215.1

SNP Detail

SNP ID Chromosome Location
rs760788284 Chr.11: 118193689-118193689 on Build GRCh38
rs780492637 Chr.11: 118193688-118193688 on Build GRCh38
rs923118560 Chr.11: 118193919-118193919 on Build GRCh38
rs367714463 Chr.11: 118193691-118193691 on Build GRCh38
rs1005430208 Chr.11: 118194049-118194049 on Build GRCh38
rs555224980 Chr.11: 118193983-118193983 on Build GRCh38
rs969547259 Chr.11: 118194047-118194047 on Build GRCh38
rs1027104277 Chr.11: 118194053-118194053 on Build GRCh38
rs983179343 Chr.11: 118193868-118193868 on Build GRCh38
rs73593809 Chr.11: 118194031-118194031 on Build GRCh38
rs982825355 Chr.11: 118194092-118194092 on Build GRCh38
rs980870066 Chr.11: 118193827-118193827 on Build GRCh38
rs619250 Chr.11: 118193867-118193867 on Build GRCh38
rs546590388 Chr.11: 118193882-118193882 on Build GRCh38
rs1028663388 Chr.11: 118193807-118193807 on Build GRCh38
rs144289711 Chr.11: 118193685-118193685 on Build GRCh38
rs561652480 Chr.11: 118193837-118193837 on Build GRCh38
rs931779360 Chr.11: 118193920-118193920 on Build GRCh38
rs182260809 Chr.11: 118193984-118193984 on Build GRCh38
rs764034034 Chr.11: 118193862-118193862 on Build GRCh38
rs1005118294 Chr.11: 118193730-118193730 on Build GRCh38
rs1025207779 Chr.11: 118194041-118194041 on Build GRCh38
rs952904172 Chr.11: 118194080-118194080 on Build GRCh38
rs903534252 Chr.11: 118193717-118193717 on Build GRCh38
rs553844327 Chr.11: 118194076-118194076 on Build GRCh38
rs769216920 Chr.11: 118193901-118193901 on Build GRCh38
rs1052514185 Chr.11: 118194020-118194020 on Build GRCh38
rs577671073 Chr.11: 118194046-118194046 on Build GRCh38
rs886640038 Chr.11: 118193716-118193716 on Build GRCh38
rs965376543 Chr.11: 118194137-118194137 on Build GRCh38
rs981315718 Chr.11: 118193902-118193902 on Build GRCh38
rs957175524 Chr.11: 118193767-118193767 on Build GRCh38
rs1002338839 Chr.11: 118194048-118194048 on Build GRCh38
rs528841125 Chr.11: 118193687-118193687 on Build GRCh38
rs957054081 Chr.11: 118194118-118194118 on Build GRCh38
rs113175724 Chr.11: 118193816-118193816 on Build GRCh38
rs145050873 Chr.11: 118194132-118194132 on Build GRCh38
rs1019592851 Chr.11: 118193759-118193759 on Build GRCh38
rs139661518 Chr.11: 118193985-118193985 on Build GRCh38
rs1000906521 Chr.11: 118193743-118193743 on Build GRCh38
rs1050144324 Chr.11: 118193939-118193939 on Build GRCh38
rs535399002 Chr.11: 118193960-118193960 on Build GRCh38
rs966330529 Chr.11: 118193764-118193764 on Build GRCh38
rs3741316 Chr.11: 118193815-118193815 on Build GRCh38
rs907824056 Chr.11: 118193880-118193880 on Build GRCh38
rs562342350 Chr.11: 118193755-118193755 on Build GRCh38
rs1014267078 Chr.11: 118194039-118194039 on Build GRCh38
rs945892704 Chr.11: 118193887-118193887 on Build GRCh38
rs529195689 Chr.11: 118193874-118193874 on Build GRCh38
rs540327786 Chr.11: 118194050-118194050 on Build GRCh38
rs77588719 Chr.11: 118193917-118193917 on Build GRCh38
rs902050953 Chr.11: 118194052-118194052 on Build GRCh38
rs992363727 Chr.11: 118193804-118193804 on Build GRCh38

View Full Product Details