Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118340297-118340806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD3D
Entrez Gene ID
915
Gene Name
CD3d molecule
Gene Aliases
CD3-DELTA, IMD19, T3D
UniGene
Hs.504048
Gene Chromosome Location
Chr.11: 118339074-118344233 on Build GRCh38
Transcripts
NM_000732.4 NM_001040651.1 XM_017018543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7279258 c.56-10delT p.? Chr.11: 118340603-118340603 on Build GRCh38
4427541 c.126A>T p.T42T Chr.11: 118340523-118340523 on Build GRCh38
4018267 c.207A>T p.G69G Chr.11: 118340442-118340442 on Build GRCh38
4534270 c.206G>A p.G69E Chr.11: 118340443-118340443 on Build GRCh38
4563132 c.95G>A p.R32K Chr.11: 118340554-118340554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529884894 Chr.11: 118340509-118340509 on Build GRCh38
rs372979468 Chr.11: 118340426-118340426 on Build GRCh38
rs730880296 Chr.11: 118340370-118340370 on Build GRCh38
rs751973236 Chr.11: 118340641-118340641 on Build GRCh38
rs529268621 Chr.11: 118340589-118340589 on Build GRCh38
rs144504840 Chr.11: 118340568-118340568 on Build GRCh38
rs531304018 Chr.11: 118340681-118340681 on Build GRCh38
rs746752914 Chr.11: 118340542-118340542 on Build GRCh38
rs1040989731 Chr.11: 118340620-118340620 on Build GRCh38
rs202077439 Chr.11: 118340610-118340610 on Build GRCh38
rs761840740 Chr.11: 118340581-118340581 on Build GRCh38
rs781625373 Chr.11: 118340507-118340507 on Build GRCh38
rs775386500 Chr.11: 118340538-118340538 on Build GRCh38
rs768551495 Chr.11: 118340553-118340553 on Build GRCh38
rs201374139 Chr.11: 118340573-118340573 on Build GRCh38
rs752718909 Chr.11: 118340354-118340354 on Build GRCh38
rs201299420 Chr.11: 118340371-118340371 on Build GRCh38
rs758839719 Chr.11: 118340377-118340377 on Build GRCh38
rs111033580 Chr.11: 118340447-118340447 on Build GRCh38
rs201889742 Chr.11: 118340402-118340402 on Build GRCh38
rs777709761 Chr.11: 118340524-118340524 on Build GRCh38
rs932612783 Chr.11: 118340688-118340688 on Build GRCh38
rs763077739 Chr.11: 118340412-118340414 on Build GRCh38
rs959296545 Chr.11: 118340708-118340708 on Build GRCh38
rs866317904 Chr.11: 118340468-118340468 on Build GRCh38
rs747613953 Chr.11: 118340389-118340389 on Build GRCh38
rs960253005 Chr.11: 118340428-118340428 on Build GRCh38
rs200037447 Chr.11: 118340497-118340497 on Build GRCh38
rs762031414 Chr.11: 118340329-118340329 on Build GRCh38
rs973204628 Chr.11: 118340715-118340715 on Build GRCh38
rs201177720 Chr.11: 118340608-118340608 on Build GRCh38
rs763754862 Chr.11: 118340359-118340359 on Build GRCh38
rs750023092 Chr.11: 118340372-118340372 on Build GRCh38
rs751800253 Chr.11: 118340493-118340493 on Build GRCh38
rs201422803 Chr.11: 118340482-118340482 on Build GRCh38
rs886047737 Chr.11: 118340528-118340528 on Build GRCh38
rs929117843 Chr.11: 118340572-118340572 on Build GRCh38
rs771969049 Chr.11: 118340424-118340424 on Build GRCh38
rs760666412 Chr.11: 118340440-118340440 on Build GRCh38
rs757787812 Chr.11: 118340638-118340638 on Build GRCh38
rs944042008 Chr.11: 118340614-118340614 on Build GRCh38
rs111841831 Chr.11: 118340379-118340379 on Build GRCh38
rs1003220858 Chr.11: 118340698-118340698 on Build GRCh38
rs563103670 Chr.11: 118340585-118340585 on Build GRCh38
rs199795476 Chr.11: 118340446-118340446 on Build GRCh38
rs200260087 Chr.11: 118340387-118340387 on Build GRCh38
rs199969175 Chr.11: 118340582-118340582 on Build GRCh38
rs959121453 Chr.11: 118340534-118340534 on Build GRCh38
rs370657743 Chr.11: 118340580-118340580 on Build GRCh38
rs766581002 Chr.11: 118340482-118340482 on Build GRCh38
rs763052940 Chr.11: 118340418-118340418 on Build GRCh38
rs1033387464 Chr.11: 118340705-118340706 on Build GRCh38
rs370928131 Chr.11: 118340351-118340351 on Build GRCh38
rs771000136 Chr.11: 118340536-118340536 on Build GRCh38
rs199695944 Chr.11: 118340349-118340349 on Build GRCh38
rs142464005 Chr.11: 118340659-118340659 on Build GRCh38
rs199636212 Chr.11: 118340383-118340383 on Build GRCh38
rs764812030 Chr.11: 118340459-118340459 on Build GRCh38
rs200409086 Chr.11: 118340363-118340363 on Build GRCh38
rs1009711254 Chr.11: 118340685-118340685 on Build GRCh38
rs542784456 Chr.11: 118340355-118340355 on Build GRCh38
rs775134685 Chr.11: 118340427-118340427 on Build GRCh38
rs572315717 Chr.11: 118340393-118340393 on Build GRCh38
rs753332565 Chr.11: 118340360-118340360 on Build GRCh38
rs377380944 Chr.11: 118340340-118340340 on Build GRCh38
rs761459276 Chr.11: 118340448-118340448 on Build GRCh38
rs759324030 Chr.11: 118340352-118340352 on Build GRCh38
rs906744452 Chr.11: 118340690-118340690 on Build GRCh38
rs748182805 Chr.11: 118340414-118340414 on Build GRCh38
rs193284900 Chr.11: 118340535-118340535 on Build GRCh38
rs762667054 Chr.11: 118340627-118340627 on Build GRCh38
rs911784494 Chr.11: 118340611-118340611 on Build GRCh38
rs561302204 Chr.11: 118340380-118340380 on Build GRCh38
rs377725940 Chr.11: 118340438-118340438 on Build GRCh38
rs773529981 Chr.11: 118340417-118340417 on Build GRCh38
rs201021372 Chr.11: 118340508-118340508 on Build GRCh38
rs528486045 Chr.11: 118340416-118340416 on Build GRCh38
rs1027786730 Chr.11: 118340717-118340717 on Build GRCh38
rs1043042909 Chr.11: 118340752-118340752 on Build GRCh38
rs200462658 Chr.11: 118340471-118340471 on Build GRCh38
rs904593972 Chr.11: 118340775-118340775 on Build GRCh38
rs879255345 Chr.11: 118340587-118340587 on Build GRCh38
rs778408963 Chr.11: 118340467-118340467 on Build GRCh38
rs2298822 Chr.11: 118340719-118340719 on Build GRCh38
rs998941574 Chr.11: 118340398-118340398 on Build GRCh38
rs374700153 Chr.11: 118340461-118340461 on Build GRCh38
rs201341699 Chr.11: 118340341-118340341 on Build GRCh38
rs1023255562 Chr.11: 118340488-118340488 on Build GRCh38
rs149264725 Chr.11: 118340462-118340462 on Build GRCh38
rs779383299 Chr.11: 118340658-118340658 on Build GRCh38
rs200910273 Chr.11: 118340394-118340394 on Build GRCh38

View Full Product Details