Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118754622-118755119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX6
Entrez Gene ID
1656
Gene Name
DEAD-box helicase 6
Gene Aliases
HLR2, P54, RCK
UniGene
Hs.408461
Gene Chromosome Location
Chr.11: 118747763-118791263 on Build GRCh38
Transcripts
NM_001257191.2 NM_004397.5 XM_005271417.3 XM_011542644.1 XM_011542645.1 XM_017017251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5019931 c.1244-11_1244-10delAT p.? Chr.11: 118754897-118754898 on Build GRCh38
7109333 c.1398G>C p.K466N Chr.11: 118754766-118754766 on Build GRCh38
3791229 c.1328G>A p.G443E Chr.11: 118754803-118754803 on Build GRCh38
5341417 c.1314C>T p.I438I Chr.11: 118754850-118754850 on Build GRCh38
5050565 c.1421A>G p.H474R Chr.11: 118754743-118754743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017559977 Chr.11: 118755065-118755065 on Build GRCh38
rs782750709 Chr.11: 118754844-118754844 on Build GRCh38
rs782285756 Chr.11: 118754906-118754906 on Build GRCh38
rs577106072 Chr.11: 118754826-118754826 on Build GRCh38
rs73001476 Chr.11: 118754960-118754960 on Build GRCh38
rs564281117 Chr.11: 118754985-118754985 on Build GRCh38
rs538001978 Chr.11: 118754894-118754894 on Build GRCh38
rs148749948 Chr.11: 118755019-118755021 on Build GRCh38
rs28572245 Chr.11: 118755055-118755055 on Build GRCh38
rs781846171 Chr.11: 118754711-118754711 on Build GRCh38
rs187620853 Chr.11: 118755020-118755020 on Build GRCh38
rs782176813 Chr.11: 118754914-118754914 on Build GRCh38
rs781965648 Chr.11: 118754906-118754907 on Build GRCh38
rs1057340150 Chr.11: 118754934-118754934 on Build GRCh38
rs890168309 Chr.11: 118755047-118755047 on Build GRCh38
rs73001477 Chr.11: 118754976-118754976 on Build GRCh38
rs782182181 Chr.11: 118754738-118754738 on Build GRCh38
rs782808001 Chr.11: 118754691-118754691 on Build GRCh38
rs782449121 Chr.11: 118754706-118754706 on Build GRCh38
rs782037097 Chr.11: 118754926-118754926 on Build GRCh38
rs993146216 Chr.11: 118755002-118755002 on Build GRCh38
rs782570283 Chr.11: 118754909-118754909 on Build GRCh38
rs561752798 Chr.11: 118754739-118754739 on Build GRCh38
rs782366111 Chr.11: 118755038-118755038 on Build GRCh38
rs1008797906 Chr.11: 118755054-118755054 on Build GRCh38
rs781873721 Chr.11: 118754853-118754853 on Build GRCh38
rs782097112 Chr.11: 118754678-118754678 on Build GRCh38
rs782707531 Chr.11: 118754821-118754821 on Build GRCh38
rs781919086 Chr.11: 118754668-118754668 on Build GRCh38
rs782403936 Chr.11: 118754908-118754908 on Build GRCh38
rs782138863 Chr.11: 118754919-118754919 on Build GRCh38
rs560524166 Chr.11: 118754931-118754931 on Build GRCh38
rs782365964 Chr.11: 118754781-118754781 on Build GRCh38
rs782258493 Chr.11: 118755028-118755028 on Build GRCh38
rs916067848 Chr.11: 118754937-118754937 on Build GRCh38
rs964510323 Chr.11: 118755075-118755075 on Build GRCh38
rs781893015 Chr.11: 118754895-118754895 on Build GRCh38
rs367848342 Chr.11: 118754929-118754929 on Build GRCh38
rs200460781 Chr.11: 118754708-118754708 on Build GRCh38
rs782373276 Chr.11: 118754921-118754921 on Build GRCh38
rs554519911 Chr.11: 118754656-118754656 on Build GRCh38
rs782698158 Chr.11: 118754707-118754707 on Build GRCh38
rs374533846 Chr.11: 118755061-118755061 on Build GRCh38
rs75327037 Chr.11: 118754836-118754836 on Build GRCh38
rs1037432799 Chr.11: 118754965-118754965 on Build GRCh38
rs782113918 Chr.11: 118754686-118754686 on Build GRCh38
rs75011565 Chr.11: 118754679-118754679 on Build GRCh38
rs782256533 Chr.11: 118754751-118754751 on Build GRCh38
rs782367731 Chr.11: 118754915-118754915 on Build GRCh38
rs782244366 Chr.11: 118754724-118754724 on Build GRCh38
rs191902541 Chr.11: 118754737-118754737 on Build GRCh38
rs565883115 Chr.11: 118754898-118754898 on Build GRCh38
rs574758152 Chr.11: 118754899-118754899 on Build GRCh38
rs781928315 Chr.11: 118754823-118754823 on Build GRCh38
rs782554652 Chr.11: 118754733-118754733 on Build GRCh38
rs78525463 Chr.11: 118754718-118754718 on Build GRCh38
rs556399553 Chr.11: 118754897-118754897 on Build GRCh38
rs782028733 Chr.11: 118754811-118754811 on Build GRCh38
rs782468835 Chr.11: 118754717-118754717 on Build GRCh38
rs782662981 Chr.11: 118754722-118754722 on Build GRCh38
rs533868092 Chr.11: 118754701-118754701 on Build GRCh38
rs781963730 Chr.11: 118754916-118754916 on Build GRCh38
rs580079 Chr.11: 118754900-118754900 on Build GRCh38
rs374184056 Chr.11: 118754819-118754819 on Build GRCh38

View Full Product Details