Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118756867-118757370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX6
Entrez Gene ID
1656
Gene Name
DEAD-box helicase 6
Gene Aliases
HLR2, P54, RCK
UniGene
Hs.408461
Gene Chromosome Location
Chr.11: 118747763-118791263 on Build GRCh38
Transcripts
NM_001257191.2 NM_004397.5 XM_005271417.3 XM_011542644.1 XM_011542645.1 XM_017017251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3808510 c.1071G>C p.M357I Chr.11: 118757177-118757177 on Build GRCh38
4942127 c.1007A>T p.Q336L Chr.11: 118757274-118757274 on Build GRCh38
5656641 c.1066C>T p.Q356* Chr.11: 118757215-118757215 on Build GRCh38
4972134 c.1025_1027delAGA p.K342delK Chr.11: 118757221-118757223 on Build GRCh38
1351707 c.1004G>A p.R335Q Chr.11: 118757244-118757244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004900981 Chr.11: 118756934-118756934 on Build GRCh38
rs531102422 Chr.11: 118757320-118757321 on Build GRCh38
rs537085403 Chr.11: 118757119-118757119 on Build GRCh38
rs782034629 Chr.11: 118757271-118757271 on Build GRCh38
rs782579204 Chr.11: 118757236-118757236 on Build GRCh38
rs782100894 Chr.11: 118757304-118757304 on Build GRCh38
rs782701187 Chr.11: 118757142-118757142 on Build GRCh38
rs782624148 Chr.11: 118757180-118757180 on Build GRCh38
rs981137067 Chr.11: 118757327-118757327 on Build GRCh38
rs1024815791 Chr.11: 118757146-118757146 on Build GRCh38
rs781948897 Chr.11: 118757255-118757255 on Build GRCh38
rs541533194 Chr.11: 118757237-118757237 on Build GRCh38
rs199834623 Chr.11: 118757276-118757276 on Build GRCh38
rs10892282 Chr.11: 118757069-118757069 on Build GRCh38
rs12281298 Chr.11: 118756992-118756992 on Build GRCh38
rs569709892 Chr.11: 118757111-118757111 on Build GRCh38
rs574025116 Chr.11: 118757153-118757153 on Build GRCh38
rs59323635 Chr.11: 118757125-118757128 on Build GRCh38
rs781836864 Chr.11: 118757148-118757148 on Build GRCh38
rs781852750 Chr.11: 118757335-118757335 on Build GRCh38
rs782286418 Chr.11: 118757183-118757183 on Build GRCh38
rs782534467 Chr.11: 118757213-118757213 on Build GRCh38
rs555739141 Chr.11: 118757138-118757138 on Build GRCh38
rs782263608 Chr.11: 118757267-118757267 on Build GRCh38
rs782195996 Chr.11: 118757068-118757068 on Build GRCh38
rs78749417 Chr.11: 118757298-118757298 on Build GRCh38
rs782716033 Chr.11: 118757285-118757285 on Build GRCh38
rs1000605637 Chr.11: 118757073-118757073 on Build GRCh38
rs761691321 Chr.11: 118757190-118757190 on Build GRCh38
rs782386385 Chr.11: 118757270-118757270 on Build GRCh38
rs138622541 Chr.11: 118756984-118756984 on Build GRCh38
rs782353147 Chr.11: 118757246-118757246 on Build GRCh38
rs781890493 Chr.11: 118757127-118757127 on Build GRCh38
rs1040610371 Chr.11: 118756982-118756982 on Build GRCh38
rs956348598 Chr.11: 118757125-118757125 on Build GRCh38
rs782665610 Chr.11: 118757008-118757008 on Build GRCh38
rs782529877 Chr.11: 118757149-118757149 on Build GRCh38
rs781923634 Chr.11: 118757290-118757290 on Build GRCh38
rs782730134 Chr.11: 118757328-118757328 on Build GRCh38

View Full Product Details