Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118893708-118894214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCR5
Entrez Gene ID
643
Gene Name
C-X-C motif chemokine receptor 5
Gene Aliases
BLR1, CD185, MDR15
UniGene
Hs.113916
Gene Chromosome Location
Chr.11: 118883766-118896271 on Build GRCh38
Transcripts
NM_001716.4 NM_032966.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
220495 c.665A>C p.Y222S Chr.11: 118894209-118894209 on Build GRCh38
4651873 c.165C>T p.P55P Chr.11: 118893709-118893709 on Build GRCh38
6288236 c.429C>T p.I143I Chr.11: 118893973-118893973 on Build GRCh38
4603058 c.387_388insA p.V131fs*178 Chr.11: 118893931-118893932 on Build GRCh38
4566981 c.528_529GG>AA p.G177S Chr.11: 118894072-118894073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953223801 Chr.11: 118893976-118893976 on Build GRCh38
rs759344464 Chr.11: 118893843-118893843 on Build GRCh38
rs200959025 Chr.11: 118894145-118894145 on Build GRCh38
rs371969699 Chr.11: 118893810-118893810 on Build GRCh38
rs955155953 Chr.11: 118893782-118893782 on Build GRCh38
rs768733962 Chr.11: 118893824-118893824 on Build GRCh38
rs781540268 Chr.11: 118893734-118893734 on Build GRCh38
rs752785973 Chr.11: 118894056-118894056 on Build GRCh38
rs144978847 Chr.11: 118894036-118894036 on Build GRCh38
rs768666087 Chr.11: 118894009-118894009 on Build GRCh38
rs140371886 Chr.11: 118893944-118893944 on Build GRCh38
rs531161857 Chr.11: 118894071-118894071 on Build GRCh38
rs759889292 Chr.11: 118894165-118894165 on Build GRCh38
rs138622461 Chr.11: 118894124-118894124 on Build GRCh38
rs770005238 Chr.11: 118893832-118893832 on Build GRCh38
rs372433399 Chr.11: 118894006-118894006 on Build GRCh38
rs774062762 Chr.11: 118893841-118893841 on Build GRCh38
rs768071139 Chr.11: 118894171-118894171 on Build GRCh38
rs747249854 Chr.11: 118893803-118893804 on Build GRCh38
rs762687395 Chr.11: 118894146-118894146 on Build GRCh38
rs1055244370 Chr.11: 118893922-118893922 on Build GRCh38
rs756510807 Chr.11: 118894179-118894179 on Build GRCh38
rs112901555 Chr.11: 118893752-118893752 on Build GRCh38
rs755680507 Chr.11: 118893783-118893783 on Build GRCh38
rs763896307 Chr.11: 118893786-118893786 on Build GRCh38
rs748559889 Chr.11: 118893801-118893801 on Build GRCh38
rs753260293 Chr.11: 118894172-118894172 on Build GRCh38
rs149961413 Chr.11: 118894034-118894034 on Build GRCh38
rs749530988 Chr.11: 118894078-118894078 on Build GRCh38
rs200335551 Chr.11: 118893805-118893805 on Build GRCh38
rs149086316 Chr.11: 118894042-118894042 on Build GRCh38
rs773283155 Chr.11: 118893756-118893756 on Build GRCh38
rs373196739 Chr.11: 118894004-118894004 on Build GRCh38
rs780599441 Chr.11: 118893914-118893914 on Build GRCh38
rs753814058 Chr.11: 118894183-118894183 on Build GRCh38
rs202087123 Chr.11: 118893788-118893788 on Build GRCh38
rs541789846 Chr.11: 118893872-118893872 on Build GRCh38
rs375559046 Chr.11: 118893827-118893827 on Build GRCh38
rs745706377 Chr.11: 118894003-118894003 on Build GRCh38
rs750080878 Chr.11: 118894033-118894033 on Build GRCh38
rs906807201 Chr.11: 118893884-118893884 on Build GRCh38
rs939499069 Chr.11: 118893892-118893892 on Build GRCh38
rs377218103 Chr.11: 118894007-118894007 on Build GRCh38
rs761963728 Chr.11: 118893736-118893736 on Build GRCh38
rs755323597 Chr.11: 118893870-118893870 on Build GRCh38
rs759410345 Chr.11: 118893745-118893745 on Build GRCh38
rs750797549 Chr.11: 118893895-118893895 on Build GRCh38
rs572403236 Chr.11: 118893991-118893991 on Build GRCh38
rs761359221 Chr.11: 118894015-118894015 on Build GRCh38
rs774735122 Chr.11: 118894153-118894153 on Build GRCh38
rs758788445 Chr.11: 118893897-118893897 on Build GRCh38
rs140701663 Chr.11: 118893826-118893826 on Build GRCh38
rs546277181 Chr.11: 118894001-118894001 on Build GRCh38
rs747672304 Chr.11: 118894129-118894129 on Build GRCh38
rs1017008492 Chr.11: 118893958-118893958 on Build GRCh38
rs369772789 Chr.11: 118893865-118893865 on Build GRCh38
rs765989231 Chr.11: 118894147-118894147 on Build GRCh38
rs777654905 Chr.11: 118894063-118894063 on Build GRCh38
rs915286322 Chr.11: 118893844-118893844 on Build GRCh38
rs767979611 Chr.11: 118894051-118894051 on Build GRCh38
rs755436171 Chr.11: 118893935-118893935 on Build GRCh38
rs764923840 Chr.11: 118893886-118893886 on Build GRCh38
rs764867894 Chr.11: 118894029-118894029 on Build GRCh38
rs768758047 Chr.11: 118894114-118894114 on Build GRCh38
rs151215585 Chr.11: 118893871-118893871 on Build GRCh38
rs201054408 Chr.11: 118893777-118893777 on Build GRCh38
rs748200994 Chr.11: 118893974-118893974 on Build GRCh38
rs773400390 Chr.11: 118893938-118893938 on Build GRCh38
rs111656136 Chr.11: 118894105-118894105 on Build GRCh38
rs202193384 Chr.11: 118893776-118893776 on Build GRCh38
rs144650313 Chr.11: 118893866-118893866 on Build GRCh38
rs776330308 Chr.11: 118894014-118894014 on Build GRCh38
rs778052526 Chr.11: 118893989-118893989 on Build GRCh38
rs73011422 Chr.11: 118893733-118893733 on Build GRCh38
rs758882989 Chr.11: 118893807-118893807 on Build GRCh38
rs1005840016 Chr.11: 118893833-118893833 on Build GRCh38
rs774152337 Chr.11: 118893740-118893740 on Build GRCh38
rs368902812 Chr.11: 118894180-118894180 on Build GRCh38
rs1044832671 Chr.11: 118894125-118894125 on Build GRCh38
rs537787473 Chr.11: 118893739-118893739 on Build GRCh38
rs752063152 Chr.11: 118893920-118893920 on Build GRCh38
rs757384100 Chr.11: 118894082-118894082 on Build GRCh38
rs763529010 Chr.11: 118894106-118894106 on Build GRCh38
rs761394882 Chr.11: 118893881-118893881 on Build GRCh38
rs530437351 Chr.11: 118893859-118893859 on Build GRCh38
rs756904573 Chr.11: 118893791-118893791 on Build GRCh38

View Full Product Details