Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118894112-118894607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCR5
Entrez Gene ID
643
Gene Name
C-X-C motif chemokine receptor 5
Gene Aliases
BLR1, CD185, MDR15
UniGene
Hs.113916
Gene Chromosome Location
Chr.11: 118883766-118896271 on Build GRCh38
Transcripts
NM_001716.4 NM_032966.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
224410 c.754C>T p.P252S Chr.11: 118894298-118894298 on Build GRCh38
6309405 c.844G>T p.D282Y Chr.11: 118894388-118894388 on Build GRCh38
220491 c.1048C>T p.Q350* Chr.11: 118894592-118894592 on Build GRCh38
6223883 c.656G>A p.R219Q Chr.11: 118894200-118894200 on Build GRCh38
923657 c.854C>T p.A285V Chr.11: 118894398-118894398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773979825 Chr.11: 118894263-118894263 on Build GRCh38
rs200959025 Chr.11: 118894145-118894145 on Build GRCh38
rs771273984 Chr.11: 118894213-118894213 on Build GRCh38
rs759128895 Chr.11: 118894265-118894265 on Build GRCh38
rs935334049 Chr.11: 118894426-118894426 on Build GRCh38
rs775130399 Chr.11: 118894316-118894316 on Build GRCh38
rs665648 Chr.11: 118894574-118894574 on Build GRCh38
rs750689771 Chr.11: 118894293-118894293 on Build GRCh38
rs891033812 Chr.11: 118894417-118894417 on Build GRCh38
rs200126792 Chr.11: 118894547-118894547 on Build GRCh38
rs771799488 Chr.11: 118894495-118894495 on Build GRCh38
rs776993079 Chr.11: 118894418-118894418 on Build GRCh38
rs914374639 Chr.11: 118894379-118894379 on Build GRCh38
rs759889292 Chr.11: 118894165-118894165 on Build GRCh38
rs775974861 Chr.11: 118894410-118894410 on Build GRCh38
rs761405561 Chr.11: 118894434-118894434 on Build GRCh38
rs147267963 Chr.11: 118894546-118894546 on Build GRCh38
rs770441359 Chr.11: 118894385-118894385 on Build GRCh38
rs371145208 Chr.11: 118894462-118894462 on Build GRCh38
rs535463325 Chr.11: 118894461-118894461 on Build GRCh38
rs753990934 Chr.11: 118894280-118894280 on Build GRCh38
rs373222312 Chr.11: 118894398-118894398 on Build GRCh38
rs768071139 Chr.11: 118894171-118894171 on Build GRCh38
rs374707658 Chr.11: 118894557-118894557 on Build GRCh38
rs762687395 Chr.11: 118894146-118894146 on Build GRCh38
rs778417636 Chr.11: 118894476-118894476 on Build GRCh38
rs572835040 Chr.11: 118894276-118894276 on Build GRCh38
rs756510807 Chr.11: 118894179-118894179 on Build GRCh38
rs373329724 Chr.11: 118894199-118894199 on Build GRCh38
rs768918941 Chr.11: 118894519-118894519 on Build GRCh38
rs767871137 Chr.11: 118894268-118894268 on Build GRCh38
rs753260293 Chr.11: 118894172-118894172 on Build GRCh38
rs12795395 Chr.11: 118894522-118894522 on Build GRCh38
rs753814058 Chr.11: 118894183-118894183 on Build GRCh38
rs201769483 Chr.11: 118894312-118894312 on Build GRCh38
rs752409830 Chr.11: 118894313-118894313 on Build GRCh38
rs750477386 Chr.11: 118894447-118894447 on Build GRCh38
rs1050747752 Chr.11: 118894580-118894580 on Build GRCh38
rs867358468 Chr.11: 118894478-118894478 on Build GRCh38
rs780794925 Chr.11: 118894217-118894217 on Build GRCh38
rs764486324 Chr.11: 118894277-118894277 on Build GRCh38
rs769274153 Chr.11: 118894412-118894412 on Build GRCh38
rs966707486 Chr.11: 118894200-118894200 on Build GRCh38
rs45513292 Chr.11: 118894573-118894573 on Build GRCh38
rs774735122 Chr.11: 118894153-118894153 on Build GRCh38
rs765416491 Chr.11: 118894437-118894437 on Build GRCh38
rs369220984 Chr.11: 118894304-118894304 on Build GRCh38
rs770635701 Chr.11: 118894259-118894259 on Build GRCh38
rs140843011 Chr.11: 118894548-118894548 on Build GRCh38
rs868477374 Chr.11: 118894520-118894520 on Build GRCh38
rs1038959105 Chr.11: 118894565-118894565 on Build GRCh38
rs566470709 Chr.11: 118894288-118894288 on Build GRCh38
rs765989231 Chr.11: 118894147-118894147 on Build GRCh38
rs145276178 Chr.11: 118894376-118894376 on Build GRCh38
rs377341193 Chr.11: 118894258-118894258 on Build GRCh38
rs769567385 Chr.11: 118894219-118894219 on Build GRCh38
rs777565094 Chr.11: 118894460-118894460 on Build GRCh38
rs765033595 Chr.11: 118894516-118894516 on Build GRCh38
rs2230321 Chr.11: 118894378-118894378 on Build GRCh38
rs766678024 Chr.11: 118894550-118894550 on Build GRCh38
rs779569993 Chr.11: 118894507-118894507 on Build GRCh38
rs182696451 Chr.11: 118894273-118894273 on Build GRCh38
rs556054471 Chr.11: 118894411-118894411 on Build GRCh38
rs761269347 Chr.11: 118894274-118894274 on Build GRCh38
rs758630504 Chr.11: 118894296-118894296 on Build GRCh38
rs113967672 Chr.11: 118894399-118894399 on Build GRCh38
rs772823257 Chr.11: 118894235-118894235 on Build GRCh38
rs144033290 Chr.11: 118894523-118894523 on Build GRCh38
rs187389271 Chr.11: 118894456-118894456 on Build GRCh38
rs754811009 Chr.11: 118894214-118894214 on Build GRCh38
rs866800308 Chr.11: 118894528-118894528 on Build GRCh38
rs766477033 Chr.11: 118894448-118894448 on Build GRCh38
rs368902812 Chr.11: 118894180-118894180 on Build GRCh38
rs598207 Chr.11: 118894558-118894558 on Build GRCh38
rs745360877 Chr.11: 118894483-118894483 on Build GRCh38
rs757234761 Chr.11: 118894571-118894571 on Build GRCh38
rs761983769 Chr.11: 118894281-118894281 on Build GRCh38
rs762803932 Chr.11: 118894344-118894346 on Build GRCh38
rs748145415 Chr.11: 118894218-118894218 on Build GRCh38
rs2230320 Chr.11: 118894372-118894372 on Build GRCh38
rs199930702 Chr.11: 118894362-118894362 on Build GRCh38

View Full Product Details