Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118897209-118897699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL9L
Entrez Gene ID
283149
Gene Name
B-cell CLL/lymphoma 9-like
Gene Aliases
BCL9-2, DLNB11
UniGene
Hs.414740
Gene Chromosome Location
Chr.11: 118896140-118926072 on Build GRCh38
Transcripts
NM_182557.2 XM_005271511.2 XM_006718815.2 XM_017017572.1

SNP Detail

SNP ID Chromosome Location
rs577783324 Chr.11: 118897380-118897380 on Build GRCh38
rs899753927 Chr.11: 118897511-118897511 on Build GRCh38
rs1000198472 Chr.11: 118897469-118897469 on Build GRCh38
rs189845413 Chr.11: 118897473-118897473 on Build GRCh38
rs981686923 Chr.11: 118897410-118897410 on Build GRCh38
rs1044979667 Chr.11: 118897333-118897333 on Build GRCh38
rs991673001 Chr.11: 118897416-118897416 on Build GRCh38
rs572994779 Chr.11: 118897482-118897483 on Build GRCh38
rs1020773330 Chr.11: 118897553-118897553 on Build GRCh38
rs899703041 Chr.11: 118897250-118897250 on Build GRCh38
rs532742279 Chr.11: 118897544-118897544 on Build GRCh38
rs930828674 Chr.11: 118897556-118897556 on Build GRCh38
rs554982192 Chr.11: 118897235-118897235 on Build GRCh38
rs1053640101 Chr.11: 118897528-118897528 on Build GRCh38
rs571607356 Chr.11: 118897540-118897540 on Build GRCh38
rs996681105 Chr.11: 118897317-118897317 on Build GRCh38
rs528455360 Chr.11: 118897452-118897452 on Build GRCh38
rs758965475 Chr.11: 118897359-118897359 on Build GRCh38
rs560669838 Chr.11: 118897281-118897281 on Build GRCh38
rs972084506 Chr.11: 118897522-118897522 on Build GRCh38
rs147283386 Chr.11: 118897320-118897321 on Build GRCh38
rs746458735 Chr.11: 118897377-118897377 on Build GRCh38
rs1011685212 Chr.11: 118897547-118897547 on Build GRCh38
rs750786126 Chr.11: 118897320-118897320 on Build GRCh38
rs984505216 Chr.11: 118897562-118897562 on Build GRCh38
rs967332766 Chr.11: 118897409-118897409 on Build GRCh38
rs182056915 Chr.11: 118897601-118897601 on Build GRCh38
rs1047236002 Chr.11: 118897451-118897451 on Build GRCh38
rs544836957 Chr.11: 118897387-118897387 on Build GRCh38
rs988792682 Chr.11: 118897513-118897513 on Build GRCh38
rs927491225 Chr.11: 118897554-118897554 on Build GRCh38
rs1024495264 Chr.11: 118897520-118897520 on Build GRCh38
rs530875574 Chr.11: 118897405-118897405 on Build GRCh38
rs777286144 Chr.11: 118897474-118897474 on Build GRCh38
rs573284920 Chr.11: 118897262-118897262 on Build GRCh38
rs901752506 Chr.11: 118897614-118897614 on Build GRCh38
rs1041023762 Chr.11: 118897483-118897483 on Build GRCh38
rs561303363 Chr.11: 118897442-118897442 on Build GRCh38
rs747699770 Chr.11: 118897443-118897443 on Build GRCh38
rs187098041 Chr.11: 118897603-118897603 on Build GRCh38
rs185500774 Chr.11: 118897391-118897391 on Build GRCh38
rs541148109 Chr.11: 118897292-118897292 on Build GRCh38
rs950223403 Chr.11: 118897319-118897319 on Build GRCh38
rs1029277859 Chr.11: 118897450-118897450 on Build GRCh38
rs1050368308 Chr.11: 118897623-118897623 on Build GRCh38
rs1040285035 Chr.11: 118897414-118897414 on Build GRCh38
rs559155341 Chr.11: 118897372-118897372 on Build GRCh38
rs542937261 Chr.11: 118897406-118897406 on Build GRCh38
rs769215527 Chr.11: 118897465-118897465 on Build GRCh38
rs34674464 Chr.11: 118897645-118897645 on Build GRCh38
rs943730461 Chr.11: 118897668-118897668 on Build GRCh38
rs967385137 Chr.11: 118897568-118897568 on Build GRCh38
rs145472817 Chr.11: 118897622-118897622 on Build GRCh38
rs901774398 Chr.11: 118897437-118897437 on Build GRCh38
rs774354710 Chr.11: 118897653-118897653 on Build GRCh38
rs780519611 Chr.11: 118897369-118897369 on Build GRCh38
rs940345796 Chr.11: 118897413-118897413 on Build GRCh38
rs569097870 Chr.11: 118897602-118897602 on Build GRCh38
rs908290346 Chr.11: 118897663-118897663 on Build GRCh38
rs958953608 Chr.11: 118897615-118897615 on Build GRCh38
rs952980795 Chr.11: 118897630-118897630 on Build GRCh38
rs770550519 Chr.11: 118897635-118897635 on Build GRCh38
rs148838473 Chr.11: 118897646-118897646 on Build GRCh38

View Full Product Details