Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119110483-119110991 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD2L
Entrez Gene ID
9854
Gene Name
C2CD2 like
Gene Aliases
DLNB23, TMEM24
UniGene
Hs.26899
Gene Chromosome Location
Chr.11: 119107061-119118543 on Build GRCh38
Transcripts
NM_001290474.1 NM_014807.4 XM_006718948.3 XM_011543076.2 XM_017018625.1 XM_017018626.1 XM_017018627.1 XM_017018628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3444104 c.470C>T p.S157F Chr.11: 119110580-119110580 on Build GRCh38
3935341 c.643C>A p.L215I Chr.11: 119110919-119110919 on Build GRCh38
3444105 c.595C>T p.P199S Chr.11: 119110871-119110871 on Build GRCh38
2106422 c.484C>T p.R162W Chr.11: 119110594-119110594 on Build GRCh38
4018467 c.581A>C p.N194T Chr.11: 119110857-119110857 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762137452 Chr.11: 119110797-119110797 on Build GRCh38
rs200204602 Chr.11: 119110816-119110816 on Build GRCh38
rs111593658 Chr.11: 119110965-119110965 on Build GRCh38
rs761928272 Chr.11: 119110929-119110929 on Build GRCh38
rs561088763 Chr.11: 119110517-119110517 on Build GRCh38
rs371781479 Chr.11: 119110717-119110717 on Build GRCh38
rs768420843 Chr.11: 119110622-119110622 on Build GRCh38
rs947778500 Chr.11: 119110681-119110681 on Build GRCh38
rs752453437 Chr.11: 119110827-119110827 on Build GRCh38
rs967566654 Chr.11: 119110910-119110910 on Build GRCh38
rs752934761 Chr.11: 119110578-119110578 on Build GRCh38
rs747795681 Chr.11: 119110626-119110626 on Build GRCh38
rs753988050 Chr.11: 119110731-119110731 on Build GRCh38
rs780951362 Chr.11: 119110687-119110687 on Build GRCh38
rs183652831 Chr.11: 119110632-119110632 on Build GRCh38
rs764698979 Chr.11: 119110525-119110525 on Build GRCh38
rs371942666 Chr.11: 119110874-119110874 on Build GRCh38
rs753913548 Chr.11: 119110653-119110653 on Build GRCh38
rs1006846802 Chr.11: 119110766-119110766 on Build GRCh38
rs937324741 Chr.11: 119110963-119110963 on Build GRCh38
rs772665465 Chr.11: 119110901-119110901 on Build GRCh38
rs781116505 Chr.11: 119110815-119110815 on Build GRCh38
rs957531295 Chr.11: 119110544-119110544 on Build GRCh38
rs201774483 Chr.11: 119110672-119110672 on Build GRCh38
rs138832456 Chr.11: 119110633-119110633 on Build GRCh38
rs773213286 Chr.11: 119110964-119110964 on Build GRCh38
rs777122511 Chr.11: 119110924-119110924 on Build GRCh38
rs756471104 Chr.11: 119110584-119110584 on Build GRCh38
rs765510665 Chr.11: 119110949-119110949 on Build GRCh38
rs776161978 Chr.11: 119110911-119110911 on Build GRCh38
rs201942374 Chr.11: 119110916-119110916 on Build GRCh38
rs529684826 Chr.11: 119110635-119110635 on Build GRCh38
rs376805740 Chr.11: 119110567-119110567 on Build GRCh38
rs375034030 Chr.11: 119110707-119110707 on Build GRCh38
rs777661325 Chr.11: 119110836-119110836 on Build GRCh38
rs145920257 Chr.11: 119110594-119110594 on Build GRCh38
rs764226444 Chr.11: 119110651-119110651 on Build GRCh38
rs755427282 Chr.11: 119110568-119110568 on Build GRCh38
rs199809079 Chr.11: 119110595-119110595 on Build GRCh38
rs757149229 Chr.11: 119110753-119110753 on Build GRCh38
rs866955180 Chr.11: 119110515-119110515 on Build GRCh38
rs746771941 Chr.11: 119110613-119110613 on Build GRCh38
rs369181429 Chr.11: 119110850-119110850 on Build GRCh38
rs753165662 Chr.11: 119110644-119110644 on Build GRCh38
rs767396451 Chr.11: 119110968-119110968 on Build GRCh38
rs142730336 Chr.11: 119110640-119110640 on Build GRCh38
rs537765742 Chr.11: 119110786-119110786 on Build GRCh38
rs777324202 Chr.11: 119110701-119110701 on Build GRCh38
rs764326317 Chr.11: 119110726-119110726 on Build GRCh38
rs1018278928 Chr.11: 119110778-119110778 on Build GRCh38
rs774695637 Chr.11: 119110638-119110638 on Build GRCh38
rs373676895 Chr.11: 119110697-119110697 on Build GRCh38
rs558453944 Chr.11: 119110734-119110734 on Build GRCh38
rs928605483 Chr.11: 119110589-119110589 on Build GRCh38
rs188365297 Chr.11: 119110744-119110744 on Build GRCh38
rs376197826 Chr.11: 119110812-119110812 on Build GRCh38
rs770671304 Chr.11: 119110858-119110858 on Build GRCh38
rs200869167 Chr.11: 119110623-119110623 on Build GRCh38
rs754856782 Chr.11: 119110814-119110814 on Build GRCh38
rs1055856596 Chr.11: 119110602-119110602 on Build GRCh38
rs77672284 Chr.11: 119110553-119110553 on Build GRCh38
rs749338039 Chr.11: 119110593-119110593 on Build GRCh38
rs761505988 Chr.11: 119110524-119110524 on Build GRCh38
rs386758016 Chr.11: 119110614-119110616 on Build GRCh38
rs200565961 Chr.11: 119110552-119110552 on Build GRCh38
rs753471771 Chr.11: 119110718-119110718 on Build GRCh38
rs768255307 Chr.11: 119110514-119110514 on Build GRCh38
rs78758351 Chr.11: 119110716-119110716 on Build GRCh38
rs768041337 Chr.11: 119110571-119110571 on Build GRCh38
rs923821011 Chr.11: 119110889-119110889 on Build GRCh38
rs772668918 Chr.11: 119110550-119110550 on Build GRCh38
rs200781372 Chr.11: 119110654-119110654 on Build GRCh38
rs377467763 Chr.11: 119110530-119110530 on Build GRCh38
rs760922364 Chr.11: 119110646-119110646 on Build GRCh38
rs1044877954 Chr.11: 119110689-119110689 on Build GRCh38
rs745374994 Chr.11: 119110888-119110888 on Build GRCh38
rs747953992 Chr.11: 119110691-119110691 on Build GRCh38
rs761151763 Chr.11: 119110724-119110724 on Build GRCh38
rs773734725 Chr.11: 119110592-119110592 on Build GRCh38
rs372583562 Chr.11: 119110686-119110686 on Build GRCh38
rs779101118 Chr.11: 119110666-119110666 on Build GRCh38
rs769498574 Chr.11: 119110627-119110627 on Build GRCh38

View Full Product Details