Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119111233-119111737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD2L
Entrez Gene ID
9854
Gene Name
C2CD2 like
Gene Aliases
DLNB23, TMEM24
UniGene
Hs.26899
Gene Chromosome Location
Chr.11: 119107061-119118543 on Build GRCh38
Transcripts
NM_001290474.1 NM_014807.4 XM_006718948.3 XM_011543076.2 XM_017018625.1 XM_017018626.1 XM_017018627.1 XM_017018628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4450870 c.910+1G>A p.? Chr.11: 119111375-119111375 on Build GRCh38
923752 c.822G>T p.M274I Chr.11: 119111286-119111286 on Build GRCh38
5703885 c.827C>T p.P276L Chr.11: 119111291-119111291 on Build GRCh38
923753 c.828C>A p.P276P Chr.11: 119111292-119111292 on Build GRCh38
5015055 c.869T>A p.L290Q Chr.11: 119111333-119111333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576674224 Chr.11: 119111474-119111474 on Build GRCh38
rs754543575 Chr.11: 119111343-119111343 on Build GRCh38
rs765619710 Chr.11: 119111648-119111648 on Build GRCh38
rs757596227 Chr.11: 119111526-119111526 on Build GRCh38
rs373082466 Chr.11: 119111652-119111652 on Build GRCh38
rs769868088 Chr.11: 119111409-119111409 on Build GRCh38
rs373570091 Chr.11: 119111539-119111539 on Build GRCh38
rs201118399 Chr.11: 119111585-119111585 on Build GRCh38
rs746125415 Chr.11: 119111385-119111385 on Build GRCh38
rs568006975 Chr.11: 119111692-119111692 on Build GRCh38
rs552219067 Chr.11: 119111639-119111639 on Build GRCh38
rs371412317 Chr.11: 119111336-119111336 on Build GRCh38
rs748325593 Chr.11: 119111363-119111363 on Build GRCh38
rs906286295 Chr.11: 119111669-119111669 on Build GRCh38
rs991791886 Chr.11: 119111494-119111494 on Build GRCh38
rs368566781 Chr.11: 119111382-119111382 on Build GRCh38
rs763642146 Chr.11: 119111636-119111636 on Build GRCh38
rs778051698 Chr.11: 119111362-119111362 on Build GRCh38
rs772157508 Chr.11: 119111395-119111395 on Build GRCh38
rs759154283 Chr.11: 119111489-119111489 on Build GRCh38
rs564506700 Chr.11: 119111455-119111455 on Build GRCh38
rs371242980 Chr.11: 119111532-119111532 on Build GRCh38
rs943270341 Chr.11: 119111614-119111614 on Build GRCh38
rs747090293 Chr.11: 119111579-119111579 on Build GRCh38
rs779183220 Chr.11: 119111527-119111527 on Build GRCh38
rs768428400 Chr.11: 119111317-119111317 on Build GRCh38
rs766640522 Chr.11: 119111405-119111405 on Build GRCh38
rs770196267 Chr.11: 119111529-119111529 on Build GRCh38
rs754312442 Chr.11: 119111638-119111638 on Build GRCh38
rs749236711 Chr.11: 119111679-119111679 on Build GRCh38
rs560238744 Chr.11: 119111597-119111597 on Build GRCh38
rs774225008 Chr.11: 119111292-119111292 on Build GRCh38
rs749653059 Chr.11: 119111376-119111376 on Build GRCh38
rs773274065 Chr.11: 119111257-119111257 on Build GRCh38
rs774937522 Chr.11: 119111601-119111601 on Build GRCh38
rs572444827 Chr.11: 119111345-119111345 on Build GRCh38
rs761830492 Chr.11: 119111396-119111396 on Build GRCh38
rs758607306 Chr.11: 119111540-119111540 on Build GRCh38
rs771152661 Chr.11: 119111291-119111291 on Build GRCh38
rs762637334 Chr.11: 119111335-119111335 on Build GRCh38
rs763284839 Chr.11: 119111389-119111389 on Build GRCh38
rs139391781 Chr.11: 119111525-119111525 on Build GRCh38
rs771097735 Chr.11: 119111379-119111379 on Build GRCh38
rs546404330 Chr.11: 119111435-119111435 on Build GRCh38
rs535890289 Chr.11: 119111706-119111706 on Build GRCh38
rs761879735 Chr.11: 119111324-119111324 on Build GRCh38
rs527382278 Chr.11: 119111603-119111603 on Build GRCh38
rs903887316 Chr.11: 119111677-119111677 on Build GRCh38
rs751206256 Chr.11: 119111482-119111482 on Build GRCh38
rs370444138 Chr.11: 119111491-119111491 on Build GRCh38
rs140133210 Chr.11: 119111587-119111587 on Build GRCh38
rs377646752 Chr.11: 119111332-119111332 on Build GRCh38
rs755090059 Chr.11: 119111662-119111662 on Build GRCh38
rs1009251798 Chr.11: 119111655-119111655 on Build GRCh38
rs972100262 Chr.11: 119111562-119111562 on Build GRCh38
rs752158483 Chr.11: 119111497-119111497 on Build GRCh38
rs368232711 Chr.11: 119111413-119111413 on Build GRCh38
rs150066907 Chr.11: 119111553-119111553 on Build GRCh38
rs758582011 Chr.11: 119111650-119111650 on Build GRCh38
rs959212270 Chr.11: 119111436-119111436 on Build GRCh38
rs767052101 Chr.11: 119111493-119111493 on Build GRCh38
rs756544253 Chr.11: 119111511-119111511 on Build GRCh38
rs548284461 Chr.11: 119111453-119111453 on Build GRCh38
rs375477516 Chr.11: 119111353-119111353 on Build GRCh38
rs765902219 Chr.11: 119111417-119111417 on Build GRCh38
rs150900283 Chr.11: 119111323-119111323 on Build GRCh38
rs1001356607 Chr.11: 119111685-119111685 on Build GRCh38
rs773074727 Chr.11: 119111414-119111414 on Build GRCh38
rs374161788 Chr.11: 119111284-119111284 on Build GRCh38
rs368029666 Chr.11: 119111674-119111674 on Build GRCh38
rs779595107 Chr.11: 119111716-119111716 on Build GRCh38
rs917807433 Chr.11: 119111505-119111505 on Build GRCh38
rs549675301 Chr.11: 119111683-119111683 on Build GRCh38
rs778278828 Chr.11: 119111518-119111518 on Build GRCh38
rs570335817 Chr.11: 119111658-119111658 on Build GRCh38
rs747037244 Chr.11: 119111659-119111659 on Build GRCh38
rs759692086 Chr.11: 119111310-119111310 on Build GRCh38
rs750768747 Chr.11: 119111649-119111649 on Build GRCh38
rs767078982 Chr.11: 119111600-119111600 on Build GRCh38
rs34673237 Chr.11: 119111586-119111586 on Build GRCh38
rs762873011 Chr.11: 119111415-119111415 on Build GRCh38

View Full Product Details