Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119112249-119112746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD2L
Entrez Gene ID
9854
Gene Name
C2CD2 like
Gene Aliases
DLNB23, TMEM24
UniGene
Hs.26899
Gene Chromosome Location
Chr.11: 119107061-119118543 on Build GRCh38
Transcripts
NM_001290474.1 NM_014807.4 XM_006718948.3 XM_011543076.2 XM_017018625.1 XM_017018626.1 XM_017018627.1 XM_017018628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
686933 c.1073G>A p.S358N Chr.11: 119112381-119112381 on Build GRCh38
6067895 c.1072A>T p.S358C Chr.11: 119112380-119112380 on Build GRCh38
923754 c.1243C>A p.L415M Chr.11: 119112730-119112730 on Build GRCh38
5714113 c.1212G>A p.E404E Chr.11: 119112609-119112609 on Build GRCh38
5763101 c.1133C>A p.P378Q Chr.11: 119112530-119112530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775097341 Chr.11: 119112328-119112328 on Build GRCh38
rs755076657 Chr.11: 119112277-119112277 on Build GRCh38
rs756122425 Chr.11: 119112503-119112503 on Build GRCh38
rs748815354 Chr.11: 119112684-119112684 on Build GRCh38
rs753228335 Chr.11: 119112693-119112693 on Build GRCh38
rs751306158 Chr.11: 119112650-119112650 on Build GRCh38
rs771581644 Chr.11: 119112584-119112584 on Build GRCh38
rs868123536 Chr.11: 119112589-119112589 on Build GRCh38
rs772557061 Chr.11: 119112709-119112709 on Build GRCh38
rs767956921 Chr.11: 119112653-119112653 on Build GRCh38
rs759996756 Chr.11: 119112600-119112600 on Build GRCh38
rs745864092 Chr.11: 119112413-119112413 on Build GRCh38
rs576025301 Chr.11: 119112322-119112322 on Build GRCh38
rs752828682 Chr.11: 119112612-119112612 on Build GRCh38
rs760748375 Chr.11: 119112716-119112716 on Build GRCh38
rs768257581 Chr.11: 119112552-119112552 on Build GRCh38
rs752029953 Chr.11: 119112414-119112415 on Build GRCh38
rs779890004 Chr.11: 119112426-119112426 on Build GRCh38
rs376786817 Chr.11: 119112549-119112549 on Build GRCh38
rs376445949 Chr.11: 119112358-119112358 on Build GRCh38
rs770287054 Chr.11: 119112435-119112435 on Build GRCh38
rs764066439 Chr.11: 119112631-119112631 on Build GRCh38
rs754746691 Chr.11: 119112659-119112659 on Build GRCh38
rs890713799 Chr.11: 119112573-119112573 on Build GRCh38
rs369410743 Chr.11: 119112592-119112592 on Build GRCh38
rs756319579 Chr.11: 119112495-119112495 on Build GRCh38
rs781411395 Chr.11: 119112364-119112364 on Build GRCh38
rs758713281 Chr.11: 119112351-119112351 on Build GRCh38
rs1037554244 Chr.11: 119112455-119112455 on Build GRCh38
rs750760202 Chr.11: 119112350-119112350 on Build GRCh38
rs773572036 Chr.11: 119112458-119112458 on Build GRCh38
rs373100062 Chr.11: 119112568-119112568 on Build GRCh38
rs372307633 Chr.11: 119112505-119112505 on Build GRCh38
rs750322797 Chr.11: 119112524-119112524 on Build GRCh38
rs529055620 Chr.11: 119112486-119112486 on Build GRCh38
rs553586760 Chr.11: 119112308-119112308 on Build GRCh38
rs954417132 Chr.11: 119112275-119112275 on Build GRCh38
rs369793976 Chr.11: 119112558-119112558 on Build GRCh38
rs751804759 Chr.11: 119112357-119112357 on Build GRCh38
rs913904396 Chr.11: 119112365-119112365 on Build GRCh38
rs746415496 Chr.11: 119112697-119112697 on Build GRCh38
rs201080086 Chr.11: 119112430-119112430 on Build GRCh38
rs747755293 Chr.11: 119112666-119112666 on Build GRCh38
rs762325460 Chr.11: 119112347-119112347 on Build GRCh38
rs941111118 Chr.11: 119112452-119112452 on Build GRCh38
rs780847077 Chr.11: 119112556-119112556 on Build GRCh38
rs748791627 Chr.11: 119112663-119112663 on Build GRCh38
rs199707788 Chr.11: 119112530-119112530 on Build GRCh38
rs545384365 Chr.11: 119112391-119112391 on Build GRCh38
rs1009603575 Chr.11: 119112620-119112620 on Build GRCh38
rs143682926 Chr.11: 119112606-119112606 on Build GRCh38
rs371031696 Chr.11: 119112667-119112667 on Build GRCh38
rs771701762 Chr.11: 119112321-119112321 on Build GRCh38
rs371073216 Chr.11: 119112311-119112311 on Build GRCh38
rs375253280 Chr.11: 119112348-119112348 on Build GRCh38
rs964443835 Chr.11: 119112344-119112344 on Build GRCh38
rs779765066 Chr.11: 119112534-119112534 on Build GRCh38
rs140734829 Chr.11: 119112641-119112641 on Build GRCh38
rs778734695 Chr.11: 119112401-119112401 on Build GRCh38
rs549880903 Chr.11: 119112646-119112647 on Build GRCh38
rs543248590 Chr.11: 119112477-119112477 on Build GRCh38
rs373042322 Chr.11: 119112725-119112725 on Build GRCh38
rs2239896 Chr.11: 119112724-119112724 on Build GRCh38
rs763588684 Chr.11: 119112336-119112336 on Build GRCh38
rs766902383 Chr.11: 119112353-119112353 on Build GRCh38
rs747885394 Chr.11: 119112560-119112560 on Build GRCh38
rs565924451 Chr.11: 119112623-119112623 on Build GRCh38
rs774721742 Chr.11: 119112340-119112340 on Build GRCh38
rs761261973 Chr.11: 119112431-119112431 on Build GRCh38
rs373719904 Chr.11: 119112634-119112634 on Build GRCh38
rs771512046 Chr.11: 119112685-119112685 on Build GRCh38
rs774972096 Chr.11: 119112696-119112696 on Build GRCh38
rs746649703 Chr.11: 119112427-119112427 on Build GRCh38
rs763559660 Chr.11: 119112470-119112470 on Build GRCh38
rs766335218 Chr.11: 119112649-119112649 on Build GRCh38
rs775979236 Chr.11: 119112711-119112711 on Build GRCh38
rs541384884 Chr.11: 119112272-119112272 on Build GRCh38
rs199820148 Chr.11: 119112569-119112569 on Build GRCh38
rs771731860 Chr.11: 119112414-119112414 on Build GRCh38
rs200367337 Chr.11: 119112483-119112483 on Build GRCh38
rs1046444131 Chr.11: 119112408-119112408 on Build GRCh38
rs777093266 Chr.11: 119112672-119112672 on Build GRCh38
rs752031053 Chr.11: 119112484-119112484 on Build GRCh38
rs371616626 Chr.11: 119112305-119112305 on Build GRCh38
rs113111158 Chr.11: 119112400-119112400 on Build GRCh38
rs763360089 Chr.11: 119112571-119112571 on Build GRCh38
rs767879925 Chr.11: 119112501-119112501 on Build GRCh38

View Full Product Details