Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119158156-119158653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG4
Entrez Gene ID
64137
Gene Name
ATP binding cassette subfamily G member 4
Gene Aliases
WHITE2
UniGene
Hs.126378
Gene Chromosome Location
Chr.11: 119149040-119162666 on Build GRCh38
Transcripts
NM_001142505.1 NM_022169.4 XM_011542952.2 XM_017018121.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4658477 c.1075G>A p.D359N Chr.11: 119158240-119158240 on Build GRCh38
1475106 c.1190T>G p.M397R Chr.11: 119158579-119158579 on Build GRCh38
1475105 c.1101C>T p.A367A Chr.11: 119158266-119158266 on Build GRCh38
3808546 c.1178A>C p.H393P Chr.11: 119158567-119158567 on Build GRCh38
5792956 c.1143C>A p.T381T Chr.11: 119158308-119158308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781361056 Chr.11: 119158571-119158571 on Build GRCh38
rs145578285 Chr.11: 119158607-119158607 on Build GRCh38
rs376407071 Chr.11: 119158599-119158599 on Build GRCh38
rs774097984 Chr.11: 119158492-119158492 on Build GRCh38
rs990057024 Chr.11: 119158205-119158205 on Build GRCh38
rs935927568 Chr.11: 119158191-119158191 on Build GRCh38
rs768567393 Chr.11: 119158568-119158568 on Build GRCh38
rs142680859 Chr.11: 119158245-119158245 on Build GRCh38
rs758270241 Chr.11: 119158254-119158254 on Build GRCh38
rs771258605 Chr.11: 119158210-119158210 on Build GRCh38
rs145359289 Chr.11: 119158632-119158632 on Build GRCh38
rs1029023621 Chr.11: 119158577-119158577 on Build GRCh38
rs140264299 Chr.11: 119158619-119158619 on Build GRCh38
rs767711713 Chr.11: 119158223-119158223 on Build GRCh38
rs755710321 Chr.11: 119158411-119158411 on Build GRCh38
rs746048701 Chr.11: 119158447-119158447 on Build GRCh38
rs760398916 Chr.11: 119158598-119158598 on Build GRCh38
rs780380500 Chr.11: 119158457-119158457 on Build GRCh38
rs916959660 Chr.11: 119158597-119158597 on Build GRCh38
rs748614293 Chr.11: 119158186-119158186 on Build GRCh38
rs750967405 Chr.11: 119158631-119158631 on Build GRCh38
rs905395186 Chr.11: 119158332-119158332 on Build GRCh38
rs139556601 Chr.11: 119158331-119158331 on Build GRCh38
rs551382399 Chr.11: 119158440-119158440 on Build GRCh38
rs202066840 Chr.11: 119158356-119158356 on Build GRCh38
rs772229533 Chr.11: 119158352-119158352 on Build GRCh38
rs762668467 Chr.11: 119158387-119158387 on Build GRCh38
rs777228793 Chr.11: 119158376-119158376 on Build GRCh38
rs760040052 Chr.11: 119158529-119158529 on Build GRCh38
rs771908038 Chr.11: 119158591-119158591 on Build GRCh38
rs751214207 Chr.11: 119158293-119158293 on Build GRCh38
rs559240628 Chr.11: 119158407-119158407 on Build GRCh38
rs759380045 Chr.11: 119158496-119158496 on Build GRCh38
rs770668309 Chr.11: 119158487-119158487 on Build GRCh38
rs137946614 Chr.11: 119158433-119158433 on Build GRCh38
rs759685520 Chr.11: 119158215-119158215 on Build GRCh38
rs779603850 Chr.11: 119158543-119158543 on Build GRCh38
rs765259348 Chr.11: 119158243-119158243 on Build GRCh38
rs371251382 Chr.11: 119158628-119158628 on Build GRCh38
rs773931620 Chr.11: 119158575-119158576 on Build GRCh38
rs866064778 Chr.11: 119158408-119158408 on Build GRCh38
rs766171479 Chr.11: 119158390-119158390 on Build GRCh38
rs564152389 Chr.11: 119158228-119158228 on Build GRCh38
rs115411701 Chr.11: 119158479-119158479 on Build GRCh38
rs758899125 Chr.11: 119158552-119158552 on Build GRCh38
rs766009741 Chr.11: 119158282-119158282 on Build GRCh38
rs770143825 Chr.11: 119158188-119158188 on Build GRCh38
rs752463582 Chr.11: 119158523-119158523 on Build GRCh38
rs988866584 Chr.11: 119158575-119158575 on Build GRCh38
rs761465370 Chr.11: 119158618-119158618 on Build GRCh38
rs773264847 Chr.11: 119158384-119158384 on Build GRCh38
rs548296437 Chr.11: 119158574-119158574 on Build GRCh38
rs780071418 Chr.11: 119158564-119158564 on Build GRCh38
rs759214504 Chr.11: 119158398-119158398 on Build GRCh38
rs377482969 Chr.11: 119158469-119158469 on Build GRCh38
rs151021657 Chr.11: 119158247-119158247 on Build GRCh38
rs754374750 Chr.11: 119158425-119158425 on Build GRCh38
rs372541424 Chr.11: 119158340-119158340 on Build GRCh38
rs762179499 Chr.11: 119158414-119158415 on Build GRCh38
rs567443652 Chr.11: 119158616-119158616 on Build GRCh38
rs965413568 Chr.11: 119158451-119158451 on Build GRCh38
rs141411331 Chr.11: 119158220-119158221 on Build GRCh38
rs774948961 Chr.11: 119158594-119158594 on Build GRCh38
rs748264266 Chr.11: 119158572-119158572 on Build GRCh38
rs768783884 Chr.11: 119158477-119158477 on Build GRCh38
rs754350774 Chr.11: 119158535-119158535 on Build GRCh38
rs751161028 Chr.11: 119158394-119158394 on Build GRCh38
rs768874082 Chr.11: 119158348-119158348 on Build GRCh38
rs761679014 Chr.11: 119158239-119158239 on Build GRCh38
rs774334992 Chr.11: 119158576-119158576 on Build GRCh38
rs543574484 Chr.11: 119158346-119158346 on Build GRCh38
rs761153066 Chr.11: 119158224-119158225 on Build GRCh38
rs192734017 Chr.11: 119158348-119158348 on Build GRCh38
rs756551714 Chr.11: 119158339-119158339 on Build GRCh38
rs772962506 Chr.11: 119158478-119158478 on Build GRCh38
rs749707366 Chr.11: 119158195-119158195 on Build GRCh38
rs149710791 Chr.11: 119158573-119158573 on Build GRCh38
rs772400069 Chr.11: 119158453-119158453 on Build GRCh38
rs778141966 Chr.11: 119158420-119158420 on Build GRCh38
rs779442281 Chr.11: 119158446-119158446 on Build GRCh38
rs765072646 Chr.11: 119158383-119158383 on Build GRCh38
rs754425127 Chr.11: 119158294-119158294 on Build GRCh38
rs766001050 Chr.11: 119158441-119158457 on Build GRCh38
rs767068594 Chr.11: 119158510-119158510 on Build GRCh38
rs747267202 Chr.11: 119158566-119158566 on Build GRCh38
rs750804052 Chr.11: 119158549-119158549 on Build GRCh38
rs199518763 Chr.11: 119158190-119158190 on Build GRCh38
rs759324902 Chr.11: 119158587-119158587 on Build GRCh38
rs780699500 Chr.11: 119158314-119158314 on Build GRCh38
rs763676136 Chr.11: 119158533-119158533 on Build GRCh38
rs868434734 Chr.11: 119158235-119158235 on Build GRCh38
rs374632897 Chr.11: 119158539-119158539 on Build GRCh38
rs776574108 Chr.11: 119158233-119158233 on Build GRCh38
rs760857462 Chr.11: 119158361-119158361 on Build GRCh38
rs145297167 Chr.11: 119158212-119158212 on Build GRCh38
rs368221871 Chr.11: 119158370-119158370 on Build GRCh38
rs112039440 Chr.11: 119158228-119158229 on Build GRCh38

View Full Product Details