Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 122867330-122867841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTAM
Entrez Gene ID
56253
Gene Name
cytotoxic and regulatory T-cell molecule
Gene Aliases
CD355
UniGene
Hs.159523
Gene Chromosome Location
Chr.11: 122838419-122872643 on Build GRCh38
Transcripts
NM_001304782.1 NM_019604.3 XM_011542900.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6131533 c.277A>G p.T93A Chr.11: 122867465-122867465 on Build GRCh38
6131531 c.279G>T p.T93T Chr.11: 122867467-122867467 on Build GRCh38
6386780 c.245T>C p.L82P Chr.11: 122867433-122867433 on Build GRCh38
3808612 c.347C>T p.A116V Chr.11: 122867535-122867535 on Build GRCh38
7362746 c.851A>G p.K284R Chr.11: 122867442-122867442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761942279 Chr.11: 122867492-122867492 on Build GRCh38
rs762050318 Chr.11: 122867377-122867377 on Build GRCh38
rs748259175 Chr.11: 122867608-122867608 on Build GRCh38
rs924304151 Chr.11: 122867744-122867744 on Build GRCh38
rs138197219 Chr.11: 122867498-122867498 on Build GRCh38
rs754027245 Chr.11: 122867355-122867357 on Build GRCh38
rs374289653 Chr.11: 122867450-122867450 on Build GRCh38
rs760946346 Chr.11: 122867368-122867368 on Build GRCh38
rs747458758 Chr.11: 122867480-122867480 on Build GRCh38
rs949710657 Chr.11: 122867808-122867808 on Build GRCh38
rs1046615719 Chr.11: 122867505-122867505 on Build GRCh38
rs373967425 Chr.11: 122867585-122867585 on Build GRCh38
rs1046664553 Chr.11: 122867811-122867811 on Build GRCh38
rs539227977 Chr.11: 122867562-122867562 on Build GRCh38
rs202125921 Chr.11: 122867369-122867369 on Build GRCh38
rs894118996 Chr.11: 122867453-122867453 on Build GRCh38
rs1055449409 Chr.11: 122867769-122867769 on Build GRCh38
rs542100512 Chr.11: 122867763-122867763 on Build GRCh38
rs906776327 Chr.11: 122867816-122867816 on Build GRCh38
rs915603245 Chr.11: 122867775-122867775 on Build GRCh38
rs1015050464 Chr.11: 122867535-122867535 on Build GRCh38
rs141494048 Chr.11: 122867466-122867466 on Build GRCh38
rs759287664 Chr.11: 122867548-122867548 on Build GRCh38
rs757884674 Chr.11: 122867560-122867560 on Build GRCh38
rs769137093 Chr.11: 122867570-122867570 on Build GRCh38
rs763802347 Chr.11: 122867539-122867539 on Build GRCh38
rs747506504 Chr.11: 122867563-122867563 on Build GRCh38
rs199542102 Chr.11: 122867550-122867550 on Build GRCh38
rs894062200 Chr.11: 122867805-122867805 on Build GRCh38
rs147908982 Chr.11: 122867458-122867458 on Build GRCh38
rs775832473 Chr.11: 122867467-122867467 on Build GRCh38
rs962821853 Chr.11: 122867652-122867652 on Build GRCh38
rs770791672 Chr.11: 122867433-122867433 on Build GRCh38
rs377141926 Chr.11: 122867591-122867591 on Build GRCh38
rs200352597 Chr.11: 122867411-122867411 on Build GRCh38
rs760421491 Chr.11: 122867725-122867725 on Build GRCh38
rs752554712 Chr.11: 122867530-122867530 on Build GRCh38
rs762904473 Chr.11: 122867418-122867418 on Build GRCh38
rs561584007 Chr.11: 122867577-122867577 on Build GRCh38
rs553505708 Chr.11: 122867356-122867357 on Build GRCh38
rs748506893 Chr.11: 122867573-122867573 on Build GRCh38
rs975278957 Chr.11: 122867655-122867655 on Build GRCh38
rs768929111 Chr.11: 122867482-122867482 on Build GRCh38
rs556135830 Chr.11: 122867693-122867693 on Build GRCh38
rs867968952 Chr.11: 122867578-122867578 on Build GRCh38
rs1005454616 Chr.11: 122867527-122867527 on Build GRCh38
rs534405642 Chr.11: 122867356-122867358 on Build GRCh38
rs764080083 Chr.11: 122867604-122867604 on Build GRCh38
rs750458554 Chr.11: 122867415-122867415 on Build GRCh38
rs553870690 Chr.11: 122867357-122867358 on Build GRCh38
rs745371172 Chr.11: 122867460-122867460 on Build GRCh38
rs574521126 Chr.11: 122867739-122867739 on Build GRCh38
rs935564858 Chr.11: 122867774-122867774 on Build GRCh38
rs1052594421 Chr.11: 122867494-122867494 on Build GRCh38
rs987412929 Chr.11: 122867712-122867712 on Build GRCh38
rs368775563 Chr.11: 122867540-122867540 on Build GRCh38
rs753644341 Chr.11: 122867607-122867607 on Build GRCh38
rs571824584 Chr.11: 122867355-122867358 on Build GRCh38
rs925689774 Chr.11: 122867375-122867375 on Build GRCh38
rs769934812 Chr.11: 122867607-122867608 on Build GRCh38
rs760611485 Chr.11: 122867537-122867537 on Build GRCh38
rs756837929 Chr.11: 122867451-122867451 on Build GRCh38
rs773481639 Chr.11: 122867502-122867502 on Build GRCh38
rs935721894 Chr.11: 122867435-122867435 on Build GRCh38
rs116385810 Chr.11: 122867592-122867592 on Build GRCh38
rs796465163 Chr.11: 122867369-122867370 on Build GRCh38
rs763119560 Chr.11: 122867504-122867504 on Build GRCh38
rs756962853 Chr.11: 122867543-122867543 on Build GRCh38
rs760543026 Chr.11: 122867603-122867603 on Build GRCh38
rs567080622 Chr.11: 122867452-122867452 on Build GRCh38
rs773535114 Chr.11: 122867583-122867583 on Build GRCh38
rs764420168 Chr.11: 122867370-122867370 on Build GRCh38
rs1050793617 Chr.11: 122867371-122867371 on Build GRCh38
rs767546285 Chr.11: 122867429-122867429 on Build GRCh38
rs752602503 Chr.11: 122867431-122867431 on Build GRCh38
rs3134422 Chr.11: 122867374-122867374 on Build GRCh38
rs2272094 Chr.11: 122867553-122867553 on Build GRCh38
rs199677419 Chr.11: 122867517-122867517 on Build GRCh38
rs1052728400 Chr.11: 122867797-122867797 on Build GRCh38
rs567393950 Chr.11: 122867358-122867358 on Build GRCh38
rs150210111 Chr.11: 122867442-122867442 on Build GRCh38
rs925773400 Chr.11: 122867758-122867758 on Build GRCh38
rs781419451 Chr.11: 122867572-122867572 on Build GRCh38
rs76087077 Chr.11: 122867358-122867358 on Build GRCh38
rs775344233 Chr.11: 122867713-122867713 on Build GRCh38
rs777530497 Chr.11: 122867441-122867441 on Build GRCh38
rs147039129 Chr.11: 122867489-122867489 on Build GRCh38
rs57365669 Chr.11: 122867355-122867356 on Build GRCh38
rs775892725 Chr.11: 122867367-122867367 on Build GRCh38

View Full Product Details