Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 122871117-122871658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTAM
Entrez Gene ID
56253
Gene Name
cytotoxic and regulatory T-cell molecule
Gene Aliases
CD355
UniGene
Hs.159523
Gene Chromosome Location
Chr.11: 122838419-122872643 on Build GRCh38
Transcripts
NM_001304782.1 NM_019604.3 XM_011542900.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3444463 c.561C>T p.I187I Chr.11: 122871375-122871375 on Build GRCh38
5349628 c.535C>A p.H179N Chr.11: 122871349-122871349 on Build GRCh38
4415027 c.1125T>C p.N375N Chr.11: 122871342-122871342 on Build GRCh38
5042609 c.463C>T p.P155S Chr.11: 122871277-122871277 on Build GRCh38
6552766 c.1067G>T p.R356L Chr.11: 122871284-122871284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374457179 Chr.11: 122871306-122871306 on Build GRCh38
rs759231974 Chr.11: 122871433-122871433 on Build GRCh38
rs538326492 Chr.11: 122871616-122871616 on Build GRCh38
rs775206778 Chr.11: 122871437-122871437 on Build GRCh38
rs749655692 Chr.11: 122871255-122871255 on Build GRCh38
rs185283159 Chr.11: 122871619-122871619 on Build GRCh38
rs752188383 Chr.11: 122871322-122871322 on Build GRCh38
rs758388478 Chr.11: 122871512-122871512 on Build GRCh38
rs551931464 Chr.11: 122871257-122871257 on Build GRCh38
rs991603718 Chr.11: 122871541-122871541 on Build GRCh38
rs865861316 Chr.11: 122871208-122871208 on Build GRCh38
rs747082536 Chr.11: 122871393-122871393 on Build GRCh38
rs758479042 Chr.11: 122871378-122871378 on Build GRCh38
rs778296301 Chr.11: 122871249-122871249 on Build GRCh38
rs756742868 Chr.11: 122871335-122871335 on Build GRCh38
rs571795531 Chr.11: 122871607-122871607 on Build GRCh38
rs762806413 Chr.11: 122871296-122871296 on Build GRCh38
rs1005609123 Chr.11: 122871160-122871160 on Build GRCh38
rs997302502 Chr.11: 122871390-122871390 on Build GRCh38
rs957876946 Chr.11: 122871530-122871530 on Build GRCh38
rs773053417 Chr.11: 122871284-122871284 on Build GRCh38
rs181554965 Chr.11: 122871583-122871583 on Build GRCh38
rs764693469 Chr.11: 122871339-122871339 on Build GRCh38
rs367838873 Chr.11: 122871442-122871442 on Build GRCh38
rs775504413 Chr.11: 122871277-122871277 on Build GRCh38
rs780116398 Chr.11: 122871389-122871389 on Build GRCh38
rs749087947 Chr.11: 122871406-122871406 on Build GRCh38
rs1017473575 Chr.11: 122871346-122871346 on Build GRCh38
rs567291383 Chr.11: 122871148-122871148 on Build GRCh38
rs34706597 Chr.11: 122871372-122871372 on Build GRCh38
rs768510985 Chr.11: 122871553-122871553 on Build GRCh38
rs759958591 Chr.11: 122871195-122871195 on Build GRCh38
rs1020632208 Chr.11: 122871430-122871430 on Build GRCh38
rs745981859 Chr.11: 122871273-122871273 on Build GRCh38
rs371011462 Chr.11: 122871439-122871439 on Build GRCh38
rs116506147 Chr.11: 122871560-122871560 on Build GRCh38
rs759062648 Chr.11: 122871317-122871317 on Build GRCh38
rs771836932 Chr.11: 122871436-122871436 on Build GRCh38
rs34242434 Chr.11: 122871342-122871342 on Build GRCh38
rs1045699468 Chr.11: 122871185-122871185 on Build GRCh38
rs756649121 Chr.11: 122871222-122871222 on Build GRCh38
rs142339889 Chr.11: 122871343-122871343 on Build GRCh38
rs779094905 Chr.11: 122871345-122871345 on Build GRCh38
rs752808081 Chr.11: 122871269-122871269 on Build GRCh38
rs371043833 Chr.11: 122871281-122871281 on Build GRCh38
rs760229744 Chr.11: 122871441-122871441 on Build GRCh38
rs941447050 Chr.11: 122871578-122871578 on Build GRCh38
rs930182254 Chr.11: 122871517-122871517 on Build GRCh38
rs780182864 Chr.11: 122871524-122871524 on Build GRCh38
rs901582059 Chr.11: 122871370-122871370 on Build GRCh38
rs956786392 Chr.11: 122871293-122871293 on Build GRCh38
rs773978972 Chr.11: 122871413-122871413 on Build GRCh38
rs746797928 Chr.11: 122871529-122871529 on Build GRCh38
rs148121861 Chr.11: 122871276-122871276 on Build GRCh38
rs56148911 Chr.11: 122871446-122871446 on Build GRCh38
rs879029825 Chr.11: 122871379-122871379 on Build GRCh38
rs774035052 Chr.11: 122871310-122871310 on Build GRCh38
rs141267818 Chr.11: 122871283-122871283 on Build GRCh38
rs1916036 Chr.11: 122871320-122871320 on Build GRCh38
rs753279951 Chr.11: 122871221-122871221 on Build GRCh38
rs1039794725 Chr.11: 122871627-122871627 on Build GRCh38
rs993523197 Chr.11: 122871447-122871447 on Build GRCh38
rs777660086 Chr.11: 122871401-122871401 on Build GRCh38
rs767769283 Chr.11: 122871216-122871216 on Build GRCh38

View Full Product Details