Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31382524-31383032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND5B
Entrez Gene ID
160518
Gene Name
DENN domain containing 5B
Gene Aliases
-
UniGene
Hs.118166
Gene Chromosome Location
Chr.12: 31382223-31591018 on Build GRCh38
Transcripts
NM_001308339.1 NM_144973.3 XM_005253326.2 XM_005253327.3 XM_005253330.3 XM_006719040.3 XM_011520590.2 XM_017018896.1 XM_017018897.1 XM_017018898.1 XM_017018899.1

SNP Detail

SNP ID Chromosome Location
rs554045069 Chr.12: 31382600-31382600 on Build GRCh38
rs78114544 Chr.12: 31382587-31382587 on Build GRCh38
rs769112000 Chr.12: 31382943-31382943 on Build GRCh38
rs916594242 Chr.12: 31382899-31382899 on Build GRCh38
rs1027431705 Chr.12: 31382872-31382872 on Build GRCh38
rs966577175 Chr.12: 31382830-31382830 on Build GRCh38
rs2374190 Chr.12: 31382813-31382813 on Build GRCh38
rs79046506 Chr.12: 31382724-31382724 on Build GRCh38
rs905483194 Chr.12: 31382846-31382846 on Build GRCh38
rs1045324703 Chr.12: 31382835-31382835 on Build GRCh38
rs940860239 Chr.12: 31382950-31382952 on Build GRCh38
rs61918600 Chr.12: 31382730-31382730 on Build GRCh38
rs1010848887 Chr.12: 31382815-31382815 on Build GRCh38
rs916811972 Chr.12: 31382578-31382578 on Build GRCh38
rs976039012 Chr.12: 31382678-31382678 on Build GRCh38
rs1028906289 Chr.12: 31382946-31382946 on Build GRCh38
rs148490237 Chr.12: 31382939-31382939 on Build GRCh38
rs1013814566 Chr.12: 31382816-31382816 on Build GRCh38
rs708204 Chr.12: 31382747-31382747 on Build GRCh38
rs1004272870 Chr.12: 31382886-31382886 on Build GRCh38
rs1014683477 Chr.12: 31382889-31382889 on Build GRCh38
rs114799686 Chr.12: 31382596-31382596 on Build GRCh38
rs997835570 Chr.12: 31382764-31382765 on Build GRCh38
rs775808595 Chr.12: 31382882-31382882 on Build GRCh38
rs184724384 Chr.12: 31382800-31382800 on Build GRCh38
rs931668753 Chr.12: 31382716-31382716 on Build GRCh38
rs533185484 Chr.12: 31382867-31382867 on Build GRCh38
rs7963635 Chr.12: 31382860-31382860 on Build GRCh38
rs546870588 Chr.12: 31382911-31382911 on Build GRCh38
rs965911407 Chr.12: 31382673-31382673 on Build GRCh38
rs887175665 Chr.12: 31382876-31382876 on Build GRCh38
rs953453414 Chr.12: 31382987-31382987 on Build GRCh38
rs79810353 Chr.12: 31382728-31382728 on Build GRCh38
rs745898887 Chr.12: 31382855-31382855 on Build GRCh38
rs752376094 Chr.12: 31382691-31382691 on Build GRCh38
rs900955751 Chr.12: 31382752-31382752 on Build GRCh38
rs780755034 Chr.12: 31382593-31382593 on Build GRCh38
rs562625487 Chr.12: 31382717-31382717 on Build GRCh38
rs867264517 Chr.12: 31382722-31382722 on Build GRCh38
rs541461819 Chr.12: 31382692-31382692 on Build GRCh38
rs755851543 Chr.12: 31382704-31382704 on Build GRCh38
rs193299739 Chr.12: 31382650-31382650 on Build GRCh38

View Full Product Details