Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31386759-31387262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND5B
Entrez Gene ID
160518
Gene Name
DENN domain containing 5B
Gene Aliases
-
UniGene
Hs.118166
Gene Chromosome Location
Chr.12: 31382223-31591018 on Build GRCh38
Transcripts
NM_001308339.1 NM_144973.3 XM_005253326.2 XM_005253327.3 XM_005253330.3 XM_006719040.3 XM_011520590.2 XM_017018896.1 XM_017018897.1 XM_017018898.1 XM_017018899.1

SNP Detail

SNP ID Chromosome Location
rs974908176 Chr.12: 31386966-31386966 on Build GRCh38
rs12372655 Chr.12: 31387216-31387216 on Build GRCh38
rs199579076 Chr.12: 31387200-31387201 on Build GRCh38
rs572086446 Chr.12: 31386896-31386896 on Build GRCh38
rs528826544 Chr.12: 31386873-31386873 on Build GRCh38
rs965749192 Chr.12: 31386937-31386937 on Build GRCh38
rs890582688 Chr.12: 31387000-31387000 on Build GRCh38
rs187774813 Chr.12: 31387149-31387149 on Build GRCh38
rs34559645 Chr.12: 31386914-31386915 on Build GRCh38
rs761733373 Chr.12: 31387199-31387200 on Build GRCh38
rs117370384 Chr.12: 31386921-31386921 on Build GRCh38
rs1055759262 Chr.12: 31386785-31386785 on Build GRCh38
rs998572225 Chr.12: 31387076-31387076 on Build GRCh38
rs113980654 Chr.12: 31387008-31387008 on Build GRCh38
rs74084448 Chr.12: 31387008-31387008 on Build GRCh38
rs951639586 Chr.12: 31386903-31386903 on Build GRCh38
rs945977696 Chr.12: 31387040-31387040 on Build GRCh38
rs569454820 Chr.12: 31387176-31387176 on Build GRCh38
rs74322203 Chr.12: 31387209-31387209 on Build GRCh38
rs4931487 Chr.12: 31386962-31386962 on Build GRCh38
rs959845818 Chr.12: 31387135-31387135 on Build GRCh38
rs1056273664 Chr.12: 31387178-31387178 on Build GRCh38
rs752775565 Chr.12: 31386933-31386933 on Build GRCh38
rs1008468354 Chr.12: 31387003-31387003 on Build GRCh38
rs960482242 Chr.12: 31386793-31386793 on Build GRCh38
rs751518159 Chr.12: 31386788-31386788 on Build GRCh38
rs1023391920 Chr.12: 31387162-31387162 on Build GRCh38
rs928472715 Chr.12: 31387162-31387163 on Build GRCh38
rs138085124 Chr.12: 31387228-31387228 on Build GRCh38
rs529773470 Chr.12: 31387007-31387007 on Build GRCh38
rs754085760 Chr.12: 31387163-31387163 on Build GRCh38
rs1035841085 Chr.12: 31387077-31387077 on Build GRCh38
rs367708048 Chr.12: 31387147-31387147 on Build GRCh38
rs576301908 Chr.12: 31387020-31387020 on Build GRCh38
rs916153081 Chr.12: 31386805-31386805 on Build GRCh38
rs749021281 Chr.12: 31387162-31387162 on Build GRCh38
rs1021270325 Chr.12: 31387042-31387042 on Build GRCh38
rs899732521 Chr.12: 31387021-31387021 on Build GRCh38
rs756228231 Chr.12: 31386983-31386983 on Build GRCh38
rs1011517240 Chr.12: 31387038-31387038 on Build GRCh38
rs370357512 Chr.12: 31386875-31386875 on Build GRCh38
rs989853760 Chr.12: 31387019-31387019 on Build GRCh38
rs995554573 Chr.12: 31386829-31386829 on Build GRCh38
rs183417309 Chr.12: 31386931-31386931 on Build GRCh38
rs967039356 Chr.12: 31387058-31387058 on Build GRCh38
rs1023110287 Chr.12: 31387200-31387200 on Build GRCh38
rs969197006 Chr.12: 31387199-31387199 on Build GRCh38
rs991705747 Chr.12: 31386803-31386803 on Build GRCh38
rs12372313 Chr.12: 31386999-31386999 on Build GRCh38

View Full Product Details