Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31398115-31398646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND5B
Entrez Gene ID
160518
Gene Name
DENN domain containing 5B
Gene Aliases
-
UniGene
Hs.118166
Gene Chromosome Location
Chr.12: 31382223-31591018 on Build GRCh38
Transcripts
NM_001308339.1 NM_144973.3 XM_005253326.2 XM_005253327.3 XM_005253330.3 XM_006719040.3 XM_011520590.2 XM_017018896.1 XM_017018897.1 XM_017018898.1 XM_017018899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6398099 c.3087G>A p.W1029* Chr.12: 31398344-31398344 on Build GRCh38
5814443 c.3130A>G p.I1044V Chr.12: 31398301-31398301 on Build GRCh38
5980273 c.3356_3357insA p.N1119fs*26 Chr.12: 31398179-31398180 on Build GRCh38
6320250 c.3209C>A p.T1070N Chr.12: 31398222-31398222 on Build GRCh38
5925671 c.3350G>A p.G1117E Chr.12: 31398186-31398186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969462227 Chr.12: 31398540-31398540 on Build GRCh38
rs371791166 Chr.12: 31398182-31398182 on Build GRCh38
rs546235138 Chr.12: 31398418-31398418 on Build GRCh38
rs372149130 Chr.12: 31398279-31398279 on Build GRCh38
rs1798769 Chr.12: 31398416-31398416 on Build GRCh38
rs888726639 Chr.12: 31398333-31398333 on Build GRCh38
rs1013455629 Chr.12: 31398526-31398526 on Build GRCh38
rs747034281 Chr.12: 31398543-31398545 on Build GRCh38
rs957681917 Chr.12: 31398217-31398217 on Build GRCh38
rs759848443 Chr.12: 31398329-31398329 on Build GRCh38
rs758574471 Chr.12: 31398506-31398507 on Build GRCh38
rs1017914398 Chr.12: 31398464-31398464 on Build GRCh38
rs771773537 Chr.12: 31398278-31398278 on Build GRCh38
rs772412748 Chr.12: 31398175-31398177 on Build GRCh38
rs189699285 Chr.12: 31398147-31398147 on Build GRCh38
rs919536705 Chr.12: 31398617-31398617 on Build GRCh38
rs568254961 Chr.12: 31398588-31398588 on Build GRCh38
rs981194426 Chr.12: 31398367-31398367 on Build GRCh38
rs560370567 Chr.12: 31398434-31398434 on Build GRCh38
rs966386070 Chr.12: 31398543-31398543 on Build GRCh38
rs1046518171 Chr.12: 31398260-31398260 on Build GRCh38
rs1055773182 Chr.12: 31398314-31398314 on Build GRCh38
rs1821238 Chr.12: 31398598-31398598 on Build GRCh38
rs754181092 Chr.12: 31398316-31398316 on Build GRCh38
rs768735757 Chr.12: 31398262-31398262 on Build GRCh38
rs1052402803 Chr.12: 31398517-31398517 on Build GRCh38
rs182449176 Chr.12: 31398436-31398436 on Build GRCh38
rs910109920 Chr.12: 31398612-31398612 on Build GRCh38
rs151059114 Chr.12: 31398348-31398348 on Build GRCh38
rs542934920 Chr.12: 31398428-31398429 on Build GRCh38
rs758905846 Chr.12: 31398369-31398369 on Build GRCh38
rs774479195 Chr.12: 31398264-31398264 on Build GRCh38
rs376506006 Chr.12: 31398284-31398284 on Build GRCh38
rs758524872 Chr.12: 31398200-31398200 on Build GRCh38
rs913570926 Chr.12: 31398538-31398538 on Build GRCh38
rs989072061 Chr.12: 31398494-31398494 on Build GRCh38
rs368562763 Chr.12: 31398419-31398419 on Build GRCh38
rs777881908 Chr.12: 31398205-31398205 on Build GRCh38
rs909426537 Chr.12: 31398402-31398402 on Build GRCh38
rs765567611 Chr.12: 31398332-31398332 on Build GRCh38
rs747263395 Chr.12: 31398218-31398218 on Build GRCh38
rs377549094 Chr.12: 31398597-31398597 on Build GRCh38
rs774928242 Chr.12: 31398286-31398286 on Build GRCh38
rs1002175578 Chr.12: 31398543-31398543 on Build GRCh38
rs761808285 Chr.12: 31398275-31398275 on Build GRCh38
rs913625090 Chr.12: 31398247-31398247 on Build GRCh38
rs775711904 Chr.12: 31398392-31398393 on Build GRCh38
rs954694513 Chr.12: 31398428-31398428 on Build GRCh38
rs998680924 Chr.12: 31398417-31398417 on Build GRCh38
rs367928196 Chr.12: 31398144-31398144 on Build GRCh38
rs779727658 Chr.12: 31398249-31398249 on Build GRCh38
rs942283732 Chr.12: 31398412-31398412 on Build GRCh38
rs1039781195 Chr.12: 31398487-31398487 on Build GRCh38
rs373674194 Chr.12: 31398243-31398243 on Build GRCh38
rs368956997 Chr.12: 31398250-31398250 on Build GRCh38
rs779356378 Chr.12: 31398160-31398160 on Build GRCh38
rs957961518 Chr.12: 31398468-31398468 on Build GRCh38
rs982259350 Chr.12: 31398567-31398567 on Build GRCh38
rs369420310 Chr.12: 31398335-31398335 on Build GRCh38
rs559926838 Chr.12: 31398176-31398176 on Build GRCh38
rs989124346 Chr.12: 31398219-31398219 on Build GRCh38
rs1037050603 Chr.12: 31398378-31398378 on Build GRCh38

View Full Product Details