Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 32328125-32328671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICD1
Entrez Gene ID
636
Gene Name
BICD cargo adaptor 1
Gene Aliases
BICD
UniGene
Hs.505202
Gene Chromosome Location
Chr.12: 32106736-32383637 on Build GRCh38
Transcripts
NM_001003398.1 NM_001714.2 XM_005253465.3 XM_005253466.2 XM_006719134.3 XM_011520812.2 XM_011520813.1 XM_011520814.2 XM_011520815.2 XM_011520816.2 XM_017019797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
693328 c.1970G>T p.R657L Chr.12: 32328425-32328425 on Build GRCh38
1705360 c.1951C>T p.R651C Chr.12: 32328406-32328406 on Build GRCh38
5478197 c.1808T>C p.V603A Chr.12: 32328263-32328263 on Build GRCh38
318924 c.1939T>G p.L647V Chr.12: 32328394-32328394 on Build GRCh38
4041423 c.1737A>G p.E579E Chr.12: 32328192-32328192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530703367 Chr.12: 32328632-32328632 on Build GRCh38
rs201636879 Chr.12: 32328420-32328420 on Build GRCh38
rs536283864 Chr.12: 32328261-32328261 on Build GRCh38
rs201557218 Chr.12: 32328346-32328346 on Build GRCh38
rs913043415 Chr.12: 32328167-32328167 on Build GRCh38
rs750334933 Chr.12: 32328293-32328293 on Build GRCh38
rs371155476 Chr.12: 32328575-32328575 on Build GRCh38
rs761183677 Chr.12: 32328260-32328260 on Build GRCh38
rs147065047 Chr.12: 32328184-32328184 on Build GRCh38
rs200686810 Chr.12: 32328425-32328425 on Build GRCh38
rs927168876 Chr.12: 32328639-32328639 on Build GRCh38
rs201096392 Chr.12: 32328456-32328456 on Build GRCh38
rs780960219 Chr.12: 32328157-32328157 on Build GRCh38
rs772921660 Chr.12: 32328585-32328585 on Build GRCh38
rs200579974 Chr.12: 32328345-32328345 on Build GRCh38
rs199910195 Chr.12: 32328537-32328537 on Build GRCh38
rs754669082 Chr.12: 32328303-32328303 on Build GRCh38
rs998314203 Chr.12: 32328389-32328389 on Build GRCh38
rs957088230 Chr.12: 32328518-32328518 on Build GRCh38
rs905754691 Chr.12: 32328251-32328251 on Build GRCh38
rs199605193 Chr.12: 32328300-32328300 on Build GRCh38
rs200110308 Chr.12: 32328440-32328440 on Build GRCh38
rs199498750 Chr.12: 32328336-32328336 on Build GRCh38
rs751383007 Chr.12: 32328155-32328155 on Build GRCh38
rs766277520 Chr.12: 32328428-32328428 on Build GRCh38
rs199952837 Chr.12: 32328154-32328154 on Build GRCh38
rs750220029 Chr.12: 32328424-32328424 on Build GRCh38
rs375672599 Chr.12: 32328461-32328461 on Build GRCh38
rs961470910 Chr.12: 32328328-32328328 on Build GRCh38
rs1009886917 Chr.12: 32328352-32328352 on Build GRCh38
rs951352633 Chr.12: 32328237-32328237 on Build GRCh38
rs981232894 Chr.12: 32328633-32328633 on Build GRCh38
rs3748276 Chr.12: 32328643-32328643 on Build GRCh38
rs765534131 Chr.12: 32328274-32328274 on Build GRCh38
rs202126956 Chr.12: 32328567-32328567 on Build GRCh38
rs949864242 Chr.12: 32328199-32328199 on Build GRCh38
rs747373437 Chr.12: 32328455-32328455 on Build GRCh38
rs201720708 Chr.12: 32328321-32328321 on Build GRCh38
rs751440161 Chr.12: 32328278-32328278 on Build GRCh38
rs139652205 Chr.12: 32328519-32328519 on Build GRCh38
rs944681292 Chr.12: 32328266-32328266 on Build GRCh38
rs774749871 Chr.12: 32328235-32328235 on Build GRCh38
rs201431305 Chr.12: 32328299-32328299 on Build GRCh38
rs1006038440 Chr.12: 32328309-32328309 on Build GRCh38
rs200444584 Chr.12: 32328310-32328310 on Build GRCh38
rs775708453 Chr.12: 32328538-32328538 on Build GRCh38
rs116009500 Chr.12: 32328279-32328279 on Build GRCh38
rs763175978 Chr.12: 32328419-32328419 on Build GRCh38
rs113004430 Chr.12: 32328534-32328534 on Build GRCh38
rs768284104 Chr.12: 32328221-32328221 on Build GRCh38
rs755181806 Chr.12: 32328152-32328152 on Build GRCh38
rs776895522 Chr.12: 32328399-32328399 on Build GRCh38
rs757331139 Chr.12: 32328307-32328307 on Build GRCh38
rs1015652371 Chr.12: 32328320-32328320 on Build GRCh38
rs148101509 Chr.12: 32328268-32328268 on Build GRCh38
rs1025672361 Chr.12: 32328628-32328628 on Build GRCh38
rs200712359 Chr.12: 32328402-32328402 on Build GRCh38
rs755594776 Chr.12: 32328180-32328180 on Build GRCh38
rs199856719 Chr.12: 32328396-32328396 on Build GRCh38
rs772445426 Chr.12: 32328238-32328238 on Build GRCh38
rs201010499 Chr.12: 32328552-32328552 on Build GRCh38
rs769953540 Chr.12: 32328406-32328406 on Build GRCh38
rs778054016 Chr.12: 32328443-32328443 on Build GRCh38
rs763144810 Chr.12: 32328553-32328553 on Build GRCh38
rs753739430 Chr.12: 32328429-32328429 on Build GRCh38
rs748485099 Chr.12: 32328236-32328236 on Build GRCh38
rs746325904 Chr.12: 32328533-32328533 on Build GRCh38
rs369094443 Chr.12: 32328407-32328407 on Build GRCh38
rs779598814 Chr.12: 32328297-32328297 on Build GRCh38
rs200346745 Chr.12: 32328292-32328292 on Build GRCh38
rs759892057 Chr.12: 32328269-32328269 on Build GRCh38
rs765101601 Chr.12: 32328526-32328526 on Build GRCh38
rs747533405 Chr.12: 32328348-32328348 on Build GRCh38
rs3748275 Chr.12: 32328159-32328159 on Build GRCh38
rs766117749 Chr.12: 32328606-32328606 on Build GRCh38
rs771451999 Chr.12: 32328397-32328397 on Build GRCh38
rs201956163 Chr.12: 32328246-32328246 on Build GRCh38
rs202218997 Chr.12: 32328168-32328168 on Build GRCh38
rs768900442 Chr.12: 32328583-32328583 on Build GRCh38
rs201348197 Chr.12: 32328211-32328211 on Build GRCh38
rs201914326 Chr.12: 32328285-32328285 on Build GRCh38
rs752293409 Chr.12: 32328438-32328438 on Build GRCh38
rs754124048 Chr.12: 32328151-32328151 on Build GRCh38
rs756076471 Chr.12: 32328296-32328296 on Build GRCh38
rs749008364 Chr.12: 32328219-32328219 on Build GRCh38
rs34447944 Chr.12: 32328435-32328435 on Build GRCh38
rs200547848 Chr.12: 32328599-32328599 on Build GRCh38
rs754508990 Chr.12: 32328433-32328433 on Build GRCh38

View Full Product Details