Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 38675248-38675787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE8
Entrez Gene ID
144402
Gene Name
copine 8
Gene Aliases
-
UniGene
Hs.40910
Gene Chromosome Location
Chr.12: 38652200-38908593 on Build GRCh38
Transcripts
NM_153634.2 XM_017018851.1 XM_017018852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4772757 c.1412T>A p.V471D Chr.12: 38675737-38675737 on Build GRCh38
6299597 c.1420G>A p.A474T Chr.12: 38675729-38675729 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs34364230 Chr.12: 38675325-38675326 on Build GRCh38
rs950029683 Chr.12: 38675361-38675361 on Build GRCh38
rs765595982 Chr.12: 38675759-38675759 on Build GRCh38
rs765058645 Chr.12: 38675622-38675622 on Build GRCh38
rs940915019 Chr.12: 38675526-38675526 on Build GRCh38
rs776141018 Chr.12: 38675700-38675700 on Build GRCh38
rs758934752 Chr.12: 38675760-38675760 on Build GRCh38
rs1022400525 Chr.12: 38675405-38675405 on Build GRCh38
rs1049672901 Chr.12: 38675501-38675501 on Build GRCh38
rs534978948 Chr.12: 38675481-38675481 on Build GRCh38
rs550104002 Chr.12: 38675319-38675319 on Build GRCh38
rs960688905 Chr.12: 38675444-38675444 on Build GRCh38
rs760058276 Chr.12: 38675690-38675690 on Build GRCh38
rs984399550 Chr.12: 38675545-38675545 on Build GRCh38
rs186964895 Chr.12: 38675456-38675456 on Build GRCh38
rs750428674 Chr.12: 38675749-38675749 on Build GRCh38
rs908991712 Chr.12: 38675567-38675567 on Build GRCh38
rs549194148 Chr.12: 38675547-38675547 on Build GRCh38
rs1033225276 Chr.12: 38675708-38675708 on Build GRCh38
rs141633945 Chr.12: 38675461-38675461 on Build GRCh38
rs1040599924 Chr.12: 38675535-38675535 on Build GRCh38
rs900111957 Chr.12: 38675276-38675276 on Build GRCh38
rs1024210753 Chr.12: 38675761-38675761 on Build GRCh38
rs568263130 Chr.12: 38675558-38675558 on Build GRCh38
rs371583800 Chr.12: 38675709-38675709 on Build GRCh38
rs755561174 Chr.12: 38675447-38675447 on Build GRCh38
rs781748726 Chr.12: 38675536-38675536 on Build GRCh38
rs988818232 Chr.12: 38675304-38675304 on Build GRCh38
rs368118379 Chr.12: 38675698-38675698 on Build GRCh38
rs968098707 Chr.12: 38675430-38675430 on Build GRCh38
rs537381857 Chr.12: 38675566-38675566 on Build GRCh38
rs886699208 Chr.12: 38675519-38675519 on Build GRCh38
rs1872638 Chr.12: 38675347-38675347 on Build GRCh38
rs955890457 Chr.12: 38675718-38675718 on Build GRCh38
rs12581365 Chr.12: 38675572-38675572 on Build GRCh38
rs980651296 Chr.12: 38675432-38675432 on Build GRCh38
rs770273458 Chr.12: 38675600-38675600 on Build GRCh38
rs367778046 Chr.12: 38675758-38675758 on Build GRCh38
rs992490817 Chr.12: 38675454-38675454 on Build GRCh38
rs10875962 Chr.12: 38675442-38675442 on Build GRCh38
rs554992718 Chr.12: 38675509-38675509 on Build GRCh38
rs1039907389 Chr.12: 38675589-38675589 on Build GRCh38
rs137976772 Chr.12: 38675291-38675291 on Build GRCh38
rs191982859 Chr.12: 38675655-38675655 on Build GRCh38
rs1010197371 Chr.12: 38675757-38675757 on Build GRCh38
rs772812928 Chr.12: 38675678-38675678 on Build GRCh38
rs34033452 Chr.12: 38675480-38675481 on Build GRCh38
rs997792395 Chr.12: 38675619-38675619 on Build GRCh38
rs1032946693 Chr.12: 38675273-38675273 on Build GRCh38

View Full Product Details