Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 39551939-39552473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCD2
Entrez Gene ID
225
Gene Name
ATP binding cassette subfamily D member 2
Gene Aliases
ABC39, ALDL1, ALDR, ALDRP, hALDR
UniGene
Hs.117852
Gene Chromosome Location
Chr.12: 39530966-39651333 on Build GRCh38
Transcripts
NM_005164.3 XM_011538027.2 XM_017018992.1

SNP Detail

SNP ID Chromosome Location
rs920290184 Chr.12: 39552042-39552042 on Build GRCh38
rs1008969921 Chr.12: 39552379-39552379 on Build GRCh38
rs1013478325 Chr.12: 39552443-39552443 on Build GRCh38
rs780785058 Chr.12: 39552315-39552316 on Build GRCh38
rs922139163 Chr.12: 39552401-39552401 on Build GRCh38
rs866252034 Chr.12: 39552096-39552096 on Build GRCh38
rs774142151 Chr.12: 39552006-39552006 on Build GRCh38
rs745667808 Chr.12: 39552108-39552108 on Build GRCh38
rs969870152 Chr.12: 39552269-39552269 on Build GRCh38
rs150538441 Chr.12: 39551966-39551966 on Build GRCh38
rs1037581115 Chr.12: 39552232-39552232 on Build GRCh38
rs1000440591 Chr.12: 39552109-39552109 on Build GRCh38
rs958958246 Chr.12: 39552418-39552418 on Build GRCh38
rs983538176 Chr.12: 39552316-39552316 on Build GRCh38
rs1022763172 Chr.12: 39551987-39551987 on Build GRCh38
rs1012292348 Chr.12: 39552218-39552218 on Build GRCh38
rs771953947 Chr.12: 39552126-39552126 on Build GRCh38
rs970522940 Chr.12: 39552167-39552167 on Build GRCh38
rs996007044 Chr.12: 39552009-39552009 on Build GRCh38
rs1042227489 Chr.12: 39552398-39552398 on Build GRCh38
rs896571287 Chr.12: 39552209-39552209 on Build GRCh38
rs924213783 Chr.12: 39552446-39552446 on Build GRCh38
rs542123305 Chr.12: 39552427-39552427 on Build GRCh38
rs375400832 Chr.12: 39552171-39552171 on Build GRCh38
rs866659820 Chr.12: 39552191-39552191 on Build GRCh38
rs920518025 Chr.12: 39552256-39552256 on Build GRCh38
rs1009287914 Chr.12: 39552062-39552062 on Build GRCh38
rs950588081 Chr.12: 39552277-39552277 on Build GRCh38
rs537606834 Chr.12: 39552271-39552271 on Build GRCh38
rs868815934 Chr.12: 39552403-39552403 on Build GRCh38
rs775443203 Chr.12: 39552138-39552138 on Build GRCh38
rs1023258454 Chr.12: 39552223-39552223 on Build GRCh38
rs890556676 Chr.12: 39552052-39552052 on Build GRCh38
rs186745926 Chr.12: 39552326-39552326 on Build GRCh38
rs576616234 Chr.12: 39552155-39552155 on Build GRCh38
rs148486913 Chr.12: 39552139-39552139 on Build GRCh38
rs746978730 Chr.12: 39552257-39552257 on Build GRCh38
rs372272161 Chr.12: 39552273-39552273 on Build GRCh38
rs765356647 Chr.12: 39552175-39552175 on Build GRCh38
rs559508926 Chr.12: 39552051-39552052 on Build GRCh38
rs1020263326 Chr.12: 39552073-39552073 on Build GRCh38
rs911647799 Chr.12: 39552091-39552091 on Build GRCh38
rs916288028 Chr.12: 39552150-39552150 on Build GRCh38
rs928941031 Chr.12: 39552046-39552046 on Build GRCh38
rs966192046 Chr.12: 39552330-39552330 on Build GRCh38
rs969964158 Chr.12: 39551989-39551989 on Build GRCh38
rs974518185 Chr.12: 39552352-39552352 on Build GRCh38
rs375367812 Chr.12: 39552410-39552410 on Build GRCh38
rs1043979462 Chr.12: 39552429-39552429 on Build GRCh38
rs902553270 Chr.12: 39552147-39552147 on Build GRCh38
rs946874674 Chr.12: 39552408-39552408 on Build GRCh38
rs533631867 Chr.12: 39552079-39552079 on Build GRCh38
rs973313537 Chr.12: 39552007-39552007 on Build GRCh38

View Full Product Details