Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 74538893-74539404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN7L3B
Entrez Gene ID
552889
Gene Name
ataxin 7 like 3B
Gene Aliases
lnc-SCA7
UniGene
Hs.744849
Gene Chromosome Location
Chr.12: 74537771-74541452 on Build GRCh38
Transcripts
NM_001136262.1

SNP Detail

SNP ID Chromosome Location
rs948512670 Chr.12: 74538954-74538954 on Build GRCh38
rs3892342 Chr.12: 74539023-74539023 on Build GRCh38
rs906934271 Chr.12: 74538957-74538957 on Build GRCh38
rs755774515 Chr.12: 74539001-74539001 on Build GRCh38
rs528324013 Chr.12: 74539082-74539082 on Build GRCh38
rs564076077 Chr.12: 74539292-74539292 on Build GRCh38
rs11557005 Chr.12: 74539242-74539242 on Build GRCh38
rs996526694 Chr.12: 74538983-74538983 on Build GRCh38
rs745525345 Chr.12: 74538971-74538971 on Build GRCh38
rs932430660 Chr.12: 74539291-74539292 on Build GRCh38
rs928403130 Chr.12: 74539211-74539211 on Build GRCh38
rs140410529 Chr.12: 74539041-74539041 on Build GRCh38
rs1019060360 Chr.12: 74539318-74539318 on Build GRCh38
rs189485719 Chr.12: 74539307-74539307 on Build GRCh38
rs67268352 Chr.12: 74539321-74539324 on Build GRCh38
rs898085199 Chr.12: 74538997-74538997 on Build GRCh38
rs930921849 Chr.12: 74539276-74539276 on Build GRCh38
rs989791819 Chr.12: 74539279-74539279 on Build GRCh38
rs192859927 Chr.12: 74539372-74539372 on Build GRCh38
rs552774473 Chr.12: 74538955-74538955 on Build GRCh38
rs966094772 Chr.12: 74539039-74539039 on Build GRCh38
rs995509786 Chr.12: 74539089-74539089 on Build GRCh38
rs1027840143 Chr.12: 74539005-74539005 on Build GRCh38
rs915708752 Chr.12: 74538944-74538944 on Build GRCh38
rs990446584 Chr.12: 74539177-74539177 on Build GRCh38
rs890939841 Chr.12: 74539011-74539011 on Build GRCh38
rs948585632 Chr.12: 74539185-74539185 on Build GRCh38
rs981271152 Chr.12: 74539194-74539194 on Build GRCh38
rs879845060 Chr.12: 74539228-74539228 on Build GRCh38
rs540617885 Chr.12: 74539249-74539249 on Build GRCh38
rs184459910 Chr.12: 74539006-74539006 on Build GRCh38
rs924342237 Chr.12: 74538962-74538962 on Build GRCh38
rs1036702205 Chr.12: 74539227-74539227 on Build GRCh38
rs561353365 Chr.12: 74539166-74539166 on Build GRCh38
rs915819710 Chr.12: 74539184-74539184 on Build GRCh38
rs957330402 Chr.12: 74539072-74539072 on Build GRCh38
rs1031686301 Chr.12: 74539331-74539331 on Build GRCh38
rs777446799 Chr.12: 74539318-74539319 on Build GRCh38
rs374422675 Chr.12: 74539081-74539081 on Build GRCh38
rs752875023 Chr.12: 74539174-74539175 on Build GRCh38
rs898130557 Chr.12: 74539243-74539243 on Build GRCh38
rs533056417 Chr.12: 74539294-74539294 on Build GRCh38
rs998874533 Chr.12: 74539326-74539326 on Build GRCh38
rs578247468 Chr.12: 74538989-74538989 on Build GRCh38
rs771967213 Chr.12: 74539321-74539321 on Build GRCh38
rs10579692 Chr.12: 74539319-74539322 on Build GRCh38
rs370342400 Chr.12: 74539037-74539037 on Build GRCh38
rs748021551 Chr.12: 74539289-74539289 on Build GRCh38
rs914208322 Chr.12: 74539286-74539286 on Build GRCh38
rs959457289 Chr.12: 74539165-74539165 on Build GRCh38
rs1019008037 Chr.12: 74539030-74539030 on Build GRCh38
rs924193653 Chr.12: 74539272-74539272 on Build GRCh38
rs569485946 Chr.12: 74538964-74538964 on Build GRCh38
rs971304128 Chr.12: 74539237-74539237 on Build GRCh38
rs571757963 Chr.12: 74539245-74539245 on Build GRCh38
rs1026965014 Chr.12: 74539135-74539135 on Build GRCh38
rs377451827 Chr.12: 74539004-74539004 on Build GRCh38
rs576107066 Chr.12: 74539040-74539040 on Build GRCh38
rs1054013811 Chr.12: 74538969-74538969 on Build GRCh38
rs939718213 Chr.12: 74539214-74539214 on Build GRCh38

View Full Product Details