Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75410306-75410845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIPR1L2
Entrez Gene ID
144321
Gene Name
GLI pathogenesis-related 1 like 2
Gene Aliases
-
UniGene
Hs.406728
Gene Chromosome Location
Chr.12: 75391070-75432688 on Build GRCh38
Transcripts
NM_001270396.1 NM_152436.2 NR_072995.1 XM_011537949.2 XM_011537950.2 XM_017018848.1 XM_017018849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4044844 c.406A>C p.S136R Chr.12: 75410605-75410605 on Build GRCh38
3465337 c.331C>T p.P111S Chr.12: 75410530-75410530 on Build GRCh38
4983555 c.479A>T p.Q160L Chr.12: 75410678-75410678 on Build GRCh38
5616598 c.327C>A p.V109V Chr.12: 75410526-75410526 on Build GRCh38
6671599 c.260C>T p.T87I Chr.12: 75410459-75410459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748365794 Chr.12: 75410504-75410504 on Build GRCh38
rs567116232 Chr.12: 75410388-75410388 on Build GRCh38
rs766112876 Chr.12: 75410387-75410388 on Build GRCh38
rs929284519 Chr.12: 75410741-75410741 on Build GRCh38
rs775030458 Chr.12: 75410750-75410750 on Build GRCh38
rs1055716301 Chr.12: 75410813-75410813 on Build GRCh38
rs763408262 Chr.12: 75410593-75410593 on Build GRCh38
rs538762994 Chr.12: 75410791-75410791 on Build GRCh38
rs200328830 Chr.12: 75410685-75410685 on Build GRCh38
rs776194115 Chr.12: 75410559-75410559 on Build GRCh38
rs760049831 Chr.12: 75410708-75410708 on Build GRCh38
rs748420826 Chr.12: 75410394-75410394 on Build GRCh38
rs202217718 Chr.12: 75410519-75410519 on Build GRCh38
rs998760195 Chr.12: 75410522-75410522 on Build GRCh38
rs114490304 Chr.12: 75410513-75410513 on Build GRCh38
rs530006793 Chr.12: 75410611-75410611 on Build GRCh38
rs1011298091 Chr.12: 75410497-75410497 on Build GRCh38
rs113617304 Chr.12: 75410602-75410602 on Build GRCh38
rs756468887 Chr.12: 75410507-75410507 on Build GRCh38
rs954587676 Chr.12: 75410538-75410538 on Build GRCh38
rs773140893 Chr.12: 75410446-75410446 on Build GRCh38
rs781468774 Chr.12: 75410495-75410495 on Build GRCh38
rs752899425 Chr.12: 75410482-75410482 on Build GRCh38
rs767699458 Chr.12: 75410603-75410603 on Build GRCh38
rs761514357 Chr.12: 75410465-75410465 on Build GRCh38
rs982075012 Chr.12: 75410748-75410748 on Build GRCh38
rs139261658 Chr.12: 75410565-75410565 on Build GRCh38
rs763320380 Chr.12: 75410753-75410753 on Build GRCh38
rs928205152 Chr.12: 75410768-75410768 on Build GRCh38
rs376778122 Chr.12: 75410492-75410492 on Build GRCh38
rs150816381 Chr.12: 75410533-75410533 on Build GRCh38
rs759383991 Chr.12: 75410445-75410445 on Build GRCh38
rs769396644 Chr.12: 75410656-75410656 on Build GRCh38
rs567529302 Chr.12: 75410566-75410566 on Build GRCh38
rs35488316 Chr.12: 75410810-75410811 on Build GRCh38
rs756344523 Chr.12: 75410621-75410621 on Build GRCh38
rs182082343 Chr.12: 75410666-75410666 on Build GRCh38
rs768586832 Chr.12: 75410636-75410636 on Build GRCh38
rs201416567 Chr.12: 75410631-75410631 on Build GRCh38
rs768144760 Chr.12: 75410542-75410542 on Build GRCh38
rs755193276 Chr.12: 75410438-75410438 on Build GRCh38
rs550898972 Chr.12: 75410549-75410549 on Build GRCh38
rs919297916 Chr.12: 75410713-75410713 on Build GRCh38
rs747679947 Chr.12: 75410644-75410644 on Build GRCh38
rs554813469 Chr.12: 75410418-75410418 on Build GRCh38
rs144818804 Chr.12: 75410474-75410474 on Build GRCh38
rs772888636 Chr.12: 75410663-75410663 on Build GRCh38
rs910628947 Chr.12: 75410355-75410355 on Build GRCh38
rs776332299 Chr.12: 75410526-75410526 on Build GRCh38
rs927121367 Chr.12: 75410782-75410782 on Build GRCh38
rs752083422 Chr.12: 75410596-75410596 on Build GRCh38
rs919246104 Chr.12: 75410419-75410419 on Build GRCh38
rs745606358 Chr.12: 75410626-75410626 on Build GRCh38
rs763638046 Chr.12: 75410456-75410456 on Build GRCh38
rs770555542 Chr.12: 75410498-75410498 on Build GRCh38
rs758194642 Chr.12: 75410627-75410627 on Build GRCh38
rs765583629 Chr.12: 75410475-75410475 on Build GRCh38
rs776247147 Chr.12: 75410461-75410461 on Build GRCh38
rs35066273 Chr.12: 75410493-75410493 on Build GRCh38
rs142539769 Chr.12: 75410455-75410455 on Build GRCh38
rs756251948 Chr.12: 75410700-75410701 on Build GRCh38
rs774801554 Chr.12: 75410447-75410447 on Build GRCh38
rs938254371 Chr.12: 75410799-75410799 on Build GRCh38
rs937208768 Chr.12: 75410812-75410812 on Build GRCh38
rs766693613 Chr.12: 75410594-75410594 on Build GRCh38
rs895478486 Chr.12: 75410423-75410423 on Build GRCh38
rs776064193 Chr.12: 75410638-75410638 on Build GRCh38
rs749807454 Chr.12: 75410714-75410716 on Build GRCh38
rs540580232 Chr.12: 75410476-75410476 on Build GRCh38
rs764371673 Chr.12: 75410808-75410808 on Build GRCh38
rs775470257 Chr.12: 75410527-75410527 on Build GRCh38
rs778075111 Chr.12: 75410623-75410623 on Build GRCh38
rs904788137 Chr.12: 75410477-75410477 on Build GRCh38
rs745684554 Chr.12: 75410517-75410517 on Build GRCh38
rs762561811 Chr.12: 75410679-75410679 on Build GRCh38
rs771852210 Chr.12: 75410523-75410523 on Build GRCh38
rs773400560 Chr.12: 75410579-75410579 on Build GRCh38
rs907772390 Chr.12: 75410588-75410588 on Build GRCh38
rs751325058 Chr.12: 75410396-75410397 on Build GRCh38
rs951374965 Chr.12: 75410541-75410541 on Build GRCh38
rs758774592 Chr.12: 75410484-75410484 on Build GRCh38
rs566324981 Chr.12: 75410766-75410766 on Build GRCh38
rs777876028 Chr.12: 75410704-75410705 on Build GRCh38
rs771782553 Chr.12: 75410395-75410395 on Build GRCh38
rs781330948 Chr.12: 75410475-75410475 on Build GRCh38
rs764905137 Chr.12: 75410698-75410698 on Build GRCh38

View Full Product Details