Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75430527-75431023 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIPR1L2
Entrez Gene ID
144321
Gene Name
GLI pathogenesis-related 1 like 2
Gene Aliases
-
UniGene
Hs.406728
Gene Chromosome Location
Chr.12: 75391070-75432688 on Build GRCh38
Transcripts
NM_001270396.1 NR_072995.1 XM_011537949.2 XM_011537950.2 XM_017018848.1 XM_017018849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6237012 c.697+10G>T p.? Chr.12: 75430751-75430751 on Build GRCh38
6520467 c.798G>A p.L266L Chr.12: 75430924-75430924 on Build GRCh38
4147460 c.842T>A p.I281N Chr.12: 75430968-75430968 on Build GRCh38
1628847 c.787T>C p.C263R Chr.12: 75430913-75430913 on Build GRCh38
161279 c.850G>A p.E284K Chr.12: 75430976-75430976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026651532 Chr.12: 75430684-75430684 on Build GRCh38
rs767411520 Chr.12: 75430710-75430710 on Build GRCh38
rs561749961 Chr.12: 75430637-75430637 on Build GRCh38
rs201431673 Chr.12: 75430994-75430994 on Build GRCh38
rs930298845 Chr.12: 75430906-75430906 on Build GRCh38
rs963756292 Chr.12: 75430678-75430678 on Build GRCh38
rs947451256 Chr.12: 75430844-75430844 on Build GRCh38
rs80096121 Chr.12: 75430830-75430830 on Build GRCh38
rs377695461 Chr.12: 75430860-75430860 on Build GRCh38
rs928521194 Chr.12: 75430746-75430746 on Build GRCh38
rs553185397 Chr.12: 75430727-75430727 on Build GRCh38
rs916031657 Chr.12: 75430837-75430837 on Build GRCh38
rs749917015 Chr.12: 75430931-75430931 on Build GRCh38
rs903473099 Chr.12: 75430614-75430614 on Build GRCh38
rs1052046429 Chr.12: 75430922-75430922 on Build GRCh38
rs146685386 Chr.12: 75430574-75430574 on Build GRCh38
rs921816786 Chr.12: 75430681-75430681 on Build GRCh38
rs573089981 Chr.12: 75430777-75430777 on Build GRCh38
rs267603670 Chr.12: 75430961-75430961 on Build GRCh38
rs569202080 Chr.12: 75430556-75430556 on Build GRCh38
rs559068079 Chr.12: 75430592-75430592 on Build GRCh38
rs374194839 Chr.12: 75430695-75430695 on Build GRCh38
rs61746545 Chr.12: 75430973-75430973 on Build GRCh38
rs777772610 Chr.12: 75430679-75430679 on Build GRCh38
rs951083609 Chr.12: 75430726-75430726 on Build GRCh38
rs760715893 Chr.12: 75430784-75430784 on Build GRCh38
rs777552643 Chr.12: 75430996-75430996 on Build GRCh38
rs953282819 Chr.12: 75430724-75430724 on Build GRCh38
rs890688766 Chr.12: 75430933-75430933 on Build GRCh38
rs537954517 Chr.12: 75430569-75430569 on Build GRCh38
rs530664023 Chr.12: 75430642-75430642 on Build GRCh38
rs564751868 Chr.12: 75430835-75430835 on Build GRCh38
rs575130692 Chr.12: 75430913-75430913 on Build GRCh38
rs765843532 Chr.12: 75430829-75430829 on Build GRCh38
rs890646944 Chr.12: 75430630-75430630 on Build GRCh38
rs967435871 Chr.12: 75430613-75430613 on Build GRCh38
rs776493562 Chr.12: 75430859-75430859 on Build GRCh38
rs555062926 Chr.12: 75430591-75430591 on Build GRCh38
rs753301739 Chr.12: 75430950-75430950 on Build GRCh38
rs879475406 Chr.12: 75430953-75430953 on Build GRCh38
rs933335969 Chr.12: 75430643-75430643 on Build GRCh38
rs934632891 Chr.12: 75430899-75430899 on Build GRCh38
rs1033411131 Chr.12: 75430651-75430651 on Build GRCh38
rs761600997 Chr.12: 75430888-75430888 on Build GRCh38
rs771099317 Chr.12: 75430696-75430696 on Build GRCh38
rs999120346 Chr.12: 75430615-75430615 on Build GRCh38
rs920358675 Chr.12: 75430893-75430893 on Build GRCh38
rs759720136 Chr.12: 75430631-75430631 on Build GRCh38
rs924644369 Chr.12: 75430864-75430864 on Build GRCh38
rs1024432863 Chr.12: 75430606-75430606 on Build GRCh38
rs765292544 Chr.12: 75430930-75430930 on Build GRCh38
rs150239550 Chr.12: 75430962-75430962 on Build GRCh38
rs141367832 Chr.12: 75430627-75430627 on Build GRCh38
rs1030967845 Chr.12: 75430629-75430629 on Build GRCh38
rs12833944 Chr.12: 75430680-75430680 on Build GRCh38

View Full Product Details