Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75499165-75499702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIPR1
Entrez Gene ID
11010
Gene Name
GLI pathogenesis related 1
Gene Aliases
CRISP7, GLIPR, RTVP1
UniGene
Hs.205558
Gene Chromosome Location
Chr.12: 75480733-75501936 on Build GRCh38
Transcripts
NM_006851.2 XM_011537775.2 XM_017018721.1 NM_007043.6

Gene Symbol
KRR1
Entrez Gene ID
11103
Gene Name
KRR1, small subunit processome component homolog
Gene Aliases
HRB2, RIP-1
UniGene
Hs.645517
Gene Chromosome Location
Chr.12: 75497639-75511638 on Build GRCh38
Transcripts
NM_006851.2 XM_011537775.2 XM_017018721.1 NM_007043.6

SNP Detail

SNP ID Chromosome Location
rs71684960 Chr.12: 75499602-75499603 on Build GRCh38
rs1056903 Chr.12: 75499624-75499624 on Build GRCh38
rs927045377 Chr.12: 75499402-75499402 on Build GRCh38
rs191349641 Chr.12: 75499417-75499417 on Build GRCh38
rs569567014 Chr.12: 75499350-75499350 on Build GRCh38
rs1009454182 Chr.12: 75499310-75499310 on Build GRCh38
rs575059547 Chr.12: 75499482-75499482 on Build GRCh38
rs1020279533 Chr.12: 75499207-75499207 on Build GRCh38
rs985273515 Chr.12: 75499336-75499336 on Build GRCh38
rs949756154 Chr.12: 75499388-75499388 on Build GRCh38
rs538747415 Chr.12: 75499406-75499406 on Build GRCh38
rs774307499 Chr.12: 75499393-75499393 on Build GRCh38
rs866195016 Chr.12: 75499424-75499424 on Build GRCh38
rs1056904 Chr.12: 75499619-75499619 on Build GRCh38
rs560837835 Chr.12: 75499197-75499197 on Build GRCh38
rs567481331 Chr.12: 75499307-75499307 on Build GRCh38
rs938715650 Chr.12: 75499261-75499261 on Build GRCh38
rs901873918 Chr.12: 75499231-75499231 on Build GRCh38
rs1056304910 Chr.12: 75499280-75499288 on Build GRCh38
rs903679463 Chr.12: 75499590-75499590 on Build GRCh38
rs573084373 Chr.12: 75499381-75499381 on Build GRCh38
rs1056905 Chr.12: 75499608-75499608 on Build GRCh38
rs187259255 Chr.12: 75499392-75499392 on Build GRCh38
rs1044509466 Chr.12: 75499238-75499238 on Build GRCh38
rs1043929114 Chr.12: 75499312-75499314 on Build GRCh38
rs747974877 Chr.12: 75499536-75499536 on Build GRCh38
rs962599629 Chr.12: 75499345-75499345 on Build GRCh38
rs547444282 Chr.12: 75499188-75499188 on Build GRCh38
rs529451464 Chr.12: 75499234-75499234 on Build GRCh38
rs530409265 Chr.12: 75499500-75499501 on Build GRCh38
rs1006870682 Chr.12: 75499648-75499648 on Build GRCh38
rs569457249 Chr.12: 75499431-75499431 on Build GRCh38
rs182562901 Chr.12: 75499370-75499370 on Build GRCh38
rs953856438 Chr.12: 75499323-75499323 on Build GRCh38
rs973095402 Chr.12: 75499348-75499348 on Build GRCh38
rs889185938 Chr.12: 75499564-75499564 on Build GRCh38
rs148145944 Chr.12: 75499529-75499529 on Build GRCh38
rs901803221 Chr.12: 75499473-75499473 on Build GRCh38
rs775574306 Chr.12: 75499429-75499430 on Build GRCh38
rs1029048618 Chr.12: 75499516-75499516 on Build GRCh38
rs893346652 Chr.12: 75499416-75499416 on Build GRCh38
rs1006300865 Chr.12: 75499630-75499630 on Build GRCh38
rs1025960658 Chr.12: 75499625-75499625 on Build GRCh38
rs539705148 Chr.12: 75499637-75499638 on Build GRCh38
rs909394068 Chr.12: 75499217-75499217 on Build GRCh38
rs182426391 Chr.12: 75499476-75499476 on Build GRCh38
rs191618831 Chr.12: 75499337-75499337 on Build GRCh38
rs553792124 Chr.12: 75499515-75499515 on Build GRCh38
rs143054392 Chr.12: 75499447-75499447 on Build GRCh38
rs965700079 Chr.12: 75499352-75499353 on Build GRCh38
rs540524025 Chr.12: 75499513-75499513 on Build GRCh38
rs186780388 Chr.12: 75499287-75499287 on Build GRCh38
rs549603090 Chr.12: 75499277-75499277 on Build GRCh38
rs553157171 Chr.12: 75499281-75499282 on Build GRCh38
rs781625494 Chr.12: 75499254-75499254 on Build GRCh38
rs749056490 Chr.12: 75499255-75499255 on Build GRCh38
rs1054607752 Chr.12: 75499407-75499407 on Build GRCh38
rs1025994913 Chr.12: 75499375-75499376 on Build GRCh38
rs1043678932 Chr.12: 75499571-75499571 on Build GRCh38
rs74986206 Chr.12: 75499257-75499257 on Build GRCh38
rs768527336 Chr.12: 75499362-75499362 on Build GRCh38
rs1029493487 Chr.12: 75499291-75499291 on Build GRCh38
rs868334604 Chr.12: 75499383-75499383 on Build GRCh38
rs35542836 Chr.12: 75499281-75499281 on Build GRCh38

View Full Product Details