Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 75499823-75500323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIPR1
Entrez Gene ID
11010
Gene Name
GLI pathogenesis related 1
Gene Aliases
CRISP7, GLIPR, RTVP1
UniGene
Hs.205558
Gene Chromosome Location
Chr.12: 75480733-75501936 on Build GRCh38
Transcripts
NM_006851.2 XM_011537775.2 XM_017018721.1 NM_007043.6

Gene Symbol
KRR1
Entrez Gene ID
11103
Gene Name
KRR1, small subunit processome component homolog
Gene Aliases
HRB2, RIP-1
UniGene
Hs.645517
Gene Chromosome Location
Chr.12: 75497639-75511638 on Build GRCh38
Transcripts
NM_006851.2 XM_011537775.2 XM_017018721.1 NM_007043.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7413896 c.1046A>G p.K349R Chr.12: 75499909-75499909 on Build GRCh38
3688444 c.1128A>C p.K376N Chr.12: 75499827-75499827 on Build GRCh38
5807521 c.1096A>G p.I366V Chr.12: 75499859-75499859 on Build GRCh38
1364228 c.1121A>G p.E374G Chr.12: 75499834-75499834 on Build GRCh38
6569549 c.1131G>A p.K377K Chr.12: 75499824-75499824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1697754 Chr.12: 75500085-75500085 on Build GRCh38
rs766222607 Chr.12: 75499877-75499877 on Build GRCh38
rs933038188 Chr.12: 75499891-75499891 on Build GRCh38
rs752622359 Chr.12: 75499888-75499890 on Build GRCh38
rs111923420 Chr.12: 75499977-75499977 on Build GRCh38
rs974679910 Chr.12: 75500205-75500205 on Build GRCh38
rs757680953 Chr.12: 75499943-75499943 on Build GRCh38
rs933560527 Chr.12: 75500169-75500169 on Build GRCh38
rs755783070 Chr.12: 75499913-75499913 on Build GRCh38
rs967544635 Chr.12: 75500070-75500070 on Build GRCh38
rs1011261153 Chr.12: 75500286-75500286 on Build GRCh38
rs779398848 Chr.12: 75499948-75499948 on Build GRCh38
rs762092968 Chr.12: 75499988-75499988 on Build GRCh38
rs776824171 Chr.12: 75499982-75499982 on Build GRCh38
rs772657477 Chr.12: 75499953-75499953 on Build GRCh38
rs772093516 Chr.12: 75499979-75499980 on Build GRCh38
rs942053798 Chr.12: 75500201-75500201 on Build GRCh38
rs993618496 Chr.12: 75500213-75500213 on Build GRCh38
rs554203139 Chr.12: 75500142-75500142 on Build GRCh38
rs7486195 Chr.12: 75500288-75500288 on Build GRCh38
rs188755117 Chr.12: 75500107-75500107 on Build GRCh38
rs969210680 Chr.12: 75500134-75500134 on Build GRCh38
rs185582999 Chr.12: 75500255-75500255 on Build GRCh38
rs376672311 Chr.12: 75499951-75499951 on Build GRCh38
rs1050368149 Chr.12: 75499857-75499857 on Build GRCh38
rs762715577 Chr.12: 75499874-75499874 on Build GRCh38
rs911227758 Chr.12: 75500195-75500195 on Build GRCh38
rs1037720127 Chr.12: 75500202-75500202 on Build GRCh38
rs77063640 Chr.12: 75499908-75499908 on Build GRCh38
rs1055804565 Chr.12: 75500272-75500272 on Build GRCh38
rs751449948 Chr.12: 75499893-75499893 on Build GRCh38
rs1035803960 Chr.12: 75500058-75500058 on Build GRCh38
rs759218744 Chr.12: 75500176-75500176 on Build GRCh38
rs1050643790 Chr.12: 75500171-75500171 on Build GRCh38
rs914343859 Chr.12: 75500051-75500051 on Build GRCh38
rs886148076 Chr.12: 75500240-75500240 on Build GRCh38
rs1034011262 Chr.12: 75500023-75500023 on Build GRCh38
rs958338096 Chr.12: 75500266-75500266 on Build GRCh38
rs546874876 Chr.12: 75499899-75499899 on Build GRCh38
rs757566131 Chr.12: 75499953-75499954 on Build GRCh38
rs958369385 Chr.12: 75500029-75500029 on Build GRCh38
rs765538704 Chr.12: 75500241-75500241 on Build GRCh38
rs943278030 Chr.12: 75500203-75500203 on Build GRCh38
rs751910697 Chr.12: 75500275-75500276 on Build GRCh38
rs566245480 Chr.12: 75499888-75499888 on Build GRCh38
rs375938543 Chr.12: 75499973-75499973 on Build GRCh38
rs372067400 Chr.12: 75499861-75499861 on Build GRCh38
rs746292748 Chr.12: 75499950-75499950 on Build GRCh38
rs1002759213 Chr.12: 75500249-75500249 on Build GRCh38
rs372351965 Chr.12: 75499929-75499929 on Build GRCh38
rs776306396 Chr.12: 75500115-75500115 on Build GRCh38
rs760760484 Chr.12: 75499870-75499870 on Build GRCh38
rs552317226 Chr.12: 75500042-75500042 on Build GRCh38
rs775643670 Chr.12: 75499853-75499853 on Build GRCh38
rs768774833 Chr.12: 75499975-75499975 on Build GRCh38
rs770649959 Chr.12: 75500001-75500001 on Build GRCh38
rs193205310 Chr.12: 75500243-75500243 on Build GRCh38
rs112185628 Chr.12: 75499987-75499987 on Build GRCh38
rs779958589 Chr.12: 75499954-75499954 on Build GRCh38
rs769087617 Chr.12: 75500004-75500021 on Build GRCh38
rs568631196 Chr.12: 75500207-75500207 on Build GRCh38
rs772040272 Chr.12: 75499852-75499852 on Build GRCh38
rs777739602 Chr.12: 75499952-75499953 on Build GRCh38
rs554912924 Chr.12: 75500185-75500185 on Build GRCh38
rs150361984 Chr.12: 75499896-75499896 on Build GRCh38
rs573051952 Chr.12: 75499981-75499982 on Build GRCh38
rs746072165 Chr.12: 75499976-75499977 on Build GRCh38
rs374297882 Chr.12: 75499900-75499900 on Build GRCh38
rs910512307 Chr.12: 75500189-75500189 on Build GRCh38
rs923086713 Chr.12: 75500208-75500208 on Build GRCh38
rs767019043 Chr.12: 75499870-75499871 on Build GRCh38
rs1002205769 Chr.12: 75500011-75500011 on Build GRCh38
rs747547186 Chr.12: 75500002-75500003 on Build GRCh38
rs1002833820 Chr.12: 75500015-75500015 on Build GRCh38
rs17853238 Chr.12: 75499940-75499940 on Build GRCh38
rs756063565 Chr.12: 75499890-75499891 on Build GRCh38
rs753590345 Chr.12: 75499934-75499934 on Build GRCh38
rs1047509951 Chr.12: 75500234-75500234 on Build GRCh38
rs149114650 Chr.12: 75499920-75499920 on Build GRCh38
rs978752814 Chr.12: 75500193-75500193 on Build GRCh38

View Full Product Details