Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 76860201-76860711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRP2
Entrez Gene ID
1466
Gene Name
cysteine and glycine rich protein 2
Gene Aliases
CRP2, LMO5, SmLIM
UniGene
Hs.530904
Gene Chromosome Location
Chr.12: 76858715-76879040 on Build GRCh38
Transcripts
NM_001300965.1 NM_001321.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3813025 c.286C>T p.Q96* Chr.12: 76860409-76860409 on Build GRCh38
3955162 c.388G>T p.E130* Chr.12: 76860307-76860307 on Build GRCh38
3465543 c.411+2T>A p.? Chr.12: 76860282-76860282 on Build GRCh38
4044897 c.411+2T>C p.? Chr.12: 76860282-76860282 on Build GRCh38
7174019 c.347C>A p.A116D Chr.12: 76860348-76860348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202134808 Chr.12: 76860444-76860444 on Build GRCh38
rs1013617484 Chr.12: 76860547-76860547 on Build GRCh38
rs528484905 Chr.12: 76860519-76860519 on Build GRCh38
rs1026593610 Chr.12: 76860610-76860610 on Build GRCh38
rs902419714 Chr.12: 76860484-76860484 on Build GRCh38
rs372796776 Chr.12: 76860465-76860466 on Build GRCh38
rs775601635 Chr.12: 76860429-76860429 on Build GRCh38
rs754382117 Chr.12: 76860548-76860549 on Build GRCh38
rs757021263 Chr.12: 76860344-76860344 on Build GRCh38
rs752307470 Chr.12: 76860451-76860451 on Build GRCh38
rs774700075 Chr.12: 76860326-76860326 on Build GRCh38
rs779627598 Chr.12: 76860284-76860284 on Build GRCh38
rs540993317 Chr.12: 76860471-76860471 on Build GRCh38
rs531884247 Chr.12: 76860663-76860663 on Build GRCh38
rs1036633215 Chr.12: 76860497-76860497 on Build GRCh38
rs981813793 Chr.12: 76860417-76860417 on Build GRCh38
rs398020212 Chr.12: 76860436-76860436 on Build GRCh38
rs779609171 Chr.12: 76860378-76860378 on Build GRCh38
rs532509477 Chr.12: 76860517-76860517 on Build GRCh38
rs972015918 Chr.12: 76860557-76860557 on Build GRCh38
rs377118692 Chr.12: 76860392-76860392 on Build GRCh38
rs778260697 Chr.12: 76860269-76860269 on Build GRCh38
rs896925399 Chr.12: 76860498-76860498 on Build GRCh38
rs761600446 Chr.12: 76860431-76860431 on Build GRCh38
rs373092557 Chr.12: 76860445-76860445 on Build GRCh38
rs750329086 Chr.12: 76860367-76860367 on Build GRCh38
rs748581903 Chr.12: 76860308-76860308 on Build GRCh38
rs201242706 Chr.12: 76860428-76860428 on Build GRCh38
rs369229551 Chr.12: 76860244-76860244 on Build GRCh38
rs754514128 Chr.12: 76860447-76860447 on Build GRCh38
rs950891288 Chr.12: 76860356-76860356 on Build GRCh38
rs997992108 Chr.12: 76860506-76860506 on Build GRCh38
rs754678842 Chr.12: 76860385-76860385 on Build GRCh38
rs548437177 Chr.12: 76860560-76860560 on Build GRCh38
rs750373212 Chr.12: 76860438-76860438 on Build GRCh38
rs928084080 Chr.12: 76860650-76860650 on Build GRCh38
rs770287082 Chr.12: 76860309-76860309 on Build GRCh38
rs765179427 Chr.12: 76860363-76860363 on Build GRCh38
rs778696932 Chr.12: 76860425-76860426 on Build GRCh38
rs1007602459 Chr.12: 76860530-76860530 on Build GRCh38
rs777024886 Chr.12: 76860307-76860307 on Build GRCh38
rs762922439 Chr.12: 76860443-76860443 on Build GRCh38
rs1050854919 Chr.12: 76860509-76860509 on Build GRCh38
rs1023428580 Chr.12: 76860364-76860364 on Build GRCh38
rs562967601 Chr.12: 76860662-76860662 on Build GRCh38
rs878981650 Chr.12: 76860590-76860590 on Build GRCh38
rs773625473 Chr.12: 76860313-76860313 on Build GRCh38
rs770061489 Chr.12: 76860393-76860393 on Build GRCh38
rs1002892068 Chr.12: 76860588-76860588 on Build GRCh38
rs780832131 Chr.12: 76860448-76860448 on Build GRCh38
rs914967443 Chr.12: 76860688-76860688 on Build GRCh38
rs771677513 Chr.12: 76860427-76860428 on Build GRCh38
rs763025095 Chr.12: 76860314-76860314 on Build GRCh38
rs535336517 Chr.12: 76860549-76860549 on Build GRCh38
rs374780862 Chr.12: 76860241-76860241 on Build GRCh38
rs990215448 Chr.12: 76860601-76860601 on Build GRCh38
rs12580988 Chr.12: 76860472-76860472 on Build GRCh38
rs745329477 Chr.12: 76860273-76860273 on Build GRCh38
rs959387030 Chr.12: 76860665-76860665 on Build GRCh38
rs766670335 Chr.12: 76860236-76860236 on Build GRCh38
rs548750115 Chr.12: 76860673-76860673 on Build GRCh38
rs759809089 Chr.12: 76860425-76860425 on Build GRCh38
rs775550090 Chr.12: 76860428-76860429 on Build GRCh38
rs180731353 Chr.12: 76860297-76860297 on Build GRCh38
rs373572345 Chr.12: 76860317-76860317 on Build GRCh38
rs759904407 Chr.12: 76860327-76860327 on Build GRCh38
rs914874200 Chr.12: 76860441-76860441 on Build GRCh38
rs769008153 Chr.12: 76860294-76860294 on Build GRCh38
rs995235946 Chr.12: 76860597-76860597 on Build GRCh38
rs757258105 Chr.12: 76860265-76860265 on Build GRCh38
rs35557984 Chr.12: 76860428-76860428 on Build GRCh38
rs111570084 Chr.12: 76860245-76860245 on Build GRCh38
rs937748359 Chr.12: 76860434-76860434 on Build GRCh38
rs376447473 Chr.12: 76860640-76860640 on Build GRCh38
rs757323816 Chr.12: 76860561-76860563 on Build GRCh38
rs369517609 Chr.12: 76860407-76860407 on Build GRCh38
rs763949587 Chr.12: 76860336-76860336 on Build GRCh38
rs963442881 Chr.12: 76860555-76860555 on Build GRCh38
rs141953524 Chr.12: 76860633-76860633 on Build GRCh38
rs780731467 Chr.12: 76860388-76860388 on Build GRCh38
rs746637559 Chr.12: 76860383-76860383 on Build GRCh38
rs370222934 Chr.12: 76860552-76860552 on Build GRCh38
rs746725152 Chr.12: 76860292-76860292 on Build GRCh38
rs1055207695 Chr.12: 76860435-76860435 on Build GRCh38
rs995206126 Chr.12: 76860505-76860505 on Build GRCh38
rs757931668 Chr.12: 76860276-76860276 on Build GRCh38
rs148553046 Chr.12: 76860514-76860514 on Build GRCh38
rs757894632 Chr.12: 76860373-76860373 on Build GRCh38
rs914446828 Chr.12: 76860608-76860608 on Build GRCh38
rs756486114 Chr.12: 76860454-76860454 on Build GRCh38
rs1018100152 Chr.12: 76860531-76860531 on Build GRCh38
rs753715128 Chr.12: 76860250-76860250 on Build GRCh38
rs990624271 Chr.12: 76860671-76860671 on Build GRCh38
rs753712264 Chr.12: 76860339-76860339 on Build GRCh38
rs200345176 Chr.12: 76860436-76860436 on Build GRCh38
rs950923635 Chr.12: 76860617-76860617 on Build GRCh38
rs374611670 Chr.12: 76860414-76860414 on Build GRCh38
rs778064378 Chr.12: 76860459-76860459 on Build GRCh38
rs201480928 Chr.12: 76860422-76860422 on Build GRCh38

View Full Product Details