Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 76866058-76866565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRP2
Entrez Gene ID
1466
Gene Name
cysteine and glycine rich protein 2
Gene Aliases
CRP2, LMO5, SmLIM
UniGene
Hs.530904
Gene Chromosome Location
Chr.12: 76858715-76879040 on Build GRCh38
Transcripts
NM_001300965.1 NM_001321.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
280190 c.95G>A p.R32H Chr.12: 76866166-76866166 on Build GRCh38
6636960 c.49G>A p.V17M Chr.12: 76866212-76866212 on Build GRCh38
6860479 c.47C>A p.T16N Chr.12: 76866214-76866214 on Build GRCh38
468914 c.75T>G p.C25W Chr.12: 76866186-76866186 on Build GRCh38
243892 c.57C>T p.H19H Chr.12: 76866204-76866204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756376429 Chr.12: 76866127-76866127 on Build GRCh38
rs768274309 Chr.12: 76866137-76866137 on Build GRCh38
rs117324885 Chr.12: 76866276-76866276 on Build GRCh38
rs374755270 Chr.12: 76866134-76866134 on Build GRCh38
rs398076763 Chr.12: 76866483-76866483 on Build GRCh38
rs375897146 Chr.12: 76866271-76866271 on Build GRCh38
rs111923859 Chr.12: 76866204-76866204 on Build GRCh38
rs562029782 Chr.12: 76866381-76866381 on Build GRCh38
rs946542028 Chr.12: 76866384-76866384 on Build GRCh38
rs973543817 Chr.12: 76866514-76866514 on Build GRCh38
rs530001479 Chr.12: 76866464-76866464 on Build GRCh38
rs746887392 Chr.12: 76866249-76866249 on Build GRCh38
rs567930922 Chr.12: 76866228-76866228 on Build GRCh38
rs534915506 Chr.12: 76866492-76866492 on Build GRCh38
rs760686315 Chr.12: 76866308-76866308 on Build GRCh38
rs1017856705 Chr.12: 76866484-76866484 on Build GRCh38
rs576261391 Chr.12: 76866359-76866359 on Build GRCh38
rs753542236 Chr.12: 76866265-76866265 on Build GRCh38
rs1030706399 Chr.12: 76866444-76866444 on Build GRCh38
rs766321037 Chr.12: 76866117-76866117 on Build GRCh38
rs34861040 Chr.12: 76866473-76866473 on Build GRCh38
rs868727730 Chr.12: 76866128-76866128 on Build GRCh38
rs952201878 Chr.12: 76866456-76866456 on Build GRCh38
rs903244567 Chr.12: 76866436-76866436 on Build GRCh38
rs769442824 Chr.12: 76866282-76866282 on Build GRCh38
rs540633349 Chr.12: 76866515-76866515 on Build GRCh38
rs759818561 Chr.12: 76866502-76866502 on Build GRCh38
rs1050593867 Chr.12: 76866277-76866277 on Build GRCh38
rs981246438 Chr.12: 76866472-76866472 on Build GRCh38
rs772727866 Chr.12: 76866283-76866283 on Build GRCh38
rs756063873 Chr.12: 76866185-76866185 on Build GRCh38
rs748336192 Chr.12: 76866126-76866126 on Build GRCh38
rs747470317 Chr.12: 76866397-76866397 on Build GRCh38
rs34508717 Chr.12: 76866093-76866094 on Build GRCh38
rs745610205 Chr.12: 76866131-76866131 on Build GRCh38
rs771322686 Chr.12: 76866453-76866453 on Build GRCh38
rs141337985 Chr.12: 76866354-76866354 on Build GRCh38
rs781318495 Chr.12: 76866108-76866108 on Build GRCh38
rs762734581 Chr.12: 76866291-76866291 on Build GRCh38
rs752723566 Chr.12: 76866213-76866213 on Build GRCh38
rs890456694 Chr.12: 76866463-76866463 on Build GRCh38
rs750317087 Chr.12: 76866105-76866105 on Build GRCh38
rs763169571 Chr.12: 76866189-76866189 on Build GRCh38
rs200827875 Chr.12: 76866473-76866473 on Build GRCh38
rs756217081 Chr.12: 76866221-76866221 on Build GRCh38
rs993541590 Chr.12: 76866393-76866393 on Build GRCh38
rs754100473 Chr.12: 76866231-76866231 on Build GRCh38
rs777992048 Chr.12: 76866130-76866130 on Build GRCh38
rs777237011 Chr.12: 76866460-76866460 on Build GRCh38
rs771901625 Chr.12: 76866133-76866133 on Build GRCh38
rs372179006 Chr.12: 76866168-76866168 on Build GRCh38
rs769534944 Chr.12: 76866167-76866167 on Build GRCh38
rs79414552 Chr.12: 76866344-76866344 on Build GRCh38
rs779962540 Chr.12: 76866233-76866233 on Build GRCh38
rs372155706 Chr.12: 76866307-76866307 on Build GRCh38
rs541728106 Chr.12: 76866380-76866380 on Build GRCh38
rs200668191 Chr.12: 76866107-76866107 on Build GRCh38
rs1011657070 Chr.12: 76866103-76866103 on Build GRCh38
rs747021861 Chr.12: 76866135-76866135 on Build GRCh38
rs780086006 Chr.12: 76866262-76866262 on Build GRCh38
rs1054963625 Chr.12: 76866373-76866373 on Build GRCh38
rs778414179 Chr.12: 76866297-76866297 on Build GRCh38
rs539186908 Chr.12: 76866086-76866086 on Build GRCh38
rs767971178 Chr.12: 76866303-76866303 on Build GRCh38
rs1045684149 Chr.12: 76866166-76866166 on Build GRCh38
rs767561672 Chr.12: 76866210-76866210 on Build GRCh38
rs771171428 Chr.12: 76866293-76866293 on Build GRCh38
rs542590119 Chr.12: 76866500-76866500 on Build GRCh38
rs998873376 Chr.12: 76866437-76866437 on Build GRCh38
rs183831969 Chr.12: 76866119-76866119 on Build GRCh38
rs768668263 Chr.12: 76866251-76866251 on Build GRCh38
rs144564754 Chr.12: 76866163-76866163 on Build GRCh38
rs762547143 Chr.12: 76866304-76866304 on Build GRCh38
rs766825277 Chr.12: 76866203-76866203 on Build GRCh38
rs368852370 Chr.12: 76866148-76866148 on Build GRCh38
rs759915566 Chr.12: 76866301-76866301 on Build GRCh38
rs758112327 Chr.12: 76866232-76866232 on Build GRCh38
rs982156122 Chr.12: 76866309-76866309 on Build GRCh38

View Full Product Details