Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119472096-119472564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC60
Entrez Gene ID
160777
Gene Name
coiled-coil domain containing 60
Gene Aliases
-
UniGene
Hs.98188
Gene Chromosome Location
Chr.12: 119334712-119541047 on Build GRCh38
Transcripts
NM_178499.4 XM_017018911.1 XM_017018912.1 XM_017018913.1

Gene Symbol
LOC105370027
Entrez Gene ID
105370027
Gene Name
uncharacterized LOC105370027
Gene Aliases
-
UniGene
Hs.720596
Gene Chromosome Location
Chr.12: 119387991-119668006 on Build GRCh38
Transcripts
NM_178499.4 XM_017018911.1 XM_017018912.1 XM_017018913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5893631 c.319C>T p.H107Y Chr.12: 119472142-119472142 on Build GRCh38
3968015 c.322T>C p.Y108H Chr.12: 119472145-119472145 on Build GRCh38
4540041 c.277G>A p.E93K Chr.12: 119472100-119472100 on Build GRCh38
5374338 c.281G>A p.R94K Chr.12: 119472104-119472104 on Build GRCh38
4887006 c.339G>T p.L113F Chr.12: 119472162-119472162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375133552 Chr.12: 119472170-119472170 on Build GRCh38
rs989219377 Chr.12: 119472390-119472390 on Build GRCh38
rs369423144 Chr.12: 119472179-119472179 on Build GRCh38
rs760451650 Chr.12: 119472188-119472188 on Build GRCh38
rs550452937 Chr.12: 119472266-119472266 on Build GRCh38
rs770291473 Chr.12: 119472151-119472151 on Build GRCh38
rs139079166 Chr.12: 119472160-119472160 on Build GRCh38
rs745410813 Chr.12: 119472126-119472126 on Build GRCh38
rs919332505 Chr.12: 119472513-119472513 on Build GRCh38
rs1026382891 Chr.12: 119472301-119472301 on Build GRCh38
rs190509203 Chr.12: 119472349-119472349 on Build GRCh38
rs202167769 Chr.12: 119472140-119472140 on Build GRCh38
rs969594657 Chr.12: 119472440-119472440 on Build GRCh38
rs753492770 Chr.12: 119472123-119472123 on Build GRCh38
rs181813532 Chr.12: 119472399-119472399 on Build GRCh38
rs114931879 Chr.12: 119472308-119472308 on Build GRCh38
rs1014234561 Chr.12: 119472314-119472314 on Build GRCh38
rs1036450897 Chr.12: 119472491-119472491 on Build GRCh38
rs748544163 Chr.12: 119472150-119472150 on Build GRCh38
rs998615234 Chr.12: 119472354-119472354 on Build GRCh38
rs781609413 Chr.12: 119472146-119472146 on Build GRCh38
rs534807331 Chr.12: 119472398-119472398 on Build GRCh38
rs899015134 Chr.12: 119472505-119472505 on Build GRCh38
rs530357507 Chr.12: 119472232-119472232 on Build GRCh38
rs536852252 Chr.12: 119472435-119472435 on Build GRCh38
rs968347846 Chr.12: 119472345-119472345 on Build GRCh38
rs997717638 Chr.12: 119472192-119472192 on Build GRCh38
rs911823524 Chr.12: 119472457-119472457 on Build GRCh38
rs980966098 Chr.12: 119472448-119472448 on Build GRCh38
rs980302028 Chr.12: 119472340-119472340 on Build GRCh38
rs563972210 Chr.12: 119472292-119472292 on Build GRCh38
rs1031799967 Chr.12: 119472381-119472381 on Build GRCh38
rs778425978 Chr.12: 119472124-119472124 on Build GRCh38
rs889022309 Chr.12: 119472243-119472243 on Build GRCh38
rs763889437 Chr.12: 119472209-119472209 on Build GRCh38
rs753449542 Chr.12: 119472212-119472212 on Build GRCh38
rs185701392 Chr.12: 119472330-119472330 on Build GRCh38
rs930448146 Chr.12: 119472531-119472531 on Build GRCh38
rs545485333 Chr.12: 119472471-119472471 on Build GRCh38
rs17482508 Chr.12: 119472156-119472156 on Build GRCh38
rs776532827 Chr.12: 119472464-119472464 on Build GRCh38
rs185900528 Chr.12: 119472437-119472437 on Build GRCh38
rs933201029 Chr.12: 119472536-119472536 on Build GRCh38
rs559574757 Chr.12: 119472155-119472155 on Build GRCh38
rs956934228 Chr.12: 119472386-119472386 on Build GRCh38
rs1051723229 Chr.12: 119472228-119472228 on Build GRCh38
rs568123958 Chr.12: 119472406-119472406 on Build GRCh38
rs773514451 Chr.12: 119472155-119472155 on Build GRCh38
rs1024577718 Chr.12: 119472327-119472327 on Build GRCh38
rs945065110 Chr.12: 119472425-119472425 on Build GRCh38
rs767474920 Chr.12: 119472176-119472176 on Build GRCh38
rs267603333 Chr.12: 119472129-119472129 on Build GRCh38
rs771203945 Chr.12: 119472504-119472504 on Build GRCh38
rs756698907 Chr.12: 119472148-119472148 on Build GRCh38

View Full Product Details