Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120528860-120529367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ5
Entrez Gene ID
84274
Gene Name
coenzyme Q5, methyltransferase
Gene Aliases
-
UniGene
Hs.17250
Gene Chromosome Location
Chr.12: 120503279-120529171 on Build GRCh38
Transcripts
NM_032314.3 XM_006719639.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3954287 c.64C>T p.R22W Chr.12: 120529078-120529078 on Build GRCh38
5950983 c.64C>A p.R22R Chr.12: 120529078-120529078 on Build GRCh38
5926822 c.14G>A p.G5E Chr.12: 120529128-120529128 on Build GRCh38
5889200 c.13G>A p.G5R Chr.12: 120529129-120529129 on Build GRCh38
3752963 c.24T>C p.A8A Chr.12: 120529118-120529118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879756420 Chr.12: 120529117-120529118 on Build GRCh38
rs781518428 Chr.12: 120528910-120528910 on Build GRCh38
rs536951046 Chr.12: 120529228-120529228 on Build GRCh38
rs149964634 Chr.12: 120529075-120529075 on Build GRCh38
rs777679148 Chr.12: 120528931-120528931 on Build GRCh38
rs754026401 Chr.12: 120529108-120529108 on Build GRCh38
rs962209633 Chr.12: 120529221-120529221 on Build GRCh38
rs566538977 Chr.12: 120529212-120529212 on Build GRCh38
rs779911285 Chr.12: 120529119-120529119 on Build GRCh38
rs758933930 Chr.12: 120528897-120528897 on Build GRCh38
rs751912676 Chr.12: 120528905-120528905 on Build GRCh38
rs776213694 Chr.12: 120528987-120528987 on Build GRCh38
rs759921272 Chr.12: 120529095-120529095 on Build GRCh38
rs984958907 Chr.12: 120529009-120529009 on Build GRCh38
rs201359580 Chr.12: 120529050-120529050 on Build GRCh38
rs553059235 Chr.12: 120529294-120529294 on Build GRCh38
rs1021251919 Chr.12: 120528892-120528892 on Build GRCh38
rs539154804 Chr.12: 120529284-120529284 on Build GRCh38
rs972287695 Chr.12: 120529229-120529229 on Build GRCh38
rs978483899 Chr.12: 120529341-120529341 on Build GRCh38
rs11611657 Chr.12: 120529163-120529163 on Build GRCh38
rs911210547 Chr.12: 120529218-120529218 on Build GRCh38
rs935486038 Chr.12: 120529116-120529116 on Build GRCh38
rs567984672 Chr.12: 120529225-120529225 on Build GRCh38
rs745726486 Chr.12: 120528953-120528953 on Build GRCh38
rs376339245 Chr.12: 120529039-120529039 on Build GRCh38
rs777925972 Chr.12: 120529030-120529030 on Build GRCh38
rs747756052 Chr.12: 120529125-120529125 on Build GRCh38
rs768491516 Chr.12: 120529120-120529120 on Build GRCh38
rs752949564 Chr.12: 120529100-120529100 on Build GRCh38
rs915595841 Chr.12: 120529275-120529275 on Build GRCh38
rs374127460 Chr.12: 120529033-120529033 on Build GRCh38
rs752838666 Chr.12: 120528933-120528934 on Build GRCh38
rs754079527 Chr.12: 120529137-120529137 on Build GRCh38
rs756490285 Chr.12: 120529105-120529105 on Build GRCh38
rs577006250 Chr.12: 120529249-120529249 on Build GRCh38
rs755392332 Chr.12: 120528907-120528907 on Build GRCh38
rs758489676 Chr.12: 120529162-120529162 on Build GRCh38
rs199752578 Chr.12: 120529042-120529042 on Build GRCh38
rs778859227 Chr.12: 120529040-120529040 on Build GRCh38
rs763567310 Chr.12: 120528996-120528996 on Build GRCh38
rs563912555 Chr.12: 120529132-120529132 on Build GRCh38
rs997985890 Chr.12: 120528949-120528949 on Build GRCh38
rs935588284 Chr.12: 120529287-120529287 on Build GRCh38
rs780569337 Chr.12: 120528901-120528901 on Build GRCh38
rs922857401 Chr.12: 120529285-120529285 on Build GRCh38
rs776407019 Chr.12: 120529081-120529081 on Build GRCh38
rs771729666 Chr.12: 120528963-120528963 on Build GRCh38
rs774261471 Chr.12: 120528941-120528941 on Build GRCh38
rs771920119 Chr.12: 120529047-120529047 on Build GRCh38
rs752994991 Chr.12: 120529021-120529021 on Build GRCh38
rs548192574 Chr.12: 120529186-120529186 on Build GRCh38
rs756226742 Chr.12: 120529010-120529011 on Build GRCh38
rs369310354 Chr.12: 120529092-120529092 on Build GRCh38
rs764369993 Chr.12: 120529107-120529107 on Build GRCh38
rs1029432880 Chr.12: 120529153-120529153 on Build GRCh38
rs964071757 Chr.12: 120529251-120529251 on Build GRCh38
rs770800882 Chr.12: 120528938-120528938 on Build GRCh38
rs748297523 Chr.12: 120528914-120528914 on Build GRCh38
rs1041345943 Chr.12: 120529102-120529102 on Build GRCh38
rs759279144 Chr.12: 120529067-120529067 on Build GRCh38
rs764373282 Chr.12: 120529061-120529062 on Build GRCh38
rs369028063 Chr.12: 120528926-120528926 on Build GRCh38
rs371704051 Chr.12: 120529134-120529134 on Build GRCh38
rs764818558 Chr.12: 120529091-120529091 on Build GRCh38
rs139585780 Chr.12: 120529006-120529006 on Build GRCh38
rs760106234 Chr.12: 120529130-120529130 on Build GRCh38
rs754256121 Chr.12: 120529274-120529274 on Build GRCh38
rs368099889 Chr.12: 120529115-120529115 on Build GRCh38
rs763690638 Chr.12: 120528986-120528986 on Build GRCh38
rs761450884 Chr.12: 120529087-120529087 on Build GRCh38
rs527630225 Chr.12: 120529129-120529129 on Build GRCh38
rs1052605872 Chr.12: 120529303-120529303 on Build GRCh38
rs760988052 Chr.12: 120529133-120529133 on Build GRCh38
rs528137911 Chr.12: 120529011-120529011 on Build GRCh38
rs193157720 Chr.12: 120529146-120529146 on Build GRCh38
rs781094775 Chr.12: 120529180-120529180 on Build GRCh38
rs552681023 Chr.12: 120529157-120529157 on Build GRCh38
rs960589868 Chr.12: 120529247-120529247 on Build GRCh38
rs750386888 Chr.12: 120529117-120529117 on Build GRCh38
rs538130753 Chr.12: 120529154-120529154 on Build GRCh38
rs377206577 Chr.12: 120529002-120529002 on Build GRCh38
rs572858563 Chr.12: 120529302-120529302 on Build GRCh38
rs760191462 Chr.12: 120528985-120528985 on Build GRCh38
rs909420689 Chr.12: 120529023-120529023 on Build GRCh38
rs966018951 Chr.12: 120528888-120528888 on Build GRCh38
rs556837234 Chr.12: 120529232-120529232 on Build GRCh38
rs749232178 Chr.12: 120528932-120528932 on Build GRCh38
rs746677755 Chr.12: 120529048-120529048 on Build GRCh38
rs972320159 Chr.12: 120529054-120529054 on Build GRCh38
rs147150296 Chr.12: 120529185-120529185 on Build GRCh38
rs991167260 Chr.12: 120529250-120529251 on Build GRCh38
rs776156410 Chr.12: 120529056-120529056 on Build GRCh38
rs984243970 Chr.12: 120529194-120529194 on Build GRCh38
rs531422278 Chr.12: 120528971-120528971 on Build GRCh38
rs369744093 Chr.12: 120529028-120529028 on Build GRCh38
rs915432370 Chr.12: 120529336-120529336 on Build GRCh38
rs1035928949 Chr.12: 120529243-120529243 on Build GRCh38
rs754753362 Chr.12: 120529171-120529171 on Build GRCh38
rs373266483 Chr.12: 120529099-120529099 on Build GRCh38
rs976764382 Chr.12: 120529277-120529277 on Build GRCh38
rs139184415 Chr.12: 120529034-120529034 on Build GRCh38
rs140043357 Chr.12: 120529126-120529126 on Build GRCh38
rs775253112 Chr.12: 120528984-120528984 on Build GRCh38
rs754066407 Chr.12: 120529074-120529075 on Build GRCh38
rs3742052 Chr.12: 120529118-120529118 on Build GRCh38
rs372890461 Chr.12: 120528948-120528948 on Build GRCh38
rs757373189 Chr.12: 120529111-120529111 on Build GRCh38
rs750410862 Chr.12: 120529161-120529161 on Build GRCh38
rs960246883 Chr.12: 120529170-120529170 on Build GRCh38

View Full Product Details