Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121003322-121003857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf43
Entrez Gene ID
64897
Gene Name
chromosome 12 open reading frame 43
Gene Aliases
-
UniGene
Hs.528671
Gene Chromosome Location
Chr.12: 121002422-121016502 on Build GRCh38
Transcripts
NM_001286191.1 NM_001286192.1 NM_001286195.1 NM_001286196.1 NM_001286197.1 NM_001286198.1 NM_022895.2 NR_104409.1 XM_017019829.1

SNP Detail

SNP ID Chromosome Location
rs544484296 Chr.12: 121003456-121003456 on Build GRCh38
rs1169312 Chr.12: 121003658-121003658 on Build GRCh38
rs184046090 Chr.12: 121003416-121003416 on Build GRCh38
rs966596699 Chr.12: 121003767-121003768 on Build GRCh38
rs191296204 Chr.12: 121003813-121003813 on Build GRCh38
rs1054727555 Chr.12: 121003744-121003745 on Build GRCh38
rs1045422485 Chr.12: 121003449-121003449 on Build GRCh38
rs773330068 Chr.12: 121003376-121003376 on Build GRCh38
rs762990157 Chr.12: 121003380-121003380 on Build GRCh38
rs1055619595 Chr.12: 121003400-121003400 on Build GRCh38
rs957887525 Chr.12: 121003508-121003508 on Build GRCh38
rs915725873 Chr.12: 121003761-121003761 on Build GRCh38
rs1033596046 Chr.12: 121003432-121003432 on Build GRCh38
rs1010840223 Chr.12: 121003760-121003760 on Build GRCh38
rs975655430 Chr.12: 121003632-121003632 on Build GRCh38
rs756050692 Chr.12: 121003649-121003649 on Build GRCh38
rs898249109 Chr.12: 121003625-121003625 on Build GRCh38
rs745510278 Chr.12: 121003730-121003730 on Build GRCh38
rs572092143 Chr.12: 121003710-121003710 on Build GRCh38
rs745944268 Chr.12: 121003745-121003745 on Build GRCh38
rs188438469 Chr.12: 121003480-121003480 on Build GRCh38
rs527691515 Chr.12: 121003671-121003671 on Build GRCh38
rs191088838 Chr.12: 121003777-121003777 on Build GRCh38
rs1019746648 Chr.12: 121003781-121003781 on Build GRCh38
rs886821297 Chr.12: 121003347-121003347 on Build GRCh38
rs991336146 Chr.12: 121003749-121003749 on Build GRCh38
rs998919676 Chr.12: 121003771-121003771 on Build GRCh38
rs967841200 Chr.12: 121003546-121003546 on Build GRCh38
rs552205524 Chr.12: 121003414-121003414 on Build GRCh38
rs1008872509 Chr.12: 121003662-121003662 on Build GRCh38
rs532204852 Chr.12: 121003361-121003361 on Build GRCh38
rs949999993 Chr.12: 121003764-121003764 on Build GRCh38
rs1001858592 Chr.12: 121003731-121003731 on Build GRCh38
rs1038562354 Chr.12: 121003544-121003544 on Build GRCh38
rs893673814 Chr.12: 121003749-121003749 on Build GRCh38
rs56383076 Chr.12: 121003530-121003530 on Build GRCh38
rs967077576 Chr.12: 121003670-121003670 on Build GRCh38
rs1032465801 Chr.12: 121003697-121003697 on Build GRCh38
rs536841352 Chr.12: 121003557-121003557 on Build GRCh38
rs759748820 Chr.12: 121003488-121003488 on Build GRCh38
rs2178464 Chr.12: 121003637-121003637 on Build GRCh38
rs3751153 Chr.12: 121003665-121003665 on Build GRCh38
rs912553311 Chr.12: 121003571-121003571 on Build GRCh38
rs370864344 Chr.12: 121003753-121003753 on Build GRCh38
rs778506891 Chr.12: 121003721-121003721 on Build GRCh38
rs965393254 Chr.12: 121003572-121003572 on Build GRCh38
rs777818991 Chr.12: 121003650-121003650 on Build GRCh38
rs1002058403 Chr.12: 121003417-121003417 on Build GRCh38
rs751805076 Chr.12: 121003434-121003434 on Build GRCh38
rs935852192 Chr.12: 121003362-121003362 on Build GRCh38
rs1011121816 Chr.12: 121003520-121003520 on Build GRCh38
rs540993355 Chr.12: 121003797-121003797 on Build GRCh38

View Full Product Details