Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121238401-121238896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMKK2
Entrez Gene ID
10645
Gene Name
calcium/calmodulin dependent protein kinase kinase 2
Gene Aliases
CAMKK, CAMKKB
UniGene
Hs.297343
Gene Chromosome Location
Chr.12: 121237692-121298308 on Build GRCh38
Transcripts
NM_001270485.1 NM_001270486.1 NM_006549.3 NM_153499.2 NM_153500.1 NM_172216.1 NM_172226.2 XM_005253822.3 XM_005253824.3 XM_011537763.1 XM_011537764.2 XM_017018694.1 XM_017018695.1 XM_017018696.1 XM_017018697.1 XM_017018698.1 XM_017018699.1 XM_017018700.1 XM_017018701.1 XM_017018702.1 XM_017018705.1 XM_017018706.1 XM_017018707.1 XM_017018709.1

SNP Detail

SNP ID Chromosome Location
rs529290312 Chr.12: 121238554-121238554 on Build GRCh38
rs1029919372 Chr.12: 121238710-121238710 on Build GRCh38
rs181795744 Chr.12: 121238585-121238585 on Build GRCh38
rs186976030 Chr.12: 121238607-121238607 on Build GRCh38
rs1017087178 Chr.12: 121238726-121238726 on Build GRCh38
rs202214622 Chr.12: 121238808-121238808 on Build GRCh38
rs1009118183 Chr.12: 121238437-121238437 on Build GRCh38
rs542804408 Chr.12: 121238782-121238782 on Build GRCh38
rs768833829 Chr.12: 121238484-121238484 on Build GRCh38
rs202049022 Chr.12: 121238817-121238817 on Build GRCh38
rs1653587 Chr.12: 121238429-121238429 on Build GRCh38
rs200102410 Chr.12: 121238752-121238752 on Build GRCh38
rs199947351 Chr.12: 121238794-121238794 on Build GRCh38
rs999040449 Chr.12: 121238488-121238488 on Build GRCh38
rs200390427 Chr.12: 121238583-121238583 on Build GRCh38
rs1653588 Chr.12: 121238863-121238863 on Build GRCh38
rs866924210 Chr.12: 121238438-121238438 on Build GRCh38
rs199672958 Chr.12: 121238634-121238634 on Build GRCh38
rs201062980 Chr.12: 121238611-121238611 on Build GRCh38
rs200839343 Chr.12: 121238762-121238762 on Build GRCh38
rs777060768 Chr.12: 121238602-121238602 on Build GRCh38
rs766409432 Chr.12: 121238518-121238518 on Build GRCh38
rs373139606 Chr.12: 121238702-121238702 on Build GRCh38
rs1019617552 Chr.12: 121238459-121238459 on Build GRCh38
rs201669623 Chr.12: 121238743-121238743 on Build GRCh38
rs201645993 Chr.12: 121238786-121238786 on Build GRCh38
rs200307689 Chr.12: 121238853-121238853 on Build GRCh38
rs957425630 Chr.12: 121238515-121238515 on Build GRCh38
rs201274597 Chr.12: 121238860-121238860 on Build GRCh38
rs201413450 Chr.12: 121238496-121238496 on Build GRCh38
rs991746028 Chr.12: 121238824-121238824 on Build GRCh38
rs971804937 Chr.12: 121238670-121238670 on Build GRCh38
rs200625078 Chr.12: 121238665-121238665 on Build GRCh38
rs538669974 Chr.12: 121238641-121238641 on Build GRCh38
rs200121162 Chr.12: 121238812-121238812 on Build GRCh38
rs191466965 Chr.12: 121238657-121238657 on Build GRCh38
rs190987444 Chr.12: 121238468-121238468 on Build GRCh38
rs982026109 Chr.12: 121238706-121238706 on Build GRCh38
rs888749048 Chr.12: 121238832-121238832 on Build GRCh38
rs559587524 Chr.12: 121238774-121238774 on Build GRCh38
rs944389376 Chr.12: 121238843-121238843 on Build GRCh38
rs920467565 Chr.12: 121238784-121238784 on Build GRCh38
rs200290846 Chr.12: 121238483-121238483 on Build GRCh38
rs940640816 Chr.12: 121238720-121238720 on Build GRCh38
rs879515827 Chr.12: 121238563-121238563 on Build GRCh38
rs981159553 Chr.12: 121238862-121238872 on Build GRCh38
rs183408592 Chr.12: 121238813-121238813 on Build GRCh38
rs202208143 Chr.12: 121238562-121238562 on Build GRCh38
rs145160159 Chr.12: 121238719-121238719 on Build GRCh38
rs62933860 Chr.12: 121238573-121238573 on Build GRCh38
rs1033209682 Chr.12: 121238510-121238510 on Build GRCh38
rs200967158 Chr.12: 121238804-121238804 on Build GRCh38
rs903327736 Chr.12: 121238469-121238469 on Build GRCh38
rs201160204 Chr.12: 121238716-121238716 on Build GRCh38
rs200637479 Chr.12: 121238738-121238738 on Build GRCh38
rs199750539 Chr.12: 121238723-121238723 on Build GRCh38

View Full Product Details