Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121327014-121327523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC5
Entrez Gene ID
51433
Gene Name
anaphase promoting complex subunit 5
Gene Aliases
APC5
UniGene
Hs.7101
Gene Chromosome Location
Chr.12: 121308245-121354209 on Build GRCh38
Transcripts
NM_001137559.1 NM_016237.4 XM_005253900.3 XM_017019423.1 XM_017019424.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2175722 c.1313C>T p.A438V Chr.12: 121327223-121327223 on Build GRCh38
4661490 c.1431C>T p.H477H Chr.12: 121327105-121327105 on Build GRCh38
7449551 c.1324G>A p.A442T Chr.12: 121327212-121327212 on Build GRCh38
4039447 c.1310T>C p.M437T Chr.12: 121327226-121327226 on Build GRCh38
7242343 c.1410C>T p.L470L Chr.12: 121327126-121327126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782047843 Chr.12: 121327169-121327169 on Build GRCh38
rs371351583 Chr.12: 121327059-121327059 on Build GRCh38
rs781870526 Chr.12: 121327243-121327244 on Build GRCh38
rs547177366 Chr.12: 121327285-121327285 on Build GRCh38
rs782286059 Chr.12: 121327102-121327102 on Build GRCh38
rs537123234 Chr.12: 121327488-121327488 on Build GRCh38
rs886528197 Chr.12: 121327337-121327337 on Build GRCh38
rs755346684 Chr.12: 121327138-121327138 on Build GRCh38
rs116227311 Chr.12: 121327168-121327168 on Build GRCh38
rs898377517 Chr.12: 121327384-121327384 on Build GRCh38
rs781931612 Chr.12: 121327343-121327343 on Build GRCh38
rs1047875444 Chr.12: 121327319-121327319 on Build GRCh38
rs568132536 Chr.12: 121327255-121327255 on Build GRCh38
rs782709603 Chr.12: 121327170-121327170 on Build GRCh38
rs955243887 Chr.12: 121327426-121327426 on Build GRCh38
rs782511441 Chr.12: 121327103-121327103 on Build GRCh38
rs185066884 Chr.12: 121327445-121327445 on Build GRCh38
rs781937762 Chr.12: 121327119-121327119 on Build GRCh38
rs782134243 Chr.12: 121327164-121327164 on Build GRCh38
rs547367671 Chr.12: 121327318-121327318 on Build GRCh38
rs1028125388 Chr.12: 121327131-121327131 on Build GRCh38
rs368484764 Chr.12: 121327116-121327116 on Build GRCh38
rs782538891 Chr.12: 121327227-121327227 on Build GRCh38
rs782311213 Chr.12: 121327117-121327117 on Build GRCh38
rs925004071 Chr.12: 121327457-121327457 on Build GRCh38
rs1037726147 Chr.12: 121327368-121327368 on Build GRCh38
rs782165073 Chr.12: 121327389-121327389 on Build GRCh38
rs782220730 Chr.12: 121327266-121327266 on Build GRCh38
rs114313098 Chr.12: 121327472-121327472 on Build GRCh38
rs781931327 Chr.12: 121327273-121327273 on Build GRCh38
rs116012709 Chr.12: 121327441-121327441 on Build GRCh38
rs1055289399 Chr.12: 121327277-121327277 on Build GRCh38
rs114730469 Chr.12: 121327132-121327132 on Build GRCh38
rs1020622091 Chr.12: 121327431-121327431 on Build GRCh38
rs782639940 Chr.12: 121327105-121327105 on Build GRCh38
rs1008024681 Chr.12: 121327430-121327430 on Build GRCh38
rs371233998 Chr.12: 121327165-121327165 on Build GRCh38
rs779233762 Chr.12: 121327177-121327177 on Build GRCh38
rs782689692 Chr.12: 121327238-121327238 on Build GRCh38
rs782600216 Chr.12: 121327260-121327260 on Build GRCh38
rs782384727 Chr.12: 121327128-121327128 on Build GRCh38
rs199680151 Chr.12: 121327082-121327082 on Build GRCh38
rs782480729 Chr.12: 121327190-121327190 on Build GRCh38
rs781877821 Chr.12: 121327073-121327073 on Build GRCh38
rs781990372 Chr.12: 121327160-121327160 on Build GRCh38
rs116523210 Chr.12: 121327297-121327297 on Build GRCh38
rs781810218 Chr.12: 121327216-121327216 on Build GRCh38
rs782298804 Chr.12: 121327267-121327267 on Build GRCh38
rs782672840 Chr.12: 121327095-121327095 on Build GRCh38
rs782580075 Chr.12: 121327194-121327194 on Build GRCh38
rs781870840 Chr.12: 121327185-121327185 on Build GRCh38
rs368729676 Chr.12: 121327432-121327432 on Build GRCh38
rs782251690 Chr.12: 121327243-121327243 on Build GRCh38
rs781784460 Chr.12: 121327081-121327081 on Build GRCh38
rs141045075 Chr.12: 121327184-121327184 on Build GRCh38
rs36095958 Chr.12: 121327120-121327120 on Build GRCh38
rs557767539 Chr.12: 121327471-121327471 on Build GRCh38
rs782157779 Chr.12: 121327178-121327178 on Build GRCh38
rs782339225 Chr.12: 121327230-121327230 on Build GRCh38
rs560097160 Chr.12: 121327074-121327074 on Build GRCh38
rs555835653 Chr.12: 121327494-121327494 on Build GRCh38
rs782798597 Chr.12: 121327395-121327395 on Build GRCh38

View Full Product Details