Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123629827-123630324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B1
Entrez Gene ID
1967
Gene Name
eukaryotic translation initiation factor 2B subunit alpha
Gene Aliases
EIF2B, EIF2BA
UniGene
Hs.741273
Gene Chromosome Location
Chr.12: 123621023-123633776 on Build GRCh38
Transcripts
NM_001414.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1605720 c.265T>C p.C89R Chr.12: 123630273-123630273 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757985338 Chr.12: 123630246-123630246 on Build GRCh38
rs863225050 Chr.12: 123630210-123630210 on Build GRCh38
rs1049401968 Chr.12: 123629970-123629970 on Build GRCh38
rs759728057 Chr.12: 123630193-123630193 on Build GRCh38
rs3834752 Chr.12: 123630101-123630101 on Build GRCh38
rs781471132 Chr.12: 123630253-123630254 on Build GRCh38
rs1022496840 Chr.12: 123630122-123630122 on Build GRCh38
rs765287085 Chr.12: 123630205-123630205 on Build GRCh38
rs183323277 Chr.12: 123629946-123629946 on Build GRCh38
rs958081254 Chr.12: 123629935-123629935 on Build GRCh38
rs775739205 Chr.12: 123630217-123630217 on Build GRCh38
rs1041124768 Chr.12: 123630170-123630170 on Build GRCh38
rs557869421 Chr.12: 123630039-123630039 on Build GRCh38
rs547710140 Chr.12: 123629945-123629945 on Build GRCh38
rs906556394 Chr.12: 123630064-123630064 on Build GRCh38
rs781504897 Chr.12: 123630130-123630130 on Build GRCh38
rs916690424 Chr.12: 123629959-123629959 on Build GRCh38
rs73416221 Chr.12: 123630124-123630124 on Build GRCh38
rs755054824 Chr.12: 123630157-123630157 on Build GRCh38
rs112712085 Chr.12: 123630029-123630029 on Build GRCh38
rs768076568 Chr.12: 123630249-123630249 on Build GRCh38
rs773170410 Chr.12: 123630165-123630165 on Build GRCh38
rs36018806 Chr.12: 123630005-123630005 on Build GRCh38
rs1032135773 Chr.12: 123629930-123629930 on Build GRCh38
rs1015524724 Chr.12: 123630030-123630030 on Build GRCh38
rs78170695 Chr.12: 123630119-123630119 on Build GRCh38
rs535392372 Chr.12: 123629934-123629934 on Build GRCh38
rs748375449 Chr.12: 123630255-123630256 on Build GRCh38
rs541360450 Chr.12: 123630131-123630131 on Build GRCh38
rs764262014 Chr.12: 123630218-123630218 on Build GRCh38
rs201194663 Chr.12: 123630125-123630125 on Build GRCh38
rs970735564 Chr.12: 123629963-123629963 on Build GRCh38
rs944357736 Chr.12: 123630151-123630151 on Build GRCh38
rs747091269 Chr.12: 123630150-123630152 on Build GRCh38
rs754382992 Chr.12: 123630143-123630143 on Build GRCh38
rs142755878 Chr.12: 123630257-123630257 on Build GRCh38
rs770912605 Chr.12: 123630173-123630173 on Build GRCh38
rs920771424 Chr.12: 123630123-123630123 on Build GRCh38
rs966610157 Chr.12: 123629868-123629868 on Build GRCh38
rs1007863823 Chr.12: 123630158-123630158 on Build GRCh38
rs1050397 Chr.12: 123630252-123630252 on Build GRCh38
rs747617594 Chr.12: 123630265-123630265 on Build GRCh38
rs568295468 Chr.12: 123629860-123629860 on Build GRCh38
rs553282463 Chr.12: 123630154-123630154 on Build GRCh38
rs1004779029 Chr.12: 123630010-123630010 on Build GRCh38
rs148513104 Chr.12: 123630167-123630167 on Build GRCh38
rs757509025 Chr.12: 123630248-123630248 on Build GRCh38
rs999289359 Chr.12: 123629920-123629920 on Build GRCh38
rs746730077 Chr.12: 123630169-123630169 on Build GRCh38
rs1004001270 Chr.12: 123630098-123630098 on Build GRCh38
rs776365178 Chr.12: 123630190-123630190 on Build GRCh38
rs778778741 Chr.12: 123630146-123630146 on Build GRCh38
rs900294299 Chr.12: 123629926-123629926 on Build GRCh38
rs188253706 Chr.12: 123630057-123630057 on Build GRCh38
rs79015071 Chr.12: 123629859-123629859 on Build GRCh38
rs982550249 Chr.12: 123629990-123629990 on Build GRCh38
rs758142806 Chr.12: 123630269-123630269 on Build GRCh38
rs865958824 Chr.12: 123630099-123630099 on Build GRCh38
rs959760069 Chr.12: 123630105-123630105 on Build GRCh38
rs370141777 Chr.12: 123630118-123630118 on Build GRCh38

View Full Product Details