Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113119347-113119845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F7
Entrez Gene ID
2155
Gene Name
coagulation factor VII
Gene Aliases
SPCA
UniGene
Hs.36989
Gene Chromosome Location
Chr.13: 113105773-113120681 on Build GRCh38
Transcripts
NM_000131.4 NM_001267554.1 NM_019616.3 NR_051961.1 XM_006719963.3 XM_011537474.2 XM_011537475.2 XM_011537476.2

SNP Detail

SNP ID Chromosome Location
rs1001159046 Chr.13: 113119512-113119512 on Build GRCh38
rs962425901 Chr.13: 113119726-113119726 on Build GRCh38
rs747160994 Chr.13: 113119634-113119636 on Build GRCh38
rs886049997 Chr.13: 113119764-113119778 on Build GRCh38
rs908318351 Chr.13: 113119426-113119426 on Build GRCh38
rs982816417 Chr.13: 113119716-113119716 on Build GRCh38
rs908339282 Chr.13: 113119473-113119473 on Build GRCh38
rs886049996 Chr.13: 113119717-113119720 on Build GRCh38
rs908286143 Chr.13: 113119590-113119590 on Build GRCh38
rs575778236 Chr.13: 113119777-113119777 on Build GRCh38
rs767798867 Chr.13: 113119801-113119801 on Build GRCh38
rs35430957 Chr.13: 113119809-113119809 on Build GRCh38
rs898219172 Chr.13: 113119710-113119711 on Build GRCh38
rs886049995 Chr.13: 113119686-113119687 on Build GRCh38
rs151109093 Chr.13: 113119628-113119628 on Build GRCh38
rs561457617 Chr.13: 113119635-113119635 on Build GRCh38
rs925503448 Chr.13: 113119655-113119656 on Build GRCh38
rs941165985 Chr.13: 113119452-113119452 on Build GRCh38
rs145837821 Chr.13: 113119688-113119688 on Build GRCh38
rs1041758570 Chr.13: 113119658-113119658 on Build GRCh38
rs971712790 Chr.13: 113119711-113119711 on Build GRCh38
rs912464043 Chr.13: 113119792-113119792 on Build GRCh38
rs1024972477 Chr.13: 113119695-113119695 on Build GRCh38
rs886049994 Chr.13: 113119636-113119638 on Build GRCh38
rs1033443053 Chr.13: 113119743-113119756 on Build GRCh38
rs1050419886 Chr.13: 113119501-113119501 on Build GRCh38
rs771890118 Chr.13: 113119629-113119629 on Build GRCh38
rs886049993 Chr.13: 113119590-113119590 on Build GRCh38
rs565588646 Chr.13: 113119728-113119728 on Build GRCh38
rs958913066 Chr.13: 113119551-113119551 on Build GRCh38
rs896676699 Chr.13: 113119373-113119373 on Build GRCh38
rs1015268173 Chr.13: 113119718-113119718 on Build GRCh38
rs886049992 Chr.13: 113119571-113119572 on Build GRCh38
rs886049991 Chr.13: 113119551-113119551 on Build GRCh38
rs144204350 Chr.13: 113119613-113119613 on Build GRCh38
rs916916252 Chr.13: 113119401-113119401 on Build GRCh38
rs926680806 Chr.13: 113119487-113119490 on Build GRCh38
rs1038249568 Chr.13: 113119453-113119453 on Build GRCh38
rs936028812 Chr.13: 113119812-113119812 on Build GRCh38
rs886049990 Chr.13: 113119433-113119433 on Build GRCh38
rs1015469305 Chr.13: 113119586-113119587 on Build GRCh38
rs115484165 Chr.13: 113119398-113119398 on Build GRCh38
rs1024585962 Chr.13: 113119563-113119563 on Build GRCh38
rs1020774717 Chr.13: 113119538-113119538 on Build GRCh38
rs962662495 Chr.13: 113119477-113119477 on Build GRCh38
rs141116208 Chr.13: 113119666-113119666 on Build GRCh38
rs1000315742 Chr.13: 113119549-113119549 on Build GRCh38
rs1056452548 Chr.13: 113119498-113119501 on Build GRCh38
rs932460248 Chr.13: 113119737-113119737 on Build GRCh38
rs3093253 Chr.13: 113119778-113119778 on Build GRCh38
rs372580388 Chr.13: 113119764-113119764 on Build GRCh38
rs560342203 Chr.13: 113119481-113119481 on Build GRCh38
rs982505268 Chr.13: 113119579-113119579 on Build GRCh38
rs1032718728 Chr.13: 113119411-113119411 on Build GRCh38
rs750788946 Chr.13: 113119811-113119811 on Build GRCh38
rs890790982 Chr.13: 113119680-113119680 on Build GRCh38
rs768906862 Chr.13: 113119531-113119532 on Build GRCh38
rs1029348918 Chr.13: 113119459-113119460 on Build GRCh38
rs973869077 Chr.13: 113119602-113119602 on Build GRCh38
rs949808081 Chr.13: 113119408-113119408 on Build GRCh38
rs568851012 Chr.13: 113119687-113119688 on Build GRCh38
rs962856176 Chr.13: 113119594-113119594 on Build GRCh38
rs932564995 Chr.13: 113119492-113119492 on Build GRCh38
rs533636983 Chr.13: 113119607-113119622 on Build GRCh38
rs543052575 Chr.13: 113119539-113119539 on Build GRCh38
rs975325547 Chr.13: 113119754-113119754 on Build GRCh38
rs941091976 Chr.13: 113119592-113119592 on Build GRCh38
rs778584652 Chr.13: 113119550-113119550 on Build GRCh38
rs149716932 Chr.13: 113119460-113119461 on Build GRCh38
rs922066692 Chr.13: 113119758-113119758 on Build GRCh38
rs541698176 Chr.13: 113119612-113119612 on Build GRCh38
rs992073312 Chr.13: 113119743-113119744 on Build GRCh38
rs974397393 Chr.13: 113119479-113119480 on Build GRCh38
rs978225636 Chr.13: 113119744-113119744 on Build GRCh38
rs543290081 Chr.13: 113119568-113119568 on Build GRCh38
rs1001484466 Chr.13: 113119409-113119409 on Build GRCh38
rs185229430 Chr.13: 113119646-113119646 on Build GRCh38
rs751688576 Chr.13: 113119709-113119709 on Build GRCh38
rs933616047 Chr.13: 113119765-113119765 on Build GRCh38
rs36209570 Chr.13: 113119489-113119490 on Build GRCh38
rs139474305 Chr.13: 113119418-113119418 on Build GRCh38
rs926548736 Chr.13: 113119603-113119604 on Build GRCh38
rs1050722703 Chr.13: 113119680-113119681 on Build GRCh38
rs567634576 Chr.13: 113119678-113119678 on Build GRCh38
rs575689457 Chr.13: 113119507-113119507 on Build GRCh38
rs539255438 Chr.13: 113119729-113119729 on Build GRCh38
rs560585327 Chr.13: 113119563-113119564 on Build GRCh38
rs781542714 Chr.13: 113119744-113119755 on Build GRCh38
rs3093250 Chr.13: 113119647-113119647 on Build GRCh38
rs936741117 Chr.13: 113119750-113119750 on Build GRCh38
rs759550994 Chr.13: 113119679-113119679 on Build GRCh38
rs113386992 Chr.13: 113119406-113119406 on Build GRCh38
rs899712922 Chr.13: 113119464-113119464 on Build GRCh38
rs772137874 Chr.13: 113119480-113119483 on Build GRCh38
rs1009755675 Chr.13: 113119377-113119377 on Build GRCh38
rs36209571 Chr.13: 113119738-113119738 on Build GRCh38
rs925378767 Chr.13: 113119748-113119748 on Build GRCh38
rs762431471 Chr.13: 113119571-113119582 on Build GRCh38
rs557518714 Chr.13: 113119735-113119735 on Build GRCh38
rs3093251 Chr.13: 113119681-113119681 on Build GRCh38
rs932530832 Chr.13: 113119618-113119618 on Build GRCh38
rs376163218 Chr.13: 113119671-113119671 on Build GRCh38
rs557431193 Chr.13: 113119506-113119506 on Build GRCh38
rs1042075478 Chr.13: 113119807-113119807 on Build GRCh38
rs3093252 Chr.13: 113119710-113119713 on Build GRCh38
rs529590090 Chr.13: 113119558-113119558 on Build GRCh38
rs904016206 Chr.13: 113119512-113119513 on Build GRCh38
rs890824020 Chr.13: 113119789-113119789 on Build GRCh38
rs150239572 Chr.13: 113119672-113119672 on Build GRCh38
rs974038858 Chr.13: 113119597-113119597 on Build GRCh38
rs116688254 Chr.13: 113119541-113119541 on Build GRCh38

View Full Product Details