Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113208496-113208987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL4A
Entrez Gene ID
8451
Gene Name
cullin 4A
Gene Aliases
-
UniGene
Hs.339735
Gene Chromosome Location
Chr.13: 113208193-113265078 on Build GRCh38
Transcripts
NM_001278513.1 NM_001278514.1 NM_003589.2 NM_001127202.3 NM_001127203.3 NM_001258213.2 NM_001320655.1 NM_001320656.1 NM_001320657.1 NM_001320659.1 NM_001320660.1 NM_018386.4 XM_005268325.3 XM_017020665.1 XM_017020666.1 XM_017020667.1 XM_017020669.1

Gene Symbol
PCID2
Entrez Gene ID
55795
Gene Name
PCI domain containing 2
Gene Aliases
F10
UniGene
Hs.508769
Gene Chromosome Location
Chr.13: 113165002-113209466 on Build GRCh38
Transcripts
NM_001278513.1 NM_001278514.1 NM_003589.2 NM_001127202.3 NM_001127203.3 NM_001258213.2 NM_001320655.1 NM_001320656.1 NM_001320657.1 NM_001320659.1 NM_001320660.1 NM_018386.4 XM_005268325.3 XM_017020665.1 XM_017020666.1 XM_017020667.1 XM_017020669.1

SNP Detail

SNP ID Chromosome Location
rs778009640 Chr.13: 113208579-113208579 on Build GRCh38
rs1025012990 Chr.13: 113208681-113208681 on Build GRCh38
rs1013811140 Chr.13: 113208527-113208527 on Build GRCh38
rs769248937 Chr.13: 113208650-113208650 on Build GRCh38
rs1032953956 Chr.13: 113208600-113208600 on Build GRCh38
rs772849316 Chr.13: 113208651-113208651 on Build GRCh38
rs989877015 Chr.13: 113208627-113208627 on Build GRCh38
rs1012052033 Chr.13: 113208784-113208784 on Build GRCh38
rs146751759 Chr.13: 113208935-113208935 on Build GRCh38
rs911010999 Chr.13: 113208934-113208934 on Build GRCh38
rs981731587 Chr.13: 113208565-113208565 on Build GRCh38
rs1046376485 Chr.13: 113208835-113208835 on Build GRCh38
rs375308286 Chr.13: 113208652-113208652 on Build GRCh38
rs377176742 Chr.13: 113208772-113208772 on Build GRCh38
rs935252569 Chr.13: 113208765-113208765 on Build GRCh38
rs142022899 Chr.13: 113208624-113208624 on Build GRCh38
rs777852240 Chr.13: 113208627-113208627 on Build GRCh38
rs1041353979 Chr.13: 113208755-113208755 on Build GRCh38
rs919835260 Chr.13: 113208968-113208968 on Build GRCh38
rs748314949 Chr.13: 113208566-113208566 on Build GRCh38
rs895223623 Chr.13: 113208812-113208812 on Build GRCh38
rs775261097 Chr.13: 113208636-113208637 on Build GRCh38
rs1003076435 Chr.13: 113208796-113208796 on Build GRCh38
rs1033016011 Chr.13: 113208886-113208886 on Build GRCh38
rs943917869 Chr.13: 113208753-113208753 on Build GRCh38
rs776116132 Chr.13: 113208641-113208641 on Build GRCh38
rs1026619927 Chr.13: 113208839-113208839 on Build GRCh38
rs561335158 Chr.13: 113208834-113208834 on Build GRCh38
rs369057180 Chr.13: 113208658-113208658 on Build GRCh38
rs138888496 Chr.13: 113208599-113208599 on Build GRCh38
rs547326301 Chr.13: 113208584-113208584 on Build GRCh38
rs976574452 Chr.13: 113208948-113208948 on Build GRCh38
rs780931516 Chr.13: 113208562-113208562 on Build GRCh38
rs931382765 Chr.13: 113208670-113208670 on Build GRCh38
rs951951204 Chr.13: 113208787-113208787 on Build GRCh38
rs970681816 Chr.13: 113208521-113208521 on Build GRCh38
rs760623421 Chr.13: 113208686-113208686 on Build GRCh38
rs939691795 Chr.13: 113208576-113208576 on Build GRCh38
rs972634708 Chr.13: 113208967-113208967 on Build GRCh38
rs961580943 Chr.13: 113208825-113208825 on Build GRCh38
rs779237331 Chr.13: 113208671-113208671 on Build GRCh38
rs148604743 Chr.13: 113208679-113208679 on Build GRCh38
rs34519626 Chr.13: 113208719-113208720 on Build GRCh38
rs545595213 Chr.13: 113208742-113208742 on Build GRCh38
rs777261094 Chr.13: 113208686-113208686 on Build GRCh38
rs989826433 Chr.13: 113208738-113208738 on Build GRCh38
rs759761696 Chr.13: 113208585-113208585 on Build GRCh38
rs749489503 Chr.13: 113208581-113208581 on Build GRCh38
rs965927303 Chr.13: 113208954-113208954 on Build GRCh38
rs982082353 Chr.13: 113208682-113208682 on Build GRCh38
rs919593534 Chr.13: 113208737-113208737 on Build GRCh38
rs911029982 Chr.13: 113208752-113208752 on Build GRCh38
rs769295690 Chr.13: 113208684-113208684 on Build GRCh38
rs372700568 Chr.13: 113208732-113208732 on Build GRCh38
rs766059086 Chr.13: 113208613-113208613 on Build GRCh38
rs746057645 Chr.13: 113208675-113208675 on Build GRCh38
rs973072260 Chr.13: 113208777-113208777 on Build GRCh38
rs756235474 Chr.13: 113208625-113208625 on Build GRCh38
rs762729429 Chr.13: 113208612-113208612 on Build GRCh38
rs56771694 Chr.13: 113208588-113208588 on Build GRCh38
rs751379841 Chr.13: 113208619-113208619 on Build GRCh38
rs1023072270 Chr.13: 113208790-113208790 on Build GRCh38
rs184355597 Chr.13: 113208743-113208743 on Build GRCh38
rs141920233 Chr.13: 113208635-113208635 on Build GRCh38
rs535329317 Chr.13: 113208710-113208710 on Build GRCh38
rs528432986 Chr.13: 113208928-113208928 on Build GRCh38
rs931014389 Chr.13: 113208740-113208740 on Build GRCh38
rs542701792 Chr.13: 113208814-113208814 on Build GRCh38
rs747545850 Chr.13: 113208645-113208645 on Build GRCh38
rs1055003747 Chr.13: 113208809-113208809 on Build GRCh38
rs998848353 Chr.13: 113208647-113208647 on Build GRCh38
rs771091575 Chr.13: 113208582-113208582 on Build GRCh38
rs773899060 Chr.13: 113208643-113208643 on Build GRCh38
rs754714207 Chr.13: 113208620-113208620 on Build GRCh38
rs567911642 Chr.13: 113208689-113208689 on Build GRCh38
rs578227456 Chr.13: 113208715-113208715 on Build GRCh38
rs758322283 Chr.13: 113208555-113208555 on Build GRCh38
rs1025233617 Chr.13: 113208578-113208578 on Build GRCh38
rs961339955 Chr.13: 113208959-113208959 on Build GRCh38
rs781253894 Chr.13: 113208621-113208621 on Build GRCh38
rs372247269 Chr.13: 113208633-113208633 on Build GRCh38
rs972986902 Chr.13: 113208827-113208827 on Build GRCh38
rs893937831 Chr.13: 113208924-113208924 on Build GRCh38
rs953784881 Chr.13: 113208947-113208947 on Build GRCh38
rs754943305 Chr.13: 113208558-113208558 on Build GRCh38
rs1053648430 Chr.13: 113208960-113208960 on Build GRCh38
rs893151478 Chr.13: 113208781-113208781 on Build GRCh38
rs752439586 Chr.13: 113208660-113208660 on Build GRCh38
rs376321867 Chr.13: 113208951-113208951 on Build GRCh38
rs532925648 Chr.13: 113208589-113208589 on Build GRCh38
rs756076160 Chr.13: 113208662-113208662 on Build GRCh38
rs138409614 Chr.13: 113208634-113208634 on Build GRCh38
rs952373834 Chr.13: 113208849-113208849 on Build GRCh38
rs895107958 Chr.13: 113208676-113208676 on Build GRCh38
rs373010760 Chr.13: 113208665-113208665 on Build GRCh38
rs1011625946 Chr.13: 113208654-113208654 on Build GRCh38
rs1053016563 Chr.13: 113208776-113208776 on Build GRCh38
rs986073059 Chr.13: 113208818-113208818 on Build GRCh38
rs906012818 Chr.13: 113208793-113208793 on Build GRCh38
rs780021015 Chr.13: 113208556-113208556 on Build GRCh38
rs902531715 Chr.13: 113208867-113208867 on Build GRCh38
rs1035973872 Chr.13: 113208704-113208704 on Build GRCh38
rs928265195 Chr.13: 113208568-113208568 on Build GRCh38
rs749908696 Chr.13: 113208607-113208607 on Build GRCh38
rs553664236 Chr.13: 113208712-113208712 on Build GRCh38
rs985603635 Chr.13: 113208880-113208880 on Build GRCh38
rs548394915 Chr.13: 113208763-113208763 on Build GRCh38
rs769858877 Chr.13: 113208572-113208572 on Build GRCh38
rs115957258 Chr.13: 113208629-113208629 on Build GRCh38
rs143182963 Chr.13: 113208893-113208893 on Build GRCh38
rs772104027 Chr.13: 113208633-113208634 on Build GRCh38

View Full Product Details