Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113423964-113424461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRHL1
Entrez Gene ID
113622
Gene Name
ADP-ribosylhydrolase like 1
Gene Aliases
ARH2
UniGene
Hs.98669
Gene Chromosome Location
Chr.13: 113421940-113453524 on Build GRCh38
Transcripts
NM_138430.4 NM_199162.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1365601 c.885T>C p.C295C Chr.13: 113424239-113424239 on Build GRCh38
3872877 c.784A>G p.K262E Chr.13: 113424340-113424340 on Build GRCh38
7400074 c.877G>C p.E293Q Chr.13: 113424247-113424247 on Build GRCh38
4455495 c.798A>T p.E266D Chr.13: 113424326-113424326 on Build GRCh38
5036793 c.826G>T p.A276S Chr.13: 113424298-113424298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900980229 Chr.13: 113424424-113424424 on Build GRCh38
rs367917104 Chr.13: 113424335-113424335 on Build GRCh38
rs781585409 Chr.13: 113424282-113424282 on Build GRCh38
rs781497547 Chr.13: 113424180-113424180 on Build GRCh38
rs376646981 Chr.13: 113424328-113424328 on Build GRCh38
rs1015210489 Chr.13: 113424019-113424019 on Build GRCh38
rs779967179 Chr.13: 113424171-113424171 on Build GRCh38
rs759583938 Chr.13: 113424214-113424214 on Build GRCh38
rs746353598 Chr.13: 113424367-113424367 on Build GRCh38
rs962376208 Chr.13: 113424025-113424025 on Build GRCh38
rs755237339 Chr.13: 113424178-113424178 on Build GRCh38
rs1039964576 Chr.13: 113424413-113424413 on Build GRCh38
rs777992462 Chr.13: 113424297-113424297 on Build GRCh38
rs771676848 Chr.13: 113424201-113424201 on Build GRCh38
rs910119866 Chr.13: 113424050-113424050 on Build GRCh38
rs149444478 Chr.13: 113424309-113424309 on Build GRCh38
rs778795138 Chr.13: 113424089-113424089 on Build GRCh38
rs904448222 Chr.13: 113424202-113424202 on Build GRCh38
rs372574116 Chr.13: 113424375-113424375 on Build GRCh38
rs749058208 Chr.13: 113424379-113424379 on Build GRCh38
rs764921219 Chr.13: 113424235-113424235 on Build GRCh38
rs756515875 Chr.13: 113424231-113424248 on Build GRCh38
rs756274050 Chr.13: 113424337-113424337 on Build GRCh38
rs749835067 Chr.13: 113424339-113424339 on Build GRCh38
rs951629723 Chr.13: 113424010-113424010 on Build GRCh38
rs552680005 Chr.13: 113424124-113424124 on Build GRCh38
rs375544824 Chr.13: 113424195-113424195 on Build GRCh38
rs187023854 Chr.13: 113424121-113424121 on Build GRCh38
rs759538373 Chr.13: 113424385-113424385 on Build GRCh38
rs953878075 Chr.13: 113424095-113424095 on Build GRCh38
rs775720750 Chr.13: 113424399-113424399 on Build GRCh38
rs141758858 Chr.13: 113424376-113424376 on Build GRCh38
rs537604938 Chr.13: 113424026-113424026 on Build GRCh38
rs756583732 Chr.13: 113424294-113424294 on Build GRCh38
rs777989480 Chr.13: 113424344-113424344 on Build GRCh38
rs189816799 Chr.13: 113424129-113424129 on Build GRCh38
rs766539065 Chr.13: 113424277-113424277 on Build GRCh38
rs749725392 Chr.13: 113424196-113424196 on Build GRCh38
rs150141456 Chr.13: 113424387-113424387 on Build GRCh38
rs140383853 Chr.13: 113423992-113423992 on Build GRCh38
rs1003953330 Chr.13: 113423995-113423995 on Build GRCh38
rs770234182 Chr.13: 113424191-113424191 on Build GRCh38
rs115924315 Chr.13: 113424037-113424037 on Build GRCh38
rs776229218 Chr.13: 113424232-113424232 on Build GRCh38
rs948982416 Chr.13: 113424410-113424410 on Build GRCh38
rs371140225 Chr.13: 113424329-113424329 on Build GRCh38
rs760737230 Chr.13: 113424401-113424401 on Build GRCh38
rs780677270 Chr.13: 113424371-113424371 on Build GRCh38
rs768306415 Chr.13: 113424229-113424229 on Build GRCh38
rs751493419 Chr.13: 113424333-113424333 on Build GRCh38
rs747324118 Chr.13: 113424175-113424175 on Build GRCh38
rs747942802 Chr.13: 113424033-113424033 on Build GRCh38
rs143083124 Chr.13: 113424310-113424310 on Build GRCh38
rs750416511 Chr.13: 113424240-113424240 on Build GRCh38
rs137900593 Chr.13: 113424302-113424302 on Build GRCh38
rs938448319 Chr.13: 113424336-113424336 on Build GRCh38
rs201515700 Chr.13: 113424301-113424301 on Build GRCh38
rs544694553 Chr.13: 113424146-113424146 on Build GRCh38
rs200663640 Chr.13: 113424380-113424380 on Build GRCh38
rs748553751 Chr.13: 113424188-113424188 on Build GRCh38
rs202204683 Chr.13: 113424363-113424363 on Build GRCh38
rs778243177 Chr.13: 113424239-113424240 on Build GRCh38
rs774846550 Chr.13: 113424206-113424206 on Build GRCh38
rs774469077 Chr.13: 113424381-113424381 on Build GRCh38
rs761425732 Chr.13: 113424321-113424321 on Build GRCh38
rs143622057 Chr.13: 113424230-113424230 on Build GRCh38
rs776317103 Chr.13: 113424318-113424318 on Build GRCh38
rs1054691159 Chr.13: 113424207-113424207 on Build GRCh38
rs147753084 Chr.13: 113424234-113424234 on Build GRCh38
rs752942843 Chr.13: 113424289-113424289 on Build GRCh38
rs1025666117 Chr.13: 113424275-113424275 on Build GRCh38
rs571598266 Chr.13: 113424132-113424132 on Build GRCh38
rs779414316 Chr.13: 113424304-113424304 on Build GRCh38
rs754154287 Chr.13: 113424353-113424353 on Build GRCh38
rs567957103 Chr.13: 113424096-113424096 on Build GRCh38
rs977528534 Chr.13: 113424131-113424131 on Build GRCh38
rs374406687 Chr.13: 113424330-113424330 on Build GRCh38
rs755067590 Chr.13: 113424281-113424281 on Build GRCh38
rs112591236 Chr.13: 113424151-113424151 on Build GRCh38
rs374715069 Chr.13: 113424364-113424364 on Build GRCh38
rs141770513 Chr.13: 113424217-113424217 on Build GRCh38
rs41315960 Chr.13: 113424370-113424370 on Build GRCh38
rs556597574 Chr.13: 113424154-113424154 on Build GRCh38
rs80197449 Chr.13: 113424278-113424278 on Build GRCh38
rs41306688 Chr.13: 113424243-113424243 on Build GRCh38

View Full Product Details