Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113444184-113444688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRHL1
Entrez Gene ID
113622
Gene Name
ADP-ribosylhydrolase like 1
Gene Aliases
ARH2
UniGene
Hs.98669
Gene Chromosome Location
Chr.13: 113421940-113453524 on Build GRCh38
Transcripts
NM_138430.4 NM_199162.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
304459 c.287A>G p.E96G Chr.13: 113444517-113444517 on Build GRCh38
7354622 c.218A>T p.Y73F Chr.13: 113444586-113444586 on Build GRCh38
945600 c.365C>T p.P122L Chr.13: 113444439-113444439 on Build GRCh38
6595357 c.292C>T p.R98W Chr.13: 113444512-113444512 on Build GRCh38
361755 c.330A>G p.L110L Chr.13: 113444474-113444474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987115783 Chr.13: 113444288-113444289 on Build GRCh38
rs981701502 Chr.13: 113444255-113444255 on Build GRCh38
rs952803832 Chr.13: 113444524-113444524 on Build GRCh38
rs775474141 Chr.13: 113444444-113444444 on Build GRCh38
rs775650091 Chr.13: 113444608-113444608 on Build GRCh38
rs1019483733 Chr.13: 113444292-113444292 on Build GRCh38
rs1028213864 Chr.13: 113444264-113444264 on Build GRCh38
rs765759644 Chr.13: 113444562-113444562 on Build GRCh38
rs781063926 Chr.13: 113444336-113444336 on Build GRCh38
rs190337242 Chr.13: 113444208-113444208 on Build GRCh38
rs940392084 Chr.13: 113444290-113444290 on Build GRCh38
rs1023805282 Chr.13: 113444391-113444391 on Build GRCh38
rs754439489 Chr.13: 113444493-113444493 on Build GRCh38
rs768306273 Chr.13: 113444401-113444401 on Build GRCh38
rs756451379 Chr.13: 113444386-113444386 on Build GRCh38
rs753033301 Chr.13: 113444385-113444385 on Build GRCh38
rs757968307 Chr.13: 113444399-113444399 on Build GRCh38
rs375112372 Chr.13: 113444590-113444590 on Build GRCh38
rs958428154 Chr.13: 113444225-113444225 on Build GRCh38
rs3814257 Chr.13: 113444222-113444222 on Build GRCh38
rs184857251 Chr.13: 113444207-113444207 on Build GRCh38
rs972850298 Chr.13: 113444314-113444314 on Build GRCh38
rs760608077 Chr.13: 113444632-113444632 on Build GRCh38
rs141618303 Chr.13: 113444571-113444571 on Build GRCh38
rs1026802823 Chr.13: 113444520-113444520 on Build GRCh38
rs756472053 Chr.13: 113444453-113444453 on Build GRCh38
rs757778064 Chr.13: 113444472-113444472 on Build GRCh38
rs866656806 Chr.13: 113444439-113444439 on Build GRCh38
rs199718907 Chr.13: 113444593-113444593 on Build GRCh38
rs368794023 Chr.13: 113444477-113444477 on Build GRCh38
rs200232170 Chr.13: 113444589-113444589 on Build GRCh38
rs970857033 Chr.13: 113444393-113444393 on Build GRCh38
rs3814256 Chr.13: 113444388-113444388 on Build GRCh38
rs1052814854 Chr.13: 113444392-113444392 on Build GRCh38
rs780756171 Chr.13: 113444499-113444499 on Build GRCh38
rs559775826 Chr.13: 113444652-113444652 on Build GRCh38
rs752352484 Chr.13: 113444579-113444579 on Build GRCh38
rs754398410 Chr.13: 113444469-113444469 on Build GRCh38
rs958347622 Chr.13: 113444383-113444383 on Build GRCh38
rs181024248 Chr.13: 113444279-113444279 on Build GRCh38
rs774575530 Chr.13: 113444431-113444431 on Build GRCh38
rs577913936 Chr.13: 113444460-113444460 on Build GRCh38
rs150080386 Chr.13: 113444533-113444533 on Build GRCh38
rs368351748 Chr.13: 113444607-113444607 on Build GRCh38
rs931801685 Chr.13: 113444335-113444335 on Build GRCh38
rs185858041 Chr.13: 113444373-113444373 on Build GRCh38
rs1031341284 Chr.13: 113444232-113444232 on Build GRCh38
rs143393814 Chr.13: 113444438-113444438 on Build GRCh38
rs768957392 Chr.13: 113444638-113444638 on Build GRCh38
rs998652439 Chr.13: 113444223-113444223 on Build GRCh38
rs112656573 Chr.13: 113444400-113444400 on Build GRCh38
rs558151919 Chr.13: 113444552-113444552 on Build GRCh38
rs778294599 Chr.13: 113444565-113444566 on Build GRCh38
rs902637862 Chr.13: 113444548-113444548 on Build GRCh38
rs1008330199 Chr.13: 113444343-113444343 on Build GRCh38
rs189928511 Chr.13: 113444420-113444420 on Build GRCh38
rs982390721 Chr.13: 113444563-113444563 on Build GRCh38
rs371791782 Chr.13: 113444403-113444403 on Build GRCh38
rs113016990 Chr.13: 113444250-113444250 on Build GRCh38
rs181145159 Chr.13: 113444452-113444452 on Build GRCh38
rs138609269 Chr.13: 113444534-113444534 on Build GRCh38
rs970509860 Chr.13: 113444254-113444254 on Build GRCh38
rs767779224 Chr.13: 113444376-113444376 on Build GRCh38
rs73579439 Chr.13: 113444340-113444340 on Build GRCh38
rs935531825 Chr.13: 113444580-113444580 on Build GRCh38
rs949694622 Chr.13: 113444357-113444357 on Build GRCh38
rs1003708036 Chr.13: 113444642-113444642 on Build GRCh38
rs151314929 Chr.13: 113444512-113444512 on Build GRCh38
rs755794055 Chr.13: 113444586-113444586 on Build GRCh38
rs186128422 Chr.13: 113444647-113444647 on Build GRCh38
rs770859746 Chr.13: 113444311-113444311 on Build GRCh38
rs576528673 Chr.13: 113444557-113444557 on Build GRCh38
rs755263614 Chr.13: 113444374-113444374 on Build GRCh38
rs973078435 Chr.13: 113444474-113444474 on Build GRCh38
rs991041466 Chr.13: 113444234-113444234 on Build GRCh38
rs569395167 Chr.13: 113444362-113444362 on Build GRCh38
rs529174452 Chr.13: 113444543-113444543 on Build GRCh38
rs902501614 Chr.13: 113444365-113444365 on Build GRCh38
rs767599544 Chr.13: 113444530-113444530 on Build GRCh38
rs529516492 Chr.13: 113444263-113444263 on Build GRCh38
rs576646323 Chr.13: 113444211-113444211 on Build GRCh38
rs749116851 Chr.13: 113444592-113444592 on Build GRCh38
rs1027341502 Chr.13: 113444660-113444660 on Build GRCh38
rs1019549462 Chr.13: 113444654-113444654 on Build GRCh38
rs745793071 Chr.13: 113444594-113444594 on Build GRCh38
rs774381207 Chr.13: 113444515-113444515 on Build GRCh38
rs750908703 Chr.13: 113444566-113444566 on Build GRCh38
rs532788136 Chr.13: 113444318-113444318 on Build GRCh38
rs919981533 Chr.13: 113444322-113444322 on Build GRCh38
rs113346581 Chr.13: 113444653-113444653 on Build GRCh38
rs747805705 Chr.13: 113444511-113444511 on Build GRCh38
rs1041038001 Chr.13: 113444346-113444346 on Build GRCh38
rs560035451 Chr.13: 113444307-113444307 on Build GRCh38
rs776312100 Chr.13: 113444402-113444402 on Build GRCh38
rs766358734 Chr.13: 113444421-113444421 on Build GRCh38
rs759648461 Chr.13: 113444437-113444437 on Build GRCh38
rs769609694 Chr.13: 113444405-113444405 on Build GRCh38
rs780750444 Chr.13: 113444574-113444574 on Build GRCh38
rs966928323 Chr.13: 113444214-113444214 on Build GRCh38
rs749767076 Chr.13: 113444397-113444397 on Build GRCh38
rs140723775 Chr.13: 113444514-113444514 on Build GRCh38
rs773064874 Chr.13: 113444409-113444409 on Build GRCh38
rs920552336 Chr.13: 113444313-113444313 on Build GRCh38
rs372352299 Chr.13: 113444482-113444482 on Build GRCh38
rs759739272 Chr.13: 113444516-113444516 on Build GRCh38
rs748215780 Chr.13: 113444502-113444502 on Build GRCh38
rs999792056 Chr.13: 113444221-113444221 on Build GRCh38
rs920474832 Chr.13: 113444505-113444505 on Build GRCh38
rs999549132 Chr.13: 113444372-113444372 on Build GRCh38

View Full Product Details