Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113823232-113823724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS6
Entrez Gene ID
2621
Gene Name
growth arrest specific 6
Gene Aliases
AXLLG, AXSF
UniGene
Hs.646346
Gene Chromosome Location
Chr.13: 113820549-113864103 on Build GRCh38
Transcripts
NM_000820.3 NR_044995.2

Gene Symbol
GAS6-AS1
Entrez Gene ID
650669
Gene Name
GAS6 antisense RNA 1
Gene Aliases
-
UniGene
Hs.369201
Gene Chromosome Location
Chr.13: 113815610-113842841 on Build GRCh38
Transcripts
NM_000820.3 NR_044995.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4046238 c.1596C>T p.A532A Chr.13: 113823432-113823432 on Build GRCh38
4809386 c.1416G>C p.W472C Chr.13: 113823450-113823450 on Build GRCh38
3981947 c.1740G>T p.V580V Chr.13: 113823417-113823417 on Build GRCh38
4046241 c.1480C>T p.R494W Chr.13: 113823548-113823548 on Build GRCh38
4046239 c.1725C>T p.A575A Chr.13: 113823432-113823432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780595570 Chr.13: 113823337-113823337 on Build GRCh38
rs571029386 Chr.13: 113823487-113823487 on Build GRCh38
rs200276205 Chr.13: 113823557-113823557 on Build GRCh38
rs545444776 Chr.13: 113823658-113823658 on Build GRCh38
rs528060429 Chr.13: 113823444-113823444 on Build GRCh38
rs60377661 Chr.13: 113823338-113823338 on Build GRCh38
rs765354911 Chr.13: 113823479-113823479 on Build GRCh38
rs74118435 Chr.13: 113823531-113823531 on Build GRCh38
rs750488474 Chr.13: 113823394-113823394 on Build GRCh38
rs374982313 Chr.13: 113823393-113823393 on Build GRCh38
rs781757797 Chr.13: 113823509-113823509 on Build GRCh38
rs973632123 Chr.13: 113823399-113823399 on Build GRCh38
rs374225474 Chr.13: 113823331-113823331 on Build GRCh38
rs199759304 Chr.13: 113823581-113823581 on Build GRCh38
rs756553660 Chr.13: 113823519-113823519 on Build GRCh38
rs375230851 Chr.13: 113823351-113823351 on Build GRCh38
rs757310622 Chr.13: 113823328-113823328 on Build GRCh38
rs973372016 Chr.13: 113823265-113823265 on Build GRCh38
rs940938512 Chr.13: 113823260-113823260 on Build GRCh38
rs189726423 Chr.13: 113823324-113823324 on Build GRCh38
rs770488795 Chr.13: 113823436-113823436 on Build GRCh38
rs141432096 Chr.13: 113823534-113823534 on Build GRCh38
rs368135800 Chr.13: 113823352-113823352 on Build GRCh38
rs776678863 Chr.13: 113823548-113823548 on Build GRCh38
rs1015373766 Chr.13: 113823539-113823539 on Build GRCh38
rs535033524 Chr.13: 113823623-113823623 on Build GRCh38
rs994966110 Chr.13: 113823701-113823701 on Build GRCh38
rs966465716 Chr.13: 113823621-113823621 on Build GRCh38
rs763176753 Chr.13: 113823560-113823560 on Build GRCh38
rs149090898 Chr.13: 113823480-113823480 on Build GRCh38
rs944503418 Chr.13: 113823287-113823287 on Build GRCh38
rs1051971242 Chr.13: 113823603-113823603 on Build GRCh38
rs7987377 Chr.13: 113823644-113823644 on Build GRCh38
rs760503213 Chr.13: 113823363-113823363 on Build GRCh38
rs761480661 Chr.13: 113823549-113823549 on Build GRCh38
rs181106746 Chr.13: 113823671-113823671 on Build GRCh38
rs375533052 Chr.13: 113823671-113823671 on Build GRCh38
rs1041570900 Chr.13: 113823298-113823298 on Build GRCh38
rs368447635 Chr.13: 113823437-113823437 on Build GRCh38
rs866625407 Chr.13: 113823299-113823299 on Build GRCh38
rs992107733 Chr.13: 113823522-113823522 on Build GRCh38
rs758116137 Chr.13: 113823596-113823596 on Build GRCh38
rs527335627 Chr.13: 113823285-113823285 on Build GRCh38
rs766380178 Chr.13: 113823563-113823563 on Build GRCh38
rs772561882 Chr.13: 113823333-113823333 on Build GRCh38
rs1000057786 Chr.13: 113823310-113823310 on Build GRCh38
rs761723369 Chr.13: 113823476-113823476 on Build GRCh38
rs763815291 Chr.13: 113823381-113823381 on Build GRCh38
rs1020452461 Chr.13: 113823327-113823327 on Build GRCh38
rs113146617 Chr.13: 113823343-113823343 on Build GRCh38
rs778664550 Chr.13: 113823529-113823529 on Build GRCh38
rs1023990013 Chr.13: 113823492-113823492 on Build GRCh38
rs747546889 Chr.13: 113823421-113823421 on Build GRCh38
rs752007610 Chr.13: 113823496-113823496 on Build GRCh38
rs752994393 Chr.13: 113823517-113823517 on Build GRCh38
rs553377066 Chr.13: 113823642-113823642 on Build GRCh38
rs781425318 Chr.13: 113823431-113823431 on Build GRCh38
rs755656552 Chr.13: 113823378-113823380 on Build GRCh38
rs757423394 Chr.13: 113823391-113823391 on Build GRCh38
rs764098672 Chr.13: 113823267-113823267 on Build GRCh38
rs7992146 Chr.13: 113823530-113823530 on Build GRCh38
rs878991605 Chr.13: 113823650-113823650 on Build GRCh38
rs1042224033 Chr.13: 113823454-113823454 on Build GRCh38
rs528731495 Chr.13: 113823704-113823704 on Build GRCh38
rs755159941 Chr.13: 113823587-113823587 on Build GRCh38
rs144928601 Chr.13: 113823448-113823448 on Build GRCh38
rs758520989 Chr.13: 113823397-113823397 on Build GRCh38
rs759722080 Chr.13: 113823573-113823573 on Build GRCh38
rs548946951 Chr.13: 113823456-113823456 on Build GRCh38
rs768463188 Chr.13: 113823541-113823541 on Build GRCh38
rs776284665 Chr.13: 113823467-113823467 on Build GRCh38
rs931992482 Chr.13: 113823284-113823284 on Build GRCh38
rs755446922 Chr.13: 113823497-113823497 on Build GRCh38
rs574119601 Chr.13: 113823588-113823588 on Build GRCh38
rs376994871 Chr.13: 113823355-113823355 on Build GRCh38
rs149907010 Chr.13: 113823415-113823415 on Build GRCh38
rs1016241854 Chr.13: 113823624-113823624 on Build GRCh38
rs753040178 Chr.13: 113823589-113823589 on Build GRCh38
rs898254597 Chr.13: 113823697-113823697 on Build GRCh38
rs147253799 Chr.13: 113823669-113823669 on Build GRCh38
rs572075008 Chr.13: 113823670-113823670 on Build GRCh38
rs911884148 Chr.13: 113823611-113823611 on Build GRCh38
rs924734010 Chr.13: 113823639-113823639 on Build GRCh38
rs977758101 Chr.13: 113823543-113823543 on Build GRCh38
rs758433119 Chr.13: 113823488-113823488 on Build GRCh38
rs751637569 Chr.13: 113823566-113823566 on Build GRCh38
rs767552365 Chr.13: 113823360-113823360 on Build GRCh38
rs569021446 Chr.13: 113823556-113823556 on Build GRCh38
rs755323426 Chr.13: 113823428-113823428 on Build GRCh38
rs559872982 Chr.13: 113823286-113823286 on Build GRCh38
rs920886286 Chr.13: 113823426-113823426 on Build GRCh38
rs768943498 Chr.13: 113823342-113823342 on Build GRCh38
rs370213484 Chr.13: 113823361-113823361 on Build GRCh38
rs370340821 Chr.13: 113823443-113823443 on Build GRCh38
rs754031122 Chr.13: 113823390-113823390 on Build GRCh38
rs201975506 Chr.13: 113823377-113823377 on Build GRCh38
rs766469912 Chr.13: 113823489-113823489 on Build GRCh38
rs111811933 Chr.13: 113823325-113823325 on Build GRCh38
rs750027416 Chr.13: 113823592-113823592 on Build GRCh38
rs1004323368 Chr.13: 113823680-113823680 on Build GRCh38
rs771766365 Chr.13: 113823501-113823501 on Build GRCh38
rs1054221995 Chr.13: 113823643-113823643 on Build GRCh38
rs763879682 Chr.13: 113823463-113823463 on Build GRCh38
rs561260552 Chr.13: 113823681-113823681 on Build GRCh38
rs201938938 Chr.13: 113823432-113823432 on Build GRCh38

View Full Product Details