Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 114243774-114244276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC16
Entrez Gene ID
8881
Gene Name
cell division cycle 16
Gene Aliases
ANAPC6, APC6, CDC16Hs, CUT9
UniGene
Hs.374127
Gene Chromosome Location
Chr.13: 114234845-114272675 on Build GRCh38
Transcripts
NM_001078645.1 NM_001318517.1 NM_001318518.1 NM_003903.3 XM_005266206.1 XM_006719992.1 XM_006719993.1 XM_011534851.1 XM_011534853.1 XM_017020830.1 XM_017020831.1 XM_017020832.1 XM_017020833.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6384849 c.650A>C p.E217A Chr.13: 114243872-114243872 on Build GRCh38
4760396 c.667T>C p.S223P Chr.13: 114243889-114243889 on Build GRCh38
6247369 c.687G>A p.E229E Chr.13: 114243909-114243909 on Build GRCh38
5376463 c.685G>A p.E229K Chr.13: 114243907-114243907 on Build GRCh38
695909 c.721C>A p.H241N Chr.13: 114243943-114243943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140428792 Chr.13: 114244021-114244021 on Build GRCh38
rs755433387 Chr.13: 114243924-114243924 on Build GRCh38
rs745747331 Chr.13: 114243982-114243982 on Build GRCh38
rs765187512 Chr.13: 114244032-114244032 on Build GRCh38
rs758627823 Chr.13: 114243822-114243822 on Build GRCh38
rs746806928 Chr.13: 114244006-114244006 on Build GRCh38
rs977887052 Chr.13: 114244129-114244129 on Build GRCh38
rs765367805 Chr.13: 114243906-114243906 on Build GRCh38
rs573527286 Chr.13: 114244040-114244040 on Build GRCh38
rs529238698 Chr.13: 114244176-114244176 on Build GRCh38
rs933886474 Chr.13: 114243895-114243895 on Build GRCh38
rs188252463 Chr.13: 114244113-114244113 on Build GRCh38
rs771725529 Chr.13: 114243864-114243864 on Build GRCh38
rs982708100 Chr.13: 114244205-114244205 on Build GRCh38
rs1024387921 Chr.13: 114244030-114244030 on Build GRCh38
rs976060489 Chr.13: 114244146-114244146 on Build GRCh38
rs773362419 Chr.13: 114244033-114244033 on Build GRCh38
rs761863907 Chr.13: 114243900-114243900 on Build GRCh38
rs368309403 Chr.13: 114243825-114243825 on Build GRCh38
rs776772943 Chr.13: 114244022-114244022 on Build GRCh38
rs546359133 Chr.13: 114244094-114244098 on Build GRCh38
rs941138461 Chr.13: 114243944-114243946 on Build GRCh38
rs1001880815 Chr.13: 114244016-114244016 on Build GRCh38
rs181861703 Chr.13: 114244042-114244042 on Build GRCh38
rs200625374 Chr.13: 114243831-114243831 on Build GRCh38
rs936231335 Chr.13: 114243948-114243948 on Build GRCh38
rs750826173 Chr.13: 114243907-114243907 on Build GRCh38
rs778518639 Chr.13: 114243975-114243975 on Build GRCh38
rs766323310 Chr.13: 114243840-114243840 on Build GRCh38
rs570514626 Chr.13: 114243927-114243927 on Build GRCh38
rs775382065 Chr.13: 114243875-114243875 on Build GRCh38
rs1054616281 Chr.13: 114244202-114244202 on Build GRCh38
rs953856919 Chr.13: 114244079-114244079 on Build GRCh38
rs1004304921 Chr.13: 114244047-114244047 on Build GRCh38
rs1028268982 Chr.13: 114244060-114244060 on Build GRCh38
rs964701255 Chr.13: 114243852-114243852 on Build GRCh38
rs757370987 Chr.13: 114243809-114243809 on Build GRCh38
rs80302281 Chr.13: 114244046-114244046 on Build GRCh38
rs756657620 Chr.13: 114243961-114243961 on Build GRCh38
rs974006616 Chr.13: 114243798-114243798 on Build GRCh38
rs776908624 Chr.13: 114243891-114243891 on Build GRCh38
rs775333815 Chr.13: 114243992-114243992 on Build GRCh38
rs764272207 Chr.13: 114243884-114243884 on Build GRCh38
rs544409136 Chr.13: 114244059-114244059 on Build GRCh38
rs373947188 Chr.13: 114243950-114243950 on Build GRCh38
rs145132276 Chr.13: 114244178-114244178 on Build GRCh38
rs563373928 Chr.13: 114244233-114244236 on Build GRCh38
rs756822413 Chr.13: 114244169-114244169 on Build GRCh38
rs750994700 Chr.13: 114243819-114243819 on Build GRCh38
rs753668256 Chr.13: 114244165-114244165 on Build GRCh38
rs760516771 Chr.13: 114243876-114243876 on Build GRCh38
rs764020668 Chr.13: 114244039-114244044 on Build GRCh38
rs770624827 Chr.13: 114243859-114243859 on Build GRCh38
rs912404358 Chr.13: 114244104-114244104 on Build GRCh38
rs755852890 Chr.13: 114243830-114243830 on Build GRCh38
rs766900868 Chr.13: 114244164-114244173 on Build GRCh38
rs540205836 Chr.13: 114244183-114244183 on Build GRCh38
rs533287763 Chr.13: 114244055-114244055 on Build GRCh38
rs780468090 Chr.13: 114243824-114243824 on Build GRCh38
rs1017657708 Chr.13: 114244106-114244106 on Build GRCh38
rs112683123 Chr.13: 114243908-114243908 on Build GRCh38
rs181601253 Chr.13: 114244252-114244252 on Build GRCh38
rs986664871 Chr.13: 114244080-114244080 on Build GRCh38
rs17291173 Chr.13: 114243909-114243909 on Build GRCh38
rs765654858 Chr.13: 114243810-114243810 on Build GRCh38
rs755093340 Chr.13: 114244007-114244007 on Build GRCh38
rs751823995 Chr.13: 114243979-114243982 on Build GRCh38
rs777555590 Chr.13: 114243929-114243929 on Build GRCh38
rs748774580 Chr.13: 114243845-114243845 on Build GRCh38
rs143058360 Chr.13: 114244111-114244111 on Build GRCh38
rs945052615 Chr.13: 114243903-114243903 on Build GRCh38
rs183920814 Chr.13: 114244058-114244058 on Build GRCh38
rs772026621 Chr.13: 114243988-114243988 on Build GRCh38
rs745823334 Chr.13: 114243863-114243863 on Build GRCh38
rs150398862 Chr.13: 114244027-114244027 on Build GRCh38
rs556404196 Chr.13: 114244131-114244131 on Build GRCh38
rs368136915 Chr.13: 114244036-114244036 on Build GRCh38
rs941220614 Chr.13: 114244243-114244243 on Build GRCh38
rs774494396 Chr.13: 114243922-114243922 on Build GRCh38

View Full Product Details