Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15438350-15438845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTRC
Entrez Gene ID
11330
Gene Name
chymotrypsin C
Gene Aliases
CLCR, ELA4
UniGene
Hs.631869
Gene Chromosome Location
Chr.1: 15438442-15449247 on Build GRCh38
Transcripts
NM_007272.2 XM_011540550.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
897554 c.27G>A p.A9A Chr.1: 15438491-15438491 on Build GRCh38
6301933 c.28C>T p.L10F Chr.1: 15438492-15438492 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371050890 Chr.1: 15438760-15438760 on Build GRCh38
rs1027307249 Chr.1: 15438673-15438673 on Build GRCh38
rs775301182 Chr.1: 15438438-15438438 on Build GRCh38
rs944802371 Chr.1: 15438536-15438536 on Build GRCh38
rs764761065 Chr.1: 15438764-15438764 on Build GRCh38
rs959377063 Chr.1: 15438625-15438625 on Build GRCh38
rs1041938209 Chr.1: 15438812-15438812 on Build GRCh38
rs567507979 Chr.1: 15438698-15438698 on Build GRCh38
rs551442042 Chr.1: 15438588-15438588 on Build GRCh38
rs758984378 Chr.1: 15438493-15438493 on Build GRCh38
rs536545126 Chr.1: 15438707-15438707 on Build GRCh38
rs188758070 Chr.1: 15438541-15438541 on Build GRCh38
rs2272910 Chr.1: 15438767-15438767 on Build GRCh38
rs767349844 Chr.1: 15438486-15438486 on Build GRCh38
rs144717165 Chr.1: 15438373-15438373 on Build GRCh38
rs945395435 Chr.1: 15438661-15438661 on Build GRCh38
rs200861519 Chr.1: 15438546-15438546 on Build GRCh38
rs1054376575 Chr.1: 15438576-15438576 on Build GRCh38
rs893416463 Chr.1: 15438581-15438582 on Build GRCh38
rs183658182 Chr.1: 15438406-15438406 on Build GRCh38
rs774163002 Chr.1: 15438488-15438488 on Build GRCh38
rs374821228 Chr.1: 15438491-15438491 on Build GRCh38
rs893113874 Chr.1: 15438779-15438779 on Build GRCh38
rs769703596 Chr.1: 15438436-15438436 on Build GRCh38
rs2272909 Chr.1: 15438679-15438679 on Build GRCh38
rs907710946 Chr.1: 15438709-15438709 on Build GRCh38
rs990515037 Chr.1: 15438642-15438642 on Build GRCh38
rs781396039 Chr.1: 15438516-15438516 on Build GRCh38
rs765895922 Chr.1: 15438569-15438569 on Build GRCh38
rs776678963 Chr.1: 15438425-15438425 on Build GRCh38
rs201225064 Chr.1: 15438539-15438539 on Build GRCh38
rs192819418 Chr.1: 15438805-15438805 on Build GRCh38
rs531236651 Chr.1: 15438584-15438584 on Build GRCh38
rs371595328 Chr.1: 15438393-15438393 on Build GRCh38
rs115057154 Chr.1: 15438697-15438697 on Build GRCh38
rs369832904 Chr.1: 15438547-15438547 on Build GRCh38
rs890047179 Chr.1: 15438510-15438510 on Build GRCh38
rs1020394621 Chr.1: 15438374-15438374 on Build GRCh38
rs761548318 Chr.1: 15438475-15438475 on Build GRCh38
rs914658483 Chr.1: 15438777-15438777 on Build GRCh38
rs371272589 Chr.1: 15438410-15438410 on Build GRCh38
rs774324080 Chr.1: 15438470-15438470 on Build GRCh38
rs769782731 Chr.1: 15438532-15438532 on Build GRCh38
rs771193384 Chr.1: 15438418-15438418 on Build GRCh38
rs746013735 Chr.1: 15438435-15438435 on Build GRCh38
rs552472301 Chr.1: 15438759-15438759 on Build GRCh38
rs923803443 Chr.1: 15438741-15438741 on Build GRCh38
rs571381490 Chr.1: 15438621-15438621 on Build GRCh38
rs762757220 Chr.1: 15438455-15438455 on Build GRCh38
rs1034815526 Chr.1: 15438613-15438613 on Build GRCh38
rs368967504 Chr.1: 15438562-15438562 on Build GRCh38
rs376195831 Chr.1: 15438398-15438398 on Build GRCh38
rs113591544 Chr.1: 15438391-15438391 on Build GRCh38
rs747431847 Chr.1: 15438503-15438503 on Build GRCh38
rs933703 Chr.1: 15438477-15438477 on Build GRCh38
rs374390708 Chr.1: 15438570-15438570 on Build GRCh38
rs537865387 Chr.1: 15438810-15438810 on Build GRCh38
rs990911316 Chr.1: 15438632-15438632 on Build GRCh38
rs778090425 Chr.1: 15438496-15438496 on Build GRCh38
rs1050217802 Chr.1: 15438818-15438818 on Build GRCh38
rs982022460 Chr.1: 15438728-15438728 on Build GRCh38
rs142888713 Chr.1: 15438528-15438528 on Build GRCh38
rs369827801 Chr.1: 15438511-15438511 on Build GRCh38
rs749888586 Chr.1: 15438489-15438489 on Build GRCh38
rs922072223 Chr.1: 15438743-15438743 on Build GRCh38
rs564455614 Chr.1: 15438514-15438514 on Build GRCh38
rs544668774 Chr.1: 15438465-15438465 on Build GRCh38
rs374402898 Chr.1: 15438582-15438582 on Build GRCh38
rs772818172 Chr.1: 15438490-15438490 on Build GRCh38
rs952456878 Chr.1: 15438592-15438592 on Build GRCh38
rs932054018 Chr.1: 15438774-15438774 on Build GRCh38
rs536481690 Chr.1: 15438776-15438776 on Build GRCh38

View Full Product Details