Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15445691-15446217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTRC
Entrez Gene ID
11330
Gene Name
chymotrypsin C
Gene Aliases
CLCR, ELA4
UniGene
Hs.631869
Gene Chromosome Location
Chr.1: 15438442-15449247 on Build GRCh38
Transcripts
NM_007272.2 XM_011540550.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7260267 c.792+10G>A p.? Chr.1: 15445759-15445759 on Build GRCh38
6360985 c.788A>T p.N263I Chr.1: 15445745-15445745 on Build GRCh38
6049065 c.748G>T p.V250L Chr.1: 15445705-15445705 on Build GRCh38
4987109 c.760C>T p.R254W Chr.1: 15445717-15445717 on Build GRCh38
1985350 c.747G>A p.P249P Chr.1: 15445704-15445704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111511942 Chr.1: 15445917-15445917 on Build GRCh38
rs764039167 Chr.1: 15445748-15445748 on Build GRCh38
rs979088679 Chr.1: 15446128-15446128 on Build GRCh38
rs78012209 Chr.1: 15445962-15445963 on Build GRCh38
rs752111877 Chr.1: 15445883-15445883 on Build GRCh38
rs867213434 Chr.1: 15445935-15445935 on Build GRCh38
rs536401908 Chr.1: 15446134-15446134 on Build GRCh38
rs768653391 Chr.1: 15445798-15445798 on Build GRCh38
rs777187660 Chr.1: 15445737-15445737 on Build GRCh38
rs201965913 Chr.1: 15445874-15445881 on Build GRCh38
rs567750027 Chr.1: 15446097-15446097 on Build GRCh38
rs35392581 Chr.1: 15445959-15445960 on Build GRCh38
rs766640945 Chr.1: 15446085-15446087 on Build GRCh38
rs772573958 Chr.1: 15445724-15445724 on Build GRCh38
rs555727973 Chr.1: 15445793-15445794 on Build GRCh38
rs121909293 Chr.1: 15445717-15445717 on Build GRCh38
rs144636721 Chr.1: 15445879-15445890 on Build GRCh38
rs561053914 Chr.1: 15445817-15445817 on Build GRCh38
rs923128612 Chr.1: 15445911-15445911 on Build GRCh38
rs371799801 Chr.1: 15445761-15445761 on Build GRCh38
rs978604492 Chr.1: 15446054-15446054 on Build GRCh38
rs756384572 Chr.1: 15445743-15445744 on Build GRCh38
rs12755729 Chr.1: 15445993-15445993 on Build GRCh38
rs143842117 Chr.1: 15445982-15445982 on Build GRCh38
rs12409952 Chr.1: 15445975-15445975 on Build GRCh38
rs780270629 Chr.1: 15445780-15445780 on Build GRCh38
rs111372278 Chr.1: 15445720-15445720 on Build GRCh38
rs12754584 Chr.1: 15446018-15446018 on Build GRCh38
rs767275938 Chr.1: 15445796-15445796 on Build GRCh38
rs960861891 Chr.1: 15446104-15446104 on Build GRCh38
rs967745620 Chr.1: 15446119-15446119 on Build GRCh38
rs71000402 Chr.1: 15445974-15445975 on Build GRCh38
rs778229929 Chr.1: 15445836-15445836 on Build GRCh38
rs934703556 Chr.1: 15445801-15445801 on Build GRCh38
rs200406696 Chr.1: 15445726-15445726 on Build GRCh38
rs1047187609 Chr.1: 15445923-15445923 on Build GRCh38
rs532265778 Chr.1: 15445945-15445945 on Build GRCh38
rs9429242 Chr.1: 15445963-15445963 on Build GRCh38
rs529469818 Chr.1: 15446105-15446105 on Build GRCh38
rs751482739 Chr.1: 15445751-15445751 on Build GRCh38
rs146762370 Chr.1: 15446003-15446004 on Build GRCh38
rs142877180 Chr.1: 15446007-15446008 on Build GRCh38
rs757062706 Chr.1: 15445714-15445714 on Build GRCh38
rs1020672584 Chr.1: 15446108-15446108 on Build GRCh38
rs1023252013 Chr.1: 15445840-15445843 on Build GRCh38
rs543606478 Chr.1: 15445865-15445865 on Build GRCh38
rs954433993 Chr.1: 15445959-15445962 on Build GRCh38
rs910213380 Chr.1: 15446182-15446182 on Build GRCh38
rs769975164 Chr.1: 15445745-15445745 on Build GRCh38
rs971755141 Chr.1: 15445944-15445947 on Build GRCh38
rs895781004 Chr.1: 15445829-15445829 on Build GRCh38
rs200931849 Chr.1: 15445899-15445900 on Build GRCh38
rs780783336 Chr.1: 15445715-15445715 on Build GRCh38
rs748753747 Chr.1: 15445719-15445719 on Build GRCh38
rs574518975 Chr.1: 15445856-15445856 on Build GRCh38
rs979036266 Chr.1: 15445778-15445778 on Build GRCh38
rs761777960 Chr.1: 15445777-15445777 on Build GRCh38
rs563202820 Chr.1: 15445916-15445916 on Build GRCh38
rs55945515 Chr.1: 15446017-15446018 on Build GRCh38
rs527722814 Chr.1: 15446027-15446027 on Build GRCh38
rs759821387 Chr.1: 15445740-15445740 on Build GRCh38
rs756754404 Chr.1: 15445835-15445835 on Build GRCh38
rs755811899 Chr.1: 15445718-15445718 on Build GRCh38
rs965028310 Chr.1: 15446188-15446188 on Build GRCh38
rs1057160478 Chr.1: 15445818-15445821 on Build GRCh38
rs888683769 Chr.1: 15445896-15445896 on Build GRCh38
rs113257816 Chr.1: 15445712-15445712 on Build GRCh38
rs750274349 Chr.1: 15445797-15445797 on Build GRCh38
rs917314635 Chr.1: 15446180-15446180 on Build GRCh38
rs762952174 Chr.1: 15445746-15445746 on Build GRCh38
rs947267758 Chr.1: 15445734-15445734 on Build GRCh38
rs961784194 Chr.1: 15445929-15445929 on Build GRCh38
rs386628830 Chr.1: 15445962-15445963 on Build GRCh38
rs540753875 Chr.1: 15445735-15445735 on Build GRCh38
rs778166573 Chr.1: 15445725-15445725 on Build GRCh38
rs370181931 Chr.1: 15445958-15445958 on Build GRCh38
rs1040647018 Chr.1: 15445905-15445905 on Build GRCh38
rs910491297 Chr.1: 15445994-15445994 on Build GRCh38
rs1038353852 Chr.1: 15445968-15445968 on Build GRCh38
rs910475501 Chr.1: 15445793-15445793 on Build GRCh38
rs146241668 Chr.1: 15445960-15445961 on Build GRCh38
rs72643663 Chr.1: 15446072-15446072 on Build GRCh38

View Full Product Details