Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15476166-15476668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELA2B
Entrez Gene ID
51032
Gene Name
chymotrypsin like elastase family member 2B
Gene Aliases
ELA2B
UniGene
Hs.631871
Gene Chromosome Location
Chr.1: 15476101-15491400 on Build GRCh38
Transcripts
NM_015849.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4223487 c.10T>C p.S4P Chr.1: 15476474-15476474 on Build GRCh38
1200676 c.67G>A p.A23T Chr.1: 15476483-15476483 on Build GRCh38
424196 c.59C>T p.S20F Chr.1: 15476475-15476475 on Build GRCh38
3385289 c.66C>T p.Y22Y Chr.1: 15476482-15476482 on Build GRCh38
4223486 c.58T>C p.S20P Chr.1: 15476474-15476474 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548200314 Chr.1: 15476622-15476622 on Build GRCh38
rs111363198 Chr.1: 15476523-15476523 on Build GRCh38
rs770231915 Chr.1: 15476581-15476581 on Build GRCh38
rs920564377 Chr.1: 15476240-15476240 on Build GRCh38
rs147610320 Chr.1: 15476531-15476531 on Build GRCh38
rs773679354 Chr.1: 15476483-15476483 on Build GRCh38
rs202244887 Chr.1: 15476477-15476477 on Build GRCh38
rs376357865 Chr.1: 15476412-15476412 on Build GRCh38
rs192348387 Chr.1: 15476315-15476315 on Build GRCh38
rs28421187 Chr.1: 15476467-15476467 on Build GRCh38
rs550716870 Chr.1: 15476504-15476504 on Build GRCh38
rs748805268 Chr.1: 15476432-15476432 on Build GRCh38
rs758303295 Chr.1: 15476457-15476457 on Build GRCh38
rs115541368 Chr.1: 15476293-15476293 on Build GRCh38
rs186936455 Chr.1: 15476326-15476326 on Build GRCh38
rs571055831 Chr.1: 15476404-15476404 on Build GRCh38
rs1034474869 Chr.1: 15476602-15476602 on Build GRCh38
rs747448590 Chr.1: 15476573-15476573 on Build GRCh38
rs199955709 Chr.1: 15476554-15476554 on Build GRCh38
rs763530195 Chr.1: 15476429-15476429 on Build GRCh38
rs548295328 Chr.1: 15476316-15476316 on Build GRCh38
rs369630964 Chr.1: 15476428-15476428 on Build GRCh38
rs143905636 Chr.1: 15476509-15476509 on Build GRCh38
rs745515344 Chr.1: 15476471-15476471 on Build GRCh38
rs368888301 Chr.1: 15476485-15476485 on Build GRCh38
rs540522507 Chr.1: 15476599-15476599 on Build GRCh38
rs183383008 Chr.1: 15476245-15476245 on Build GRCh38
rs922050142 Chr.1: 15476513-15476513 on Build GRCh38
rs754424208 Chr.1: 15476551-15476551 on Build GRCh38
rs1010504914 Chr.1: 15476535-15476535 on Build GRCh38
rs750202039 Chr.1: 15476524-15476524 on Build GRCh38
rs769338268 Chr.1: 15476472-15476472 on Build GRCh38
rs759744443 Chr.1: 15476500-15476500 on Build GRCh38
rs566478686 Chr.1: 15476470-15476470 on Build GRCh38
rs900574612 Chr.1: 15476325-15476325 on Build GRCh38
rs907670597 Chr.1: 15476292-15476292 on Build GRCh38
rs746249389 Chr.1: 15476576-15476576 on Build GRCh38
rs754807126 Chr.1: 15476333-15476333 on Build GRCh38
rs1003257836 Chr.1: 15476426-15476426 on Build GRCh38
rs751526115 Chr.1: 15476516-15476516 on Build GRCh38
rs185079918 Chr.1: 15476623-15476623 on Build GRCh38
rs762317283 Chr.1: 15476212-15476212 on Build GRCh38
rs765294081 Chr.1: 15476429-15476429 on Build GRCh38
rs749012037 Chr.1: 15476547-15476547 on Build GRCh38
rs769337523 Chr.1: 15476196-15476196 on Build GRCh38
rs761125750 Chr.1: 15476597-15476598 on Build GRCh38
rs189080505 Chr.1: 15476280-15476280 on Build GRCh38
rs772842415 Chr.1: 15476393-15476393 on Build GRCh38
rs150801399 Chr.1: 15476520-15476520 on Build GRCh38
rs137951823 Chr.1: 15476493-15476493 on Build GRCh38
rs559330194 Chr.1: 15476246-15476246 on Build GRCh38
rs996215569 Chr.1: 15476337-15476337 on Build GRCh38
rs111952850 Chr.1: 15476341-15476341 on Build GRCh38
rs375665242 Chr.1: 15476201-15476201 on Build GRCh38
rs762072378 Chr.1: 15476643-15476643 on Build GRCh38
rs771440993 Chr.1: 15476574-15476574 on Build GRCh38
rs754038650 Chr.1: 15476427-15476427 on Build GRCh38
rs765398067 Chr.1: 15476501-15476501 on Build GRCh38
rs145776598 Chr.1: 15476521-15476521 on Build GRCh38
rs750962246 Chr.1: 15476565-15476567 on Build GRCh38
rs533723072 Chr.1: 15476418-15476418 on Build GRCh38
rs761060552 Chr.1: 15476416-15476416 on Build GRCh38
rs894904737 Chr.1: 15476610-15476610 on Build GRCh38
rs762536068 Chr.1: 15476414-15476414 on Build GRCh38
rs191347695 Chr.1: 15476361-15476361 on Build GRCh38
rs202079732 Chr.1: 15476586-15476586 on Build GRCh38
rs990536910 Chr.1: 15476439-15476439 on Build GRCh38
rs72643675 Chr.1: 15476430-15476430 on Build GRCh38
rs751583802 Chr.1: 15476469-15476469 on Build GRCh38
rs779561397 Chr.1: 15476474-15476474 on Build GRCh38
rs764268530 Chr.1: 15476596-15476596 on Build GRCh38
rs182357930 Chr.1: 15476323-15476323 on Build GRCh38
rs535515233 Chr.1: 15476484-15476484 on Build GRCh38
rs1040453283 Chr.1: 15476296-15476296 on Build GRCh38
rs780805131 Chr.1: 15476340-15476340 on Build GRCh38
rs375371978 Chr.1: 15476463-15476463 on Build GRCh38
rs772594074 Chr.1: 15476482-15476482 on Build GRCh38

View Full Product Details