Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15559234-15559757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC16
Entrez Gene ID
23341
Gene Name
DnaJ heat shock protein family (Hsp40) member C16
Gene Aliases
-
UniGene
Hs.655410
Gene Chromosome Location
Chr.1: 15526694-15571733 on Build GRCh38
Transcripts
NM_001287811.1 NM_015291.3 NR_109898.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7183557 c.1131G>C p.V377V Chr.1: 15559633-15559633 on Build GRCh38
1661517 c.1027C>G p.R343G Chr.1: 15559529-15559529 on Build GRCh38
6202521 c.1122_1125delCTGC p.C375fs*2 Chr.1: 15559624-15559627 on Build GRCh38
3863189 c.1038G>A p.K346K Chr.1: 15559540-15559540 on Build GRCh38
6202555 c.1121_1122insAACT p.C375fs*21 Chr.1: 15559623-15559624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547558705 Chr.1: 15559272-15559272 on Build GRCh38
rs764517505 Chr.1: 15559690-15559690 on Build GRCh38
rs890056284 Chr.1: 15559323-15559323 on Build GRCh38
rs981961610 Chr.1: 15559465-15559465 on Build GRCh38
rs144974415 Chr.1: 15559448-15559448 on Build GRCh38
rs61750325 Chr.1: 15559579-15559579 on Build GRCh38
rs779958588 Chr.1: 15559631-15559631 on Build GRCh38
rs760283118 Chr.1: 15559670-15559670 on Build GRCh38
rs897211377 Chr.1: 15559297-15559297 on Build GRCh38
rs201914457 Chr.1: 15559545-15559545 on Build GRCh38
rs779040358 Chr.1: 15559641-15559641 on Build GRCh38
rs760819775 Chr.1: 15559353-15559353 on Build GRCh38
rs764328213 Chr.1: 15559555-15559555 on Build GRCh38
rs752124680 Chr.1: 15559702-15559702 on Build GRCh38
rs749861776 Chr.1: 15559653-15559654 on Build GRCh38
rs187373625 Chr.1: 15559378-15559378 on Build GRCh38
rs534068030 Chr.1: 15559383-15559383 on Build GRCh38
rs1041352888 Chr.1: 15559348-15559348 on Build GRCh38
rs377391712 Chr.1: 15559569-15559569 on Build GRCh38
rs769256769 Chr.1: 15559668-15559670 on Build GRCh38
rs897416669 Chr.1: 15559628-15559628 on Build GRCh38
rs948359590 Chr.1: 15559473-15559473 on Build GRCh38
rs970570220 Chr.1: 15559456-15559456 on Build GRCh38
rs757327581 Chr.1: 15559600-15559600 on Build GRCh38
rs148404423 Chr.1: 15559646-15559646 on Build GRCh38
rs761717982 Chr.1: 15559535-15559535 on Build GRCh38
rs767854848 Chr.1: 15559307-15559307 on Build GRCh38
rs909763264 Chr.1: 15559510-15559510 on Build GRCh38
rs914941570 Chr.1: 15559553-15559553 on Build GRCh38
rs769829795 Chr.1: 15559488-15559488 on Build GRCh38
rs762839879 Chr.1: 15559528-15559528 on Build GRCh38
rs746127431 Chr.1: 15559617-15559617 on Build GRCh38
rs753886753 Chr.1: 15559453-15559453 on Build GRCh38
rs774438587 Chr.1: 15559642-15559642 on Build GRCh38
rs980061314 Chr.1: 15559477-15559477 on Build GRCh38
rs1012359105 Chr.1: 15559411-15559411 on Build GRCh38
rs148785407 Chr.1: 15559392-15559392 on Build GRCh38
rs941204665 Chr.1: 15559519-15559519 on Build GRCh38
rs569629402 Chr.1: 15559319-15559319 on Build GRCh38
rs758787062 Chr.1: 15559568-15559568 on Build GRCh38
rs567382850 Chr.1: 15559283-15559283 on Build GRCh38
rs781387811 Chr.1: 15559591-15559591 on Build GRCh38
rs144343992 Chr.1: 15559300-15559300 on Build GRCh38
rs150027781 Chr.1: 15559298-15559298 on Build GRCh38
rs201645810 Chr.1: 15559643-15559643 on Build GRCh38
rs994575319 Chr.1: 15559362-15559362 on Build GRCh38
rs775478249 Chr.1: 15559522-15559522 on Build GRCh38
rs753893665 Chr.1: 15559597-15559597 on Build GRCh38
rs767214972 Chr.1: 15559537-15559537 on Build GRCh38
rs947550826 Chr.1: 15559296-15559296 on Build GRCh38
rs1037079032 Chr.1: 15559293-15559294 on Build GRCh38
rs1012280169 Chr.1: 15559706-15559706 on Build GRCh38
rs372775770 Chr.1: 15559619-15559619 on Build GRCh38
rs968228239 Chr.1: 15559374-15559374 on Build GRCh38
rs576810470 Chr.1: 15559640-15559640 on Build GRCh38
rs753235339 Chr.1: 15559688-15559688 on Build GRCh38
rs753031415 Chr.1: 15559334-15559334 on Build GRCh38
rs554323594 Chr.1: 15559384-15559384 on Build GRCh38
rs375931534 Chr.1: 15559623-15559623 on Build GRCh38
rs755634565 Chr.1: 15559551-15559551 on Build GRCh38
rs138617449 Chr.1: 15559556-15559556 on Build GRCh38
rs769692010 Chr.1: 15559374-15559375 on Build GRCh38
rs759059453 Chr.1: 15559689-15559689 on Build GRCh38
rs182007546 Chr.1: 15559330-15559330 on Build GRCh38
rs537802379 Chr.1: 15559322-15559322 on Build GRCh38
rs914993548 Chr.1: 15559292-15559292 on Build GRCh38
rs866850795 Chr.1: 15559637-15559637 on Build GRCh38
rs745892913 Chr.1: 15559480-15559480 on Build GRCh38
rs373355549 Chr.1: 15559529-15559529 on Build GRCh38
rs772896520 Chr.1: 15559647-15559647 on Build GRCh38
rs1029659901 Chr.1: 15559313-15559313 on Build GRCh38
rs755208026 Chr.1: 15559461-15559461 on Build GRCh38
rs757620971 Chr.1: 15559589-15559589 on Build GRCh38
rs911384227 Chr.1: 15559573-15559573 on Build GRCh38
rs766082605 Chr.1: 15559678-15559678 on Build GRCh38
rs149620751 Chr.1: 15559530-15559530 on Build GRCh38

View Full Product Details