Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15564093-15564492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC16
Entrez Gene ID
23341
Gene Name
DnaJ heat shock protein family (Hsp40) member C16
Gene Aliases
-
UniGene
Hs.655410
Gene Chromosome Location
Chr.1: 15526694-15571733 on Build GRCh38
Transcripts
NM_001287811.1 NM_015291.3 NR_109898.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4223730 c.1506C>T p.T502T Chr.1: 15564096-15564096 on Build GRCh38
6871460 c.1569C>A p.D523E Chr.1: 15564330-15564330 on Build GRCh38
5968904 c.1579A>G p.S527G Chr.1: 15564340-15564340 on Build GRCh38
4023881 c.1551T>C p.A517A Chr.1: 15564312-15564312 on Build GRCh38
1295305 c.1553C>G p.S518C Chr.1: 15564314-15564314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372999348 Chr.1: 15564134-15564134 on Build GRCh38
rs373558461 Chr.1: 15564149-15564149 on Build GRCh38
rs746548524 Chr.1: 15564345-15564345 on Build GRCh38
rs924432172 Chr.1: 15564384-15564384 on Build GRCh38
rs138963469 Chr.1: 15564343-15564343 on Build GRCh38
rs774505420 Chr.1: 15564243-15564243 on Build GRCh38
rs180930595 Chr.1: 15564381-15564381 on Build GRCh38
rs901782381 Chr.1: 15564374-15564374 on Build GRCh38
rs749662406 Chr.1: 15564372-15564372 on Build GRCh38
rs775955775 Chr.1: 15564154-15564154 on Build GRCh38
rs772315924 Chr.1: 15564398-15564398 on Build GRCh38
rs561865092 Chr.1: 15564399-15564399 on Build GRCh38
rs890862720 Chr.1: 15564201-15564201 on Build GRCh38
rs186230120 Chr.1: 15564389-15564389 on Build GRCh38
rs754972440 Chr.1: 15564412-15564413 on Build GRCh38
rs748411849 Chr.1: 15564397-15564397 on Build GRCh38
rs530972634 Chr.1: 15564413-15564413 on Build GRCh38
rs373388480 Chr.1: 15564233-15564233 on Build GRCh38
rs746424009 Chr.1: 15564144-15564144 on Build GRCh38
rs753607084 Chr.1: 15564296-15564296 on Build GRCh38
rs756780343 Chr.1: 15564306-15564306 on Build GRCh38
rs372276264 Chr.1: 15564358-15564358 on Build GRCh38
rs753474280 Chr.1: 15564242-15564242 on Build GRCh38
rs368247216 Chr.1: 15564283-15564283 on Build GRCh38
rs546177652 Chr.1: 15564280-15564280 on Build GRCh38
rs115273779 Chr.1: 15564226-15564226 on Build GRCh38
rs112221157 Chr.1: 15564262-15564262 on Build GRCh38
rs564753258 Chr.1: 15564462-15564462 on Build GRCh38
rs1004411132 Chr.1: 15564206-15564206 on Build GRCh38
rs765141971 Chr.1: 15564266-15564267 on Build GRCh38
rs752568297 Chr.1: 15564350-15564352 on Build GRCh38
rs1011380786 Chr.1: 15564416-15564416 on Build GRCh38
rs567284851 Chr.1: 15564305-15564305 on Build GRCh38
rs757958441 Chr.1: 15564340-15564340 on Build GRCh38
rs754606922 Chr.1: 15564118-15564118 on Build GRCh38
rs766232973 Chr.1: 15564405-15564405 on Build GRCh38
rs544409137 Chr.1: 15564436-15564436 on Build GRCh38
rs950160261 Chr.1: 15564365-15564365 on Build GRCh38
rs780430853 Chr.1: 15564363-15564363 on Build GRCh38
rs376881628 Chr.1: 15564121-15564121 on Build GRCh38
rs996530752 Chr.1: 15564158-15564158 on Build GRCh38
rs373868243 Chr.1: 15564120-15564120 on Build GRCh38
rs761941683 Chr.1: 15564246-15564246 on Build GRCh38
rs962193103 Chr.1: 15564238-15564238 on Build GRCh38
rs760428984 Chr.1: 15564138-15564138 on Build GRCh38
rs760453217 Chr.1: 15564175-15564175 on Build GRCh38
rs752348564 Chr.1: 15564333-15564333 on Build GRCh38
rs764935840 Chr.1: 15564319-15564319 on Build GRCh38
rs569045004 Chr.1: 15564122-15564122 on Build GRCh38
rs201786695 Chr.1: 15564266-15564266 on Build GRCh38
rs754732504 Chr.1: 15564300-15564300 on Build GRCh38
rs185568333 Chr.1: 15564162-15564162 on Build GRCh38
rs111776669 Chr.1: 15564408-15564408 on Build GRCh38
rs760753937 Chr.1: 15564403-15564403 on Build GRCh38
rs376253947 Chr.1: 15564254-15564254 on Build GRCh38

View Full Product Details