Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15633333-15633834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDI2
Entrez Gene ID
84301
Gene Name
DNA damage inducible 1 homolog 2
Gene Aliases
-
UniGene
Hs.718857
Gene Chromosome Location
Chr.1: 15617458-15661057 on Build GRCh38
Transcripts
NM_032341.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7143869 c.589C>G p.P197A Chr.1: 15633522-15633522 on Build GRCh38
3863224 c.590C>T p.P197L Chr.1: 15633523-15633523 on Build GRCh38
6888768 c.523C>T p.L175L Chr.1: 15633456-15633456 on Build GRCh38
6058742 c.517A>G p.R173G Chr.1: 15633450-15633450 on Build GRCh38
6641330 c.571C>T p.R191C Chr.1: 15633504-15633504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762035169 Chr.1: 15633792-15633792 on Build GRCh38
rs533542972 Chr.1: 15633657-15633657 on Build GRCh38
rs771993539 Chr.1: 15633409-15633409 on Build GRCh38
rs3795763 Chr.1: 15633482-15633482 on Build GRCh38
rs41270275 Chr.1: 15633805-15633805 on Build GRCh38
rs1051077664 Chr.1: 15633776-15633776 on Build GRCh38
rs978262057 Chr.1: 15633721-15633721 on Build GRCh38
rs776589135 Chr.1: 15633585-15633585 on Build GRCh38
rs763877509 Chr.1: 15633759-15633759 on Build GRCh38
rs891138735 Chr.1: 15633786-15633786 on Build GRCh38
rs777089239 Chr.1: 15633477-15633477 on Build GRCh38
rs773160744 Chr.1: 15633505-15633505 on Build GRCh38
rs765865868 Chr.1: 15633491-15633491 on Build GRCh38
rs756842831 Chr.1: 15633770-15633770 on Build GRCh38
rs532356503 Chr.1: 15633419-15633419 on Build GRCh38
rs760446474 Chr.1: 15633618-15633619 on Build GRCh38
rs996898503 Chr.1: 15633366-15633366 on Build GRCh38
rs3795762 Chr.1: 15633391-15633391 on Build GRCh38
rs773511522 Chr.1: 15633800-15633800 on Build GRCh38
rs748318737 Chr.1: 15633501-15633501 on Build GRCh38
rs749243084 Chr.1: 15633401-15633401 on Build GRCh38
rs995337282 Chr.1: 15633802-15633802 on Build GRCh38
rs768784608 Chr.1: 15633539-15633539 on Build GRCh38
rs541380916 Chr.1: 15633387-15633387 on Build GRCh38
rs1035978975 Chr.1: 15633729-15633729 on Build GRCh38
rs1022574015 Chr.1: 15633751-15633752 on Build GRCh38
rs148012684 Chr.1: 15633808-15633808 on Build GRCh38
rs552595753 Chr.1: 15633680-15633681 on Build GRCh38
rs758094417 Chr.1: 15633458-15633458 on Build GRCh38
rs369245119 Chr.1: 15633430-15633430 on Build GRCh38
rs753617568 Chr.1: 15633425-15633425 on Build GRCh38
rs773351875 Chr.1: 15633412-15633412 on Build GRCh38
rs770750343 Chr.1: 15633564-15633564 on Build GRCh38
rs769099374 Chr.1: 15633484-15633484 on Build GRCh38
rs189803561 Chr.1: 15633402-15633402 on Build GRCh38
rs182701846 Chr.1: 15633737-15633737 on Build GRCh38
rs78386550 Chr.1: 15633745-15633745 on Build GRCh38
rs749651325 Chr.1: 15633483-15633483 on Build GRCh38
rs1030143287 Chr.1: 15633372-15633372 on Build GRCh38
rs1042497054 Chr.1: 15633667-15633668 on Build GRCh38
rs760459097 Chr.1: 15633417-15633417 on Build GRCh38
rs749741205 Chr.1: 15633392-15633392 on Build GRCh38
rs945115101 Chr.1: 15633669-15633669 on Build GRCh38
rs895735738 Chr.1: 15633738-15633738 on Build GRCh38
rs917644203 Chr.1: 15633638-15633638 on Build GRCh38
rs773121698 Chr.1: 15633598-15633599 on Build GRCh38
rs774765639 Chr.1: 15633404-15633404 on Build GRCh38
rs756578455 Chr.1: 15633478-15633478 on Build GRCh38
rs922851617 Chr.1: 15633476-15633476 on Build GRCh38
rs762155354 Chr.1: 15633407-15633407 on Build GRCh38
rs764964935 Chr.1: 15633597-15633597 on Build GRCh38
rs530979717 Chr.1: 15633428-15633429 on Build GRCh38
rs200566001 Chr.1: 15633545-15633545 on Build GRCh38
rs752502711 Chr.1: 15633601-15633601 on Build GRCh38
rs372521898 Chr.1: 15633608-15633608 on Build GRCh38
rs535791063 Chr.1: 15633758-15633758 on Build GRCh38
rs772339357 Chr.1: 15633504-15633504 on Build GRCh38
rs759502820 Chr.1: 15633592-15633592 on Build GRCh38
rs991860646 Chr.1: 15633621-15633621 on Build GRCh38
rs751024321 Chr.1: 15633764-15633764 on Build GRCh38
rs199886079 Chr.1: 15633547-15633547 on Build GRCh38
rs772156794 Chr.1: 15633796-15633796 on Build GRCh38
rs551131914 Chr.1: 15633453-15633453 on Build GRCh38
rs769847151 Chr.1: 15633796-15633798 on Build GRCh38
rs764838063 Chr.1: 15633429-15633429 on Build GRCh38

View Full Product Details