Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16046428-16046918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCNKB
Entrez Gene ID
1188
Gene Name
chloride voltage-gated channel Kb
Gene Aliases
CLCKB, ClC-K2, ClC-Kb
UniGene
Hs.352243
Gene Chromosome Location
Chr.1: 16043736-16057326 on Build GRCh38
Transcripts
NM_000085.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3802676 c.287G>A p.W96* Chr.1: 16046592-16046592 on Build GRCh38
898849 c.292G>A p.V98M Chr.1: 16046597-16046597 on Build GRCh38
7106224 c.336G>A p.Q112Q Chr.1: 16046641-16046641 on Build GRCh38
6629938 c.296A>G p.Y99C Chr.1: 16046601-16046601 on Build GRCh38
3863378 c.306C>T p.A102A Chr.1: 16046611-16046611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552199269 Chr.1: 16046779-16046779 on Build GRCh38
rs768956168 Chr.1: 16046595-16046595 on Build GRCh38
rs764958294 Chr.1: 16046572-16046572 on Build GRCh38
rs2014841 Chr.1: 16046476-16046476 on Build GRCh38
rs957653019 Chr.1: 16046827-16046827 on Build GRCh38
rs772105967 Chr.1: 16046545-16046545 on Build GRCh38
rs554794449 Chr.1: 16046534-16046534 on Build GRCh38
rs550466653 Chr.1: 16046725-16046725 on Build GRCh38
rs756113217 Chr.1: 16046504-16046504 on Build GRCh38
rs770999043 Chr.1: 16046620-16046620 on Build GRCh38
rs13306250 Chr.1: 16046795-16046795 on Build GRCh38
rs561537219 Chr.1: 16046703-16046703 on Build GRCh38
rs201876924 Chr.1: 16046585-16046585 on Build GRCh38
rs758066023 Chr.1: 16046576-16046576 on Build GRCh38
rs181505214 Chr.1: 16046825-16046825 on Build GRCh38
rs201499472 Chr.1: 16046682-16046682 on Build GRCh38
rs373764706 Chr.1: 16046542-16046542 on Build GRCh38
rs752627060 Chr.1: 16046696-16046696 on Build GRCh38
rs35820664 Chr.1: 16046705-16046707 on Build GRCh38
rs143339401 Chr.1: 16046563-16046563 on Build GRCh38
rs5256 Chr.1: 16046567-16046567 on Build GRCh38
rs112236280 Chr.1: 16046751-16046752 on Build GRCh38
rs534860798 Chr.1: 16046529-16046529 on Build GRCh38
rs765097463 Chr.1: 16046682-16046685 on Build GRCh38
rs753667002 Chr.1: 16046508-16046508 on Build GRCh38
rs202202425 Chr.1: 16046559-16046559 on Build GRCh38
rs143215915 Chr.1: 16046536-16046536 on Build GRCh38
rs747102358 Chr.1: 16046707-16046707 on Build GRCh38
rs752352309 Chr.1: 16046575-16046575 on Build GRCh38
rs530206349 Chr.1: 16046720-16046720 on Build GRCh38
rs758728854 Chr.1: 16046688-16046688 on Build GRCh38
rs5257 Chr.1: 16046629-16046629 on Build GRCh38
rs758155386 Chr.1: 16046703-16046705 on Build GRCh38
rs199657323 Chr.1: 16046683-16046683 on Build GRCh38
rs1019198040 Chr.1: 16046746-16046746 on Build GRCh38
rs796899671 Chr.1: 16046724-16046724 on Build GRCh38
rs72506933 Chr.1: 16046706-16046708 on Build GRCh38
rs754684742 Chr.1: 16046517-16046517 on Build GRCh38
rs757789729 Chr.1: 16046522-16046522 on Build GRCh38
rs777305169 Chr.1: 16046579-16046579 on Build GRCh38
rs752506201 Chr.1: 16046650-16046650 on Build GRCh38
rs184681243 Chr.1: 16046842-16046842 on Build GRCh38
rs774519177 Chr.1: 16046490-16046490 on Build GRCh38
rs762535125 Chr.1: 16046655-16046655 on Build GRCh38
rs749663302 Chr.1: 16046587-16046587 on Build GRCh38
rs750440346 Chr.1: 16046503-16046503 on Build GRCh38
rs776654219 Chr.1: 16046628-16046628 on Build GRCh38
rs773390010 Chr.1: 16046548-16046548 on Build GRCh38
rs777325023 Chr.1: 16046525-16046525 on Build GRCh38
rs375546236 Chr.1: 16046707-16046709 on Build GRCh38
rs558720338 Chr.1: 16046506-16046506 on Build GRCh38
rs780664952 Chr.1: 16046679-16046679 on Build GRCh38
rs35530360 Chr.1: 16046615-16046615 on Build GRCh38
rs189669011 Chr.1: 16046808-16046808 on Build GRCh38
rs527773080 Chr.1: 16046643-16046643 on Build GRCh38
rs184305891 Chr.1: 16046475-16046475 on Build GRCh38
rs2095540 Chr.1: 16046787-16046787 on Build GRCh38
rs766373009 Chr.1: 16046560-16046560 on Build GRCh38
rs570238493 Chr.1: 16046729-16046729 on Build GRCh38
rs761729975 Chr.1: 16046493-16046493 on Build GRCh38
rs774375793 Chr.1: 16046537-16046537 on Build GRCh38
rs867091784 Chr.1: 16046749-16046750 on Build GRCh38
rs779191112 Chr.1: 16046684-16046684 on Build GRCh38
rs779327388 Chr.1: 16046600-16046600 on Build GRCh38
rs748354862 Chr.1: 16046609-16046609 on Build GRCh38
rs748501046 Chr.1: 16046687-16046687 on Build GRCh38
rs751020964 Chr.1: 16046580-16046580 on Build GRCh38
rs139178074 Chr.1: 16046875-16046875 on Build GRCh38
rs762384991 Chr.1: 16046704-16046705 on Build GRCh38
rs751072753 Chr.1: 16046673-16046673 on Build GRCh38
rs950996593 Chr.1: 16046552-16046552 on Build GRCh38
rs763695231 Chr.1: 16046661-16046661 on Build GRCh38
rs906487651 Chr.1: 16046695-16046695 on Build GRCh38
rs778077832 Chr.1: 16046694-16046694 on Build GRCh38
rs561475719 Chr.1: 16046788-16046788 on Build GRCh38
rs764173213 Chr.1: 16046486-16046486 on Build GRCh38
rs370343548 Chr.1: 16046521-16046521 on Build GRCh38
rs756697320 Chr.1: 16046677-16046677 on Build GRCh38
rs146627440 Chr.1: 16046535-16046535 on Build GRCh38
rs763211059 Chr.1: 16046485-16046485 on Build GRCh38
rs772320740 Chr.1: 16046614-16046614 on Build GRCh38
rs201091415 Chr.1: 16046502-16046502 on Build GRCh38
rs759619946 Chr.1: 16046595-16046596 on Build GRCh38
rs532173719 Chr.1: 16046738-16046738 on Build GRCh38

View Full Product Details