Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16048581-16049082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCNKB
Entrez Gene ID
1188
Gene Name
chloride voltage-gated channel Kb
Gene Aliases
CLCKB, ClC-K2, ClC-Kb
UniGene
Hs.352243
Gene Chromosome Location
Chr.1: 16043736-16057326 on Build GRCh38
Transcripts
NM_000085.4 NM_001165945.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6527511 c.67G>C p.E23Q Chr.1: 16049038-16049038 on Build GRCh38
5528726 c.81G>A p.W27* Chr.1: 16049052-16049052 on Build GRCh38
6629935 c.39T>C p.A13A Chr.1: 16049010-16049010 on Build GRCh38
7114483 c.65G>A p.R22K Chr.1: 16049036-16049036 on Build GRCh38
6382631 c.56G>T p.R19I Chr.1: 16049027-16049027 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs9633514 Chr.1: 16048715-16048715 on Build GRCh38
rs759136255 Chr.1: 16049059-16049059 on Build GRCh38
rs901284357 Chr.1: 16048684-16048684 on Build GRCh38
rs755859196 Chr.1: 16048627-16048627 on Build GRCh38
rs542184080 Chr.1: 16048742-16048742 on Build GRCh38
rs754631794 Chr.1: 16048986-16048986 on Build GRCh38
rs563794131 Chr.1: 16048630-16048651 on Build GRCh38
rs765316636 Chr.1: 16049050-16049051 on Build GRCh38
rs12118326 Chr.1: 16048637-16048637 on Build GRCh38
rs984660656 Chr.1: 16048638-16048638 on Build GRCh38
rs568263244 Chr.1: 16049021-16049021 on Build GRCh38
rs976930301 Chr.1: 16048853-16048853 on Build GRCh38
rs570996042 Chr.1: 16048952-16048952 on Build GRCh38
rs767468243 Chr.1: 16048623-16048623 on Build GRCh38
rs539900379 Chr.1: 16048817-16048817 on Build GRCh38
rs530836325 Chr.1: 16048928-16048928 on Build GRCh38
rs766129316 Chr.1: 16049048-16049048 on Build GRCh38
rs952393915 Chr.1: 16049041-16049041 on Build GRCh38
rs750243998 Chr.1: 16048973-16048973 on Build GRCh38
rs113883907 Chr.1: 16048948-16048948 on Build GRCh38
rs529057477 Chr.1: 16048909-16048909 on Build GRCh38
rs749637923 Chr.1: 16048932-16048932 on Build GRCh38
rs771279134 Chr.1: 16048610-16048610 on Build GRCh38
rs984387190 Chr.1: 16048791-16048791 on Build GRCh38
rs754507967 Chr.1: 16048929-16048929 on Build GRCh38
rs9442227 Chr.1: 16048859-16048859 on Build GRCh38
rs536033843 Chr.1: 16049017-16049017 on Build GRCh38
rs752155216 Chr.1: 16048989-16048989 on Build GRCh38
rs200548651 Chr.1: 16048734-16048734 on Build GRCh38
rs757792735 Chr.1: 16049002-16049002 on Build GRCh38
rs878987928 Chr.1: 16048728-16048728 on Build GRCh38
rs553616251 Chr.1: 16048647-16048647 on Build GRCh38
rs774466700 Chr.1: 16048969-16048969 on Build GRCh38
rs867329175 Chr.1: 16048602-16048602 on Build GRCh38
rs998669958 Chr.1: 16048738-16048738 on Build GRCh38
rs371785153 Chr.1: 16048966-16048966 on Build GRCh38
rs773338509 Chr.1: 16049045-16049045 on Build GRCh38
rs750190985 Chr.1: 16048624-16048624 on Build GRCh38
rs767517701 Chr.1: 16048972-16048972 on Build GRCh38
rs373871181 Chr.1: 16048752-16048752 on Build GRCh38
rs763231500 Chr.1: 16048963-16048963 on Build GRCh38
rs553477442 Chr.1: 16049012-16049012 on Build GRCh38
rs575166643 Chr.1: 16048757-16048757 on Build GRCh38
rs1050159789 Chr.1: 16048902-16048902 on Build GRCh38
rs959795929 Chr.1: 16048759-16048759 on Build GRCh38
rs552719787 Chr.1: 16048744-16048744 on Build GRCh38
rs779782013 Chr.1: 16048629-16048629 on Build GRCh38
rs1021997619 Chr.1: 16048717-16048717 on Build GRCh38
rs769027001 Chr.1: 16048933-16048933 on Build GRCh38
rs752208315 Chr.1: 16049061-16049061 on Build GRCh38
rs759707624 Chr.1: 16048979-16048980 on Build GRCh38
rs768873661 Chr.1: 16049019-16049019 on Build GRCh38
rs1022636929 Chr.1: 16048786-16048786 on Build GRCh38
rs932723861 Chr.1: 16048867-16048867 on Build GRCh38
rs751517658 Chr.1: 16048611-16048611 on Build GRCh38
rs908811104 Chr.1: 16048792-16048792 on Build GRCh38
rs569115410 Chr.1: 16048917-16048917 on Build GRCh38
rs180938125 Chr.1: 16048910-16048910 on Build GRCh38
rs543783756 Chr.1: 16048766-16048766 on Build GRCh38
rs761914199 Chr.1: 16049032-16049032 on Build GRCh38
rs539945007 Chr.1: 16048970-16048970 on Build GRCh38
rs778340257 Chr.1: 16048987-16048987 on Build GRCh38
rs555815017 Chr.1: 16049057-16049057 on Build GRCh38
rs761809836 Chr.1: 16048621-16048621 on Build GRCh38
rs537759207 Chr.1: 16049005-16049005 on Build GRCh38
rs746246914 Chr.1: 16049006-16049006 on Build GRCh38
rs940210840 Chr.1: 16048653-16048653 on Build GRCh38
rs11588392 Chr.1: 16049015-16049015 on Build GRCh38
rs781559822 Chr.1: 16048949-16048949 on Build GRCh38
rs532855049 Chr.1: 16048789-16048789 on Build GRCh38
rs868411596 Chr.1: 16048709-16048709 on Build GRCh38
rs766179347 Chr.1: 16048982-16048982 on Build GRCh38
rs747514773 Chr.1: 16048945-16048945 on Build GRCh38
rs903016365 Chr.1: 16048719-16048719 on Build GRCh38
rs1041130663 Chr.1: 16048682-16048682 on Build GRCh38
rs753503431 Chr.1: 16048985-16048985 on Build GRCh38
rs775517871 Chr.1: 16048954-16048954 on Build GRCh38
rs932608729 Chr.1: 16048691-16048691 on Build GRCh38
rs764904972 Chr.1: 16049060-16049060 on Build GRCh38
rs12059889 Chr.1: 16048767-16048767 on Build GRCh38
rs778485953 Chr.1: 16048934-16048934 on Build GRCh38
rs760485269 Chr.1: 16048979-16048979 on Build GRCh38
rs959682023 Chr.1: 16048616-16048616 on Build GRCh38
rs774322435 Chr.1: 16049022-16049022 on Build GRCh38

View Full Product Details