Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16050426-16050923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCNKB
Entrez Gene ID
1188
Gene Name
chloride voltage-gated channel Kb
Gene Aliases
CLCKB, ClC-K2, ClC-Kb
UniGene
Hs.352243
Gene Chromosome Location
Chr.1: 16043736-16057326 on Build GRCh38
Transcripts
NM_000085.4 NM_001165945.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6385010 c.1058C>T p.S353F Chr.1: 16050879-16050879 on Build GRCh38
4672663 c.474C>T p.Y158Y Chr.1: 16050528-16050528 on Build GRCh38
5804581 c.592T>A p.W198R Chr.1: 16050920-16050920 on Build GRCh38
3802678 c.572C>A p.S191* Chr.1: 16050900-16050900 on Build GRCh38
5083780 c.1029C>T p.S343S Chr.1: 16050576-16050576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372482510 Chr.1: 16050845-16050845 on Build GRCh38
rs756565605 Chr.1: 16050560-16050560 on Build GRCh38
rs569630570 Chr.1: 16050797-16050797 on Build GRCh38
rs535357685 Chr.1: 16050674-16050674 on Build GRCh38
rs779218071 Chr.1: 16050618-16050618 on Build GRCh38
rs1052669520 Chr.1: 16050514-16050514 on Build GRCh38
rs121909134 Chr.1: 16050593-16050593 on Build GRCh38
rs988333756 Chr.1: 16050755-16050755 on Build GRCh38
rs772885597 Chr.1: 16050516-16050516 on Build GRCh38
rs777685114 Chr.1: 16050629-16050629 on Build GRCh38
rs754542076 Chr.1: 16050477-16050477 on Build GRCh38
rs35745997 Chr.1: 16050644-16050644 on Build GRCh38
rs918198207 Chr.1: 16050760-16050761 on Build GRCh38
rs201097879 Chr.1: 16050505-16050505 on Build GRCh38
rs529744442 Chr.1: 16050706-16050706 on Build GRCh38
rs368997387 Chr.1: 16050807-16050807 on Build GRCh38
rs562688255 Chr.1: 16050585-16050585 on Build GRCh38
rs10927895 Chr.1: 16050458-16050458 on Build GRCh38
rs769400472 Chr.1: 16050589-16050589 on Build GRCh38
rs752956789 Chr.1: 16050620-16050620 on Build GRCh38
rs775187299 Chr.1: 16050828-16050828 on Build GRCh38
rs763504960 Chr.1: 16050609-16050609 on Build GRCh38
rs746917976 Chr.1: 16050583-16050583 on Build GRCh38
rs538743005 Chr.1: 16050811-16050811 on Build GRCh38
rs755255246 Chr.1: 16050616-16050616 on Build GRCh38
rs140705060 Chr.1: 16050901-16050901 on Build GRCh38
rs185451420 Chr.1: 16050868-16050868 on Build GRCh38
rs201357287 Chr.1: 16050508-16050508 on Build GRCh38
rs756300742 Chr.1: 16050492-16050492 on Build GRCh38
rs754158660 Chr.1: 16050575-16050575 on Build GRCh38
rs949556066 Chr.1: 16050763-16050763 on Build GRCh38
rs368104316 Chr.1: 16050540-16050540 on Build GRCh38
rs935036505 Chr.1: 16050737-16050737 on Build GRCh38
rs74057567 Chr.1: 16050670-16050670 on Build GRCh38
rs201245211 Chr.1: 16050892-16050892 on Build GRCh38
rs768258386 Chr.1: 16050837-16050837 on Build GRCh38
rs751558214 Chr.1: 16050883-16050883 on Build GRCh38
rs888026091 Chr.1: 16050826-16050826 on Build GRCh38
rs781053314 Chr.1: 16050899-16050899 on Build GRCh38
rs752100309 Chr.1: 16050484-16050484 on Build GRCh38
rs911001307 Chr.1: 16050449-16050449 on Build GRCh38
rs749463748 Chr.1: 16050501-16050501 on Build GRCh38
rs765987918 Chr.1: 16050470-16050470 on Build GRCh38
rs35480349 Chr.1: 16050613-16050613 on Build GRCh38
rs758571826 Chr.1: 16050623-16050623 on Build GRCh38
rs371276597 Chr.1: 16050531-16050531 on Build GRCh38
rs547416967 Chr.1: 16050682-16050682 on Build GRCh38
rs372390371 Chr.1: 16050759-16050759 on Build GRCh38
rs527849197 Chr.1: 16050660-16050660 on Build GRCh38
rs776413329 Chr.1: 16050532-16050532 on Build GRCh38
rs774550737 Chr.1: 16050506-16050506 on Build GRCh38
rs764723252 Chr.1: 16050481-16050481 on Build GRCh38
rs764780584 Chr.1: 16050539-16050539 on Build GRCh38
rs761059666 Chr.1: 16050611-16050611 on Build GRCh38
rs777826819 Chr.1: 16050580-16050580 on Build GRCh38
rs200117310 Chr.1: 16050467-16050467 on Build GRCh38
rs368794099 Chr.1: 16050758-16050758 on Build GRCh38
rs781435713 Chr.1: 16050490-16050490 on Build GRCh38
rs777146902 Chr.1: 16050851-16050851 on Build GRCh38
rs201486977 Chr.1: 16050485-16050485 on Build GRCh38
rs572225114 Chr.1: 16050464-16050464 on Build GRCh38
rs1045254484 Chr.1: 16050766-16050766 on Build GRCh38
rs925487065 Chr.1: 16050495-16050495 on Build GRCh38
rs565896323 Chr.1: 16050846-16050846 on Build GRCh38
rs747015849 Chr.1: 16050631-16050631 on Build GRCh38
rs761275599 Chr.1: 16050847-16050847 on Build GRCh38
rs368504008 Chr.1: 16050598-16050598 on Build GRCh38
rs897827928 Chr.1: 16050806-16050806 on Build GRCh38
rs762559176 Chr.1: 16050831-16050831 on Build GRCh38
rs375061718 Chr.1: 16050656-16050656 on Build GRCh38
rs112933556 Chr.1: 16050789-16050789 on Build GRCh38
rs1008435493 Chr.1: 16050840-16050840 on Build GRCh38
rs374911075 Chr.1: 16050867-16050867 on Build GRCh38
rs754214144 Chr.1: 16050615-16050615 on Build GRCh38
rs373877250 Chr.1: 16050533-16050533 on Build GRCh38
rs748171371 Chr.1: 16050577-16050577 on Build GRCh38
rs780378358 Chr.1: 16050499-16050499 on Build GRCh38
rs748118246 Chr.1: 16050507-16050507 on Build GRCh38
rs567844969 Chr.1: 16050696-16050696 on Build GRCh38
rs770728253 Chr.1: 16050639-16050639 on Build GRCh38
rs371548242 Chr.1: 16050553-16050553 on Build GRCh38
rs765694706 Chr.1: 16050860-16050860 on Build GRCh38
rs747388630 Chr.1: 16050615-16050616 on Build GRCh38
rs375017657 Chr.1: 16050579-16050579 on Build GRCh38
rs764256848 Chr.1: 16050875-16050875 on Build GRCh38
rs942137627 Chr.1: 16050796-16050796 on Build GRCh38
rs757796536 Chr.1: 16050543-16050543 on Build GRCh38
rs549793997 Chr.1: 16050736-16050736 on Build GRCh38
rs766728474 Chr.1: 16050850-16050850 on Build GRCh38
rs147338414 Chr.1: 16050576-16050576 on Build GRCh38
rs769451791 Chr.1: 16050825-16050825 on Build GRCh38
rs753303041 Chr.1: 16050472-16050472 on Build GRCh38
rs569936954 Chr.1: 16050812-16050812 on Build GRCh38
rs5251 Chr.1: 16050547-16050547 on Build GRCh38
rs150740382 Chr.1: 16050520-16050520 on Build GRCh38
rs780246914 Chr.1: 16050571-16050571 on Build GRCh38
rs770800260 Chr.1: 16050584-16050584 on Build GRCh38
rs760033582 Chr.1: 16050852-16050852 on Build GRCh38
rs745599423 Chr.1: 16050646-16050646 on Build GRCh38
rs759212842 Chr.1: 16050586-16050586 on Build GRCh38

View Full Product Details