Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16367006-16367508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SZRD1
Entrez Gene ID
26099
Gene Name
SUZ RNA binding domain containing 1
Gene Aliases
C1orf144
UniGene
Hs.252967
Gene Chromosome Location
Chr.1: 16367030-16398148 on Build GRCh38
Transcripts
NM_001114600.2 NM_001271869.1 NR_073500.1 NR_073501.1 NR_073502.1 NR_073503.1

SNP Detail

SNP ID Chromosome Location
rs115326907 Chr.1: 16367042-16367042 on Build GRCh38
rs17551926 Chr.1: 16367235-16367235 on Build GRCh38
rs879037823 Chr.1: 16367230-16367230 on Build GRCh38
rs1035867860 Chr.1: 16367398-16367398 on Build GRCh38
rs755626121 Chr.1: 16367275-16367275 on Build GRCh38
rs1041490519 Chr.1: 16367479-16367479 on Build GRCh38
rs916280578 Chr.1: 16367174-16367174 on Build GRCh38
rs982905302 Chr.1: 16367383-16367383 on Build GRCh38
rs945879941 Chr.1: 16367463-16367463 on Build GRCh38
rs754691807 Chr.1: 16367469-16367469 on Build GRCh38
rs1036370587 Chr.1: 16367136-16367136 on Build GRCh38
rs572864283 Chr.1: 16367231-16367231 on Build GRCh38
rs780071456 Chr.1: 16367353-16367353 on Build GRCh38
rs903326191 Chr.1: 16367339-16367339 on Build GRCh38
rs767138163 Chr.1: 16367209-16367209 on Build GRCh38
rs969105830 Chr.1: 16367175-16367175 on Build GRCh38
rs72904127 Chr.1: 16367109-16367109 on Build GRCh38
rs748993850 Chr.1: 16367321-16367321 on Build GRCh38
rs746045736 Chr.1: 16367324-16367324 on Build GRCh38
rs990459779 Chr.1: 16367450-16367450 on Build GRCh38
rs35380209 Chr.1: 16367040-16367040 on Build GRCh38
rs375741964 Chr.1: 16367302-16367302 on Build GRCh38
rs754459073 Chr.1: 16367439-16367439 on Build GRCh38
rs769788715 Chr.1: 16367341-16367341 on Build GRCh38
rs991792783 Chr.1: 16367165-16367165 on Build GRCh38
rs747301008 Chr.1: 16367231-16367231 on Build GRCh38
rs750021715 Chr.1: 16367223-16367223 on Build GRCh38
rs1009970226 Chr.1: 16367247-16367247 on Build GRCh38
rs386628913 Chr.1: 16367316-16367317 on Build GRCh38
rs1012114690 Chr.1: 16367097-16367097 on Build GRCh38
rs567369521 Chr.1: 16367425-16367425 on Build GRCh38
rs909019402 Chr.1: 16367187-16367187 on Build GRCh38
rs764764633 Chr.1: 16367443-16367443 on Build GRCh38
rs757636810 Chr.1: 16367281-16367281 on Build GRCh38
rs1041440326 Chr.1: 16367273-16367273 on Build GRCh38
rs920067530 Chr.1: 16367170-16367170 on Build GRCh38
rs760113510 Chr.1: 16367224-16367224 on Build GRCh38
rs974272838 Chr.1: 16367163-16367163 on Build GRCh38
rs1027186604 Chr.1: 16367331-16367331 on Build GRCh38
rs1050415745 Chr.1: 16367253-16367253 on Build GRCh38
rs372595789 Chr.1: 16367271-16367271 on Build GRCh38
rs781212243 Chr.1: 16367234-16367235 on Build GRCh38
rs1013158202 Chr.1: 16367452-16367452 on Build GRCh38
rs142845497 Chr.1: 16367230-16367231 on Build GRCh38
rs892941647 Chr.1: 16367235-16367235 on Build GRCh38
rs759012023 Chr.1: 16367248-16367248 on Build GRCh38
rs1002495089 Chr.1: 16367293-16367293 on Build GRCh38
rs997521956 Chr.1: 16367036-16367036 on Build GRCh38
rs931941973 Chr.1: 16367428-16367428 on Build GRCh38
rs901822674 Chr.1: 16367033-16367033 on Build GRCh38
rs999430457 Chr.1: 16367384-16367384 on Build GRCh38
rs996135855 Chr.1: 16367317-16367317 on Build GRCh38
rs977672802 Chr.1: 16367128-16367128 on Build GRCh38
rs959180642 Chr.1: 16367156-16367156 on Build GRCh38
rs969476010 Chr.1: 16367455-16367455 on Build GRCh38
rs1023236251 Chr.1: 16367453-16367453 on Build GRCh38
rs575179908 Chr.1: 16367395-16367395 on Build GRCh38
rs960322728 Chr.1: 16367137-16367137 on Build GRCh38
rs984968250 Chr.1: 16367178-16367178 on Build GRCh38
rs967978467 Chr.1: 16367119-16367119 on Build GRCh38
rs762363231 Chr.1: 16367188-16367188 on Build GRCh38
rs1043547404 Chr.1: 16367320-16367320 on Build GRCh38
rs889196904 Chr.1: 16367066-16367066 on Build GRCh38
rs1036244829 Chr.1: 16367194-16367194 on Build GRCh38
rs894737908 Chr.1: 16367312-16367312 on Build GRCh38
rs932836220 Chr.1: 16367196-16367213 on Build GRCh38
rs984535209 Chr.1: 16367482-16367482 on Build GRCh38

View Full Product Details