Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16921849-16922345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROCC
Entrez Gene ID
9696
Gene Name
ciliary rootlet coiled-coil, rootletin
Gene Aliases
ROLT
UniGene
Hs.309403
Gene Chromosome Location
Chr.1: 16914082-16972979 on Build GRCh38
Transcripts
NM_014675.4 XM_006711058.3 XM_011542468.1 XM_011542470.2 XM_017002913.1 XM_017002914.1 XM_017002915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4912973 c.5G>A p.S2N Chr.1: 16922023-16922023 on Build GRCh38
4542536 c.31G>A p.V11M Chr.1: 16922049-16922049 on Build GRCh38
7322761 c.18G>A p.A6A Chr.1: 16922036-16922036 on Build GRCh38
7362735 c.27G>T p.Q9H Chr.1: 16922045-16922045 on Build GRCh38
6563612 c.41C>T p.T14I Chr.1: 16922059-16922059 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs193268849 Chr.1: 16922123-16922123 on Build GRCh38
rs373018626 Chr.1: 16922155-16922155 on Build GRCh38
rs74227514 Chr.1: 16921888-16921888 on Build GRCh38
rs551490314 Chr.1: 16922179-16922179 on Build GRCh38
rs770865682 Chr.1: 16922018-16922018 on Build GRCh38
rs565026275 Chr.1: 16922182-16922182 on Build GRCh38
rs940675904 Chr.1: 16922194-16922194 on Build GRCh38
rs752307032 Chr.1: 16922046-16922046 on Build GRCh38
rs959883975 Chr.1: 16921920-16921920 on Build GRCh38
rs527746856 Chr.1: 16922188-16922188 on Build GRCh38
rs769543740 Chr.1: 16921932-16921932 on Build GRCh38
rs760872290 Chr.1: 16922031-16922031 on Build GRCh38
rs745891830 Chr.1: 16922021-16922021 on Build GRCh38
rs560830205 Chr.1: 16922068-16922068 on Build GRCh38
rs972168381 Chr.1: 16922256-16922256 on Build GRCh38
rs998647699 Chr.1: 16921903-16921903 on Build GRCh38
rs34370934 Chr.1: 16921888-16921888 on Build GRCh38
rs745803786 Chr.1: 16922086-16922086 on Build GRCh38
rs760964506 Chr.1: 16921997-16921997 on Build GRCh38
rs558681376 Chr.1: 16922019-16922019 on Build GRCh38
rs1035388486 Chr.1: 16921907-16921907 on Build GRCh38
rs576521358 Chr.1: 16922004-16922004 on Build GRCh38
rs184446252 Chr.1: 16921985-16921985 on Build GRCh38
rs755763548 Chr.1: 16922053-16922053 on Build GRCh38
rs201470726 Chr.1: 16922011-16922012 on Build GRCh38
rs772144277 Chr.1: 16922024-16922024 on Build GRCh38
rs375594926 Chr.1: 16922115-16922115 on Build GRCh38
rs767129032 Chr.1: 16922043-16922043 on Build GRCh38
rs775614983 Chr.1: 16922026-16922026 on Build GRCh38
rs984444214 Chr.1: 16922048-16922048 on Build GRCh38
rs11203422 Chr.1: 16922302-16922302 on Build GRCh38
rs768972632 Chr.1: 16922257-16922257 on Build GRCh38
rs572098800 Chr.1: 16922036-16922036 on Build GRCh38
rs567503711 Chr.1: 16922289-16922289 on Build GRCh38
rs749187204 Chr.1: 16922066-16922066 on Build GRCh38
rs112869817 Chr.1: 16921893-16921893 on Build GRCh38
rs769920686 Chr.1: 16921900-16921900 on Build GRCh38
rs369775795 Chr.1: 16921994-16921994 on Build GRCh38
rs6586566 Chr.1: 16922037-16922037 on Build GRCh38
rs888672427 Chr.1: 16922238-16922238 on Build GRCh38
rs530021629 Chr.1: 16922095-16922095 on Build GRCh38
rs902845374 Chr.1: 16921875-16921875 on Build GRCh38
rs780931244 Chr.1: 16922014-16922014 on Build GRCh38
rs777325939 Chr.1: 16922054-16922054 on Build GRCh38
rs570276740 Chr.1: 16922305-16922305 on Build GRCh38
rs757017220 Chr.1: 16922070-16922070 on Build GRCh38
rs59062737 Chr.1: 16921892-16921893 on Build GRCh38
rs1022845168 Chr.1: 16922022-16922022 on Build GRCh38
rs554081684 Chr.1: 16921873-16921873 on Build GRCh38
rs115619445 Chr.1: 16922295-16922295 on Build GRCh38
rs547409672 Chr.1: 16922209-16922209 on Build GRCh38
rs761789281 Chr.1: 16922122-16922122 on Build GRCh38
rs778756821 Chr.1: 16922078-16922078 on Build GRCh38
rs192333233 Chr.1: 16921935-16921935 on Build GRCh38
rs747897295 Chr.1: 16922088-16922088 on Build GRCh38
rs368239934 Chr.1: 16922035-16922035 on Build GRCh38
rs923325582 Chr.1: 16922262-16922262 on Build GRCh38
rs991255202 Chr.1: 16921970-16921970 on Build GRCh38
rs764514062 Chr.1: 16922001-16922001 on Build GRCh38
rs6683035 Chr.1: 16922005-16922005 on Build GRCh38
rs773275943 Chr.1: 16922247-16922247 on Build GRCh38
rs778769958 Chr.1: 16922012-16922012 on Build GRCh38
rs753797239 Chr.1: 16921986-16921987 on Build GRCh38
rs754469347 Chr.1: 16922042-16922042 on Build GRCh38
rs754211720 Chr.1: 16922002-16922002 on Build GRCh38
rs190520832 Chr.1: 16922016-16922016 on Build GRCh38
rs754553604 Chr.1: 16922013-16922013 on Build GRCh38
rs772053922 Chr.1: 16922102-16922102 on Build GRCh38
rs372566108 Chr.1: 16922112-16922112 on Build GRCh38

View Full Product Details