Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16929778-16930280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROCC
Entrez Gene ID
9696
Gene Name
ciliary rootlet coiled-coil, rootletin
Gene Aliases
ROLT
UniGene
Hs.309403
Gene Chromosome Location
Chr.1: 16914082-16972979 on Build GRCh38
Transcripts
NM_014675.4 XM_006711058.3 XM_011542468.1 XM_011542470.2 XM_017002913.1 XM_017002914.1 XM_017002915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
463429 c.614G>A p.R205Q Chr.1: 16930200-16930200 on Build GRCh38
6281492 c.523C>T p.R175W Chr.1: 16930017-16930017 on Build GRCh38
1336294 c.504G>A p.R168R Chr.1: 16929998-16929998 on Build GRCh38
319677 c.613C>T p.R205W Chr.1: 16930199-16930199 on Build GRCh38
4143033 c.375G>A p.E125E Chr.1: 16929869-16929869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371693254 Chr.1: 16930169-16930169 on Build GRCh38
rs748463836 Chr.1: 16930010-16930010 on Build GRCh38
rs376189880 Chr.1: 16929997-16929997 on Build GRCh38
rs768153372 Chr.1: 16930112-16930112 on Build GRCh38
rs371339969 Chr.1: 16930036-16930036 on Build GRCh38
rs763939335 Chr.1: 16929952-16929954 on Build GRCh38
rs764157364 Chr.1: 16930227-16930227 on Build GRCh38
rs761524383 Chr.1: 16930054-16930054 on Build GRCh38
rs749210063 Chr.1: 16929847-16929847 on Build GRCh38
rs772052541 Chr.1: 16929928-16929928 on Build GRCh38
rs757060672 Chr.1: 16929982-16929982 on Build GRCh38
rs746441825 Chr.1: 16930104-16930104 on Build GRCh38
rs777692189 Chr.1: 16929819-16929819 on Build GRCh38
rs752777079 Chr.1: 16930220-16930220 on Build GRCh38
rs779310710 Chr.1: 16930246-16930246 on Build GRCh38
rs574287994 Chr.1: 16930223-16930223 on Build GRCh38
rs112455398 Chr.1: 16929917-16929917 on Build GRCh38
rs149095491 Chr.1: 16930150-16930150 on Build GRCh38
rs567840594 Chr.1: 16930135-16930135 on Build GRCh38
rs760213919 Chr.1: 16929961-16929961 on Build GRCh38
rs767766123 Chr.1: 16929833-16929833 on Build GRCh38
rs771296501 Chr.1: 16930088-16930088 on Build GRCh38
rs777101709 Chr.1: 16929938-16929938 on Build GRCh38
rs530869723 Chr.1: 16930082-16930082 on Build GRCh38
rs988039505 Chr.1: 16929988-16929988 on Build GRCh38
rs758235052 Chr.1: 16930058-16930058 on Build GRCh38
rs773399438 Chr.1: 16929909-16929910 on Build GRCh38
rs375998499 Chr.1: 16929890-16929890 on Build GRCh38
rs752300095 Chr.1: 16929960-16929960 on Build GRCh38
rs775591348 Chr.1: 16929886-16929886 on Build GRCh38
rs542701023 Chr.1: 16930052-16930052 on Build GRCh38
rs140045815 Chr.1: 16930200-16930200 on Build GRCh38
rs72644305 Chr.1: 16929911-16929911 on Build GRCh38
rs573658825 Chr.1: 16929834-16929834 on Build GRCh38
rs781587289 Chr.1: 16930007-16930007 on Build GRCh38
rs760799047 Chr.1: 16929940-16929942 on Build GRCh38
rs780737471 Chr.1: 16929808-16929808 on Build GRCh38
rs759138411 Chr.1: 16929910-16929910 on Build GRCh38
rs111787883 Chr.1: 16930181-16930181 on Build GRCh38
rs765000550 Chr.1: 16930055-16930055 on Build GRCh38
rs369720716 Chr.1: 16929887-16929887 on Build GRCh38
rs773868044 Chr.1: 16929942-16929942 on Build GRCh38
rs770905379 Chr.1: 16929858-16929858 on Build GRCh38
rs753585132 Chr.1: 16929971-16929971 on Build GRCh38
rs751542308 Chr.1: 16930066-16930066 on Build GRCh38
rs183343569 Chr.1: 16930113-16930113 on Build GRCh38
rs755098704 Chr.1: 16930067-16930067 on Build GRCh38
rs1018450259 Chr.1: 16929852-16929852 on Build GRCh38
rs750789276 Chr.1: 16930251-16930251 on Build GRCh38
rs763623893 Chr.1: 16929944-16929944 on Build GRCh38
rs772235900 Chr.1: 16929880-16929880 on Build GRCh38
rs767595366 Chr.1: 16930219-16930219 on Build GRCh38
rs781497430 Chr.1: 16930079-16930079 on Build GRCh38
rs766367358 Chr.1: 16930060-16930060 on Build GRCh38
rs762384312 Chr.1: 16929899-16929899 on Build GRCh38
rs767683184 Chr.1: 16930139-16930139 on Build GRCh38
rs556236891 Chr.1: 16929909-16929909 on Build GRCh38
rs747183546 Chr.1: 16929935-16929935 on Build GRCh38
rs373731769 Chr.1: 16929901-16929901 on Build GRCh38
rs373961830 Chr.1: 16930120-16930120 on Build GRCh38
rs113287485 Chr.1: 16930006-16930006 on Build GRCh38
rs554315283 Chr.1: 16929828-16929828 on Build GRCh38
rs963792605 Chr.1: 16929893-16929893 on Build GRCh38
rs147890555 Chr.1: 16929964-16929964 on Build GRCh38
rs529355496 Chr.1: 16930022-16930022 on Build GRCh38
rs116462658 Chr.1: 16930097-16930097 on Build GRCh38
rs753789622 Chr.1: 16930162-16930167 on Build GRCh38
rs930106220 Chr.1: 16930161-16930161 on Build GRCh38
rs374418441 Chr.1: 16930221-16930221 on Build GRCh38
rs762585948 Chr.1: 16930186-16930186 on Build GRCh38
rs1025883835 Chr.1: 16929800-16929800 on Build GRCh38
rs747926367 Chr.1: 16929813-16929813 on Build GRCh38
rs762827216 Chr.1: 16930127-16930127 on Build GRCh38
rs763598204 Chr.1: 16930046-16930046 on Build GRCh38
rs768967686 Chr.1: 16929937-16929937 on Build GRCh38
rs115882423 Chr.1: 16930131-16930131 on Build GRCh38
rs752864946 Chr.1: 16930141-16930141 on Build GRCh38
rs774824069 Chr.1: 16930038-16930038 on Build GRCh38
rs777106541 Chr.1: 16929897-16929897 on Build GRCh38
rs9435710 Chr.1: 16929818-16929818 on Build GRCh38
rs369256326 Chr.1: 16929918-16929918 on Build GRCh38
rs760257698 Chr.1: 16930043-16930043 on Build GRCh38
rs981432233 Chr.1: 16930072-16930072 on Build GRCh38
rs370828942 Chr.1: 16930115-16930115 on Build GRCh38
rs756366820 Chr.1: 16930142-16930142 on Build GRCh38
rs374116934 Chr.1: 16930256-16930256 on Build GRCh38
rs750263860 Chr.1: 16930057-16930057 on Build GRCh38
rs572252498 Chr.1: 16930199-16930199 on Build GRCh38
rs757482405 Chr.1: 16930232-16930232 on Build GRCh38
rs201015613 Chr.1: 16930017-16930017 on Build GRCh38
rs745982861 Chr.1: 16929869-16929869 on Build GRCh38
rs749695435 Chr.1: 16930084-16930084 on Build GRCh38
rs545352105 Chr.1: 16929927-16929927 on Build GRCh38
rs750352189 Chr.1: 16929992-16929992 on Build GRCh38
rs367916126 Chr.1: 16930149-16930149 on Build GRCh38
rs754953271 Chr.1: 16930226-16930245 on Build GRCh38
rs778165437 Chr.1: 16930144-16930144 on Build GRCh38
rs747685540 Chr.1: 16930176-16930176 on Build GRCh38
rs192079216 Chr.1: 16930083-16930083 on Build GRCh38
rs773772066 Chr.1: 16930018-16930018 on Build GRCh38
rs772840380 Chr.1: 16930125-16930125 on Build GRCh38
rs754936246 Chr.1: 16930137-16930137 on Build GRCh38
rs766925565 Chr.1: 16929959-16929959 on Build GRCh38
rs753674078 Chr.1: 16929914-16929914 on Build GRCh38
rs558553781 Chr.1: 16929931-16929931 on Build GRCh38
rs373118266 Chr.1: 16929996-16929996 on Build GRCh38
rs769384312 Chr.1: 16930179-16930179 on Build GRCh38
rs536313354 Chr.1: 16929862-16929862 on Build GRCh38
rs536400276 Chr.1: 16930146-16930146 on Build GRCh38
rs202022351 Chr.1: 16930064-16930064 on Build GRCh38
rs12409496 Chr.1: 16929875-16929875 on Build GRCh38
rs572267346 Chr.1: 16929989-16929989 on Build GRCh38
rs755612877 Chr.1: 16930230-16930230 on Build GRCh38
rs780644769 Chr.1: 16930175-16930175 on Build GRCh38
rs770755010 Chr.1: 16930190-16930190 on Build GRCh38

View Full Product Details